Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18753409-18753670 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRTC1
Entrez Gene ID
23373
Gene Name
CREB regulated transcription coactivator 1
Gene Aliases
MECT1, TORC-1, TORC1, WAMTP1
UniGene
Hs.371096
Gene Chromosome Location
Chr.19: 18683615-18782333 on Build GRCh38
Transcripts
NM_001098482.1 NM_015321.2 XM_005259833.3 XM_005259834.1 XM_005259835.3 XM_005259836.3 XM_006722710.3 XM_011527842.2 XM_017026536.1 XM_017026537.1 XM_017026538.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1589852 c.618G>T p.E206D Chr.19: 18753531-18753531 on Build GRCh38
3680822 c.539-1G>C p.? Chr.19: 18753499-18753499 on Build GRCh38
3680820 c.539-2A>C p.? Chr.19: 18753498-18753498 on Build GRCh38
3680821 c.587-1G>C p.? Chr.19: 18753499-18753499 on Build GRCh38
7182090 c.570G>C p.E190D Chr.19: 18753531-18753531 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754549459 Chr.19: 18753623-18753623 on Build GRCh38
rs139391552 Chr.19: 18753525-18753525 on Build GRCh38
rs750579512 Chr.19: 18753446-18753446 on Build GRCh38
rs150056965 Chr.19: 18753551-18753551 on Build GRCh38
rs767007423 Chr.19: 18753465-18753465 on Build GRCh38
rs374705037 Chr.19: 18753442-18753442 on Build GRCh38
rs772925998 Chr.19: 18753612-18753612 on Build GRCh38
rs750119863 Chr.19: 18753479-18753479 on Build GRCh38
rs970596133 Chr.19: 18753549-18753549 on Build GRCh38
rs933576341 Chr.19: 18753438-18753438 on Build GRCh38
rs56300365 Chr.19: 18753562-18753562 on Build GRCh38
rs748466051 Chr.19: 18753514-18753514 on Build GRCh38
rs140269231 Chr.19: 18753490-18753490 on Build GRCh38
rs761606316 Chr.19: 18753453-18753453 on Build GRCh38
rs768521639 Chr.19: 18753564-18753564 on Build GRCh38
rs1012270364 Chr.19: 18753637-18753637 on Build GRCh38
rs747279199 Chr.19: 18753526-18753526 on Build GRCh38
rs762163960 Chr.19: 18753433-18753433 on Build GRCh38
rs571740767 Chr.19: 18753431-18753431 on Build GRCh38
rs1011856482 Chr.19: 18753536-18753536 on Build GRCh38
rs748500891 Chr.19: 18753619-18753619 on Build GRCh38
rs774272041 Chr.19: 18753581-18753581 on Build GRCh38
rs146700853 Chr.19: 18753449-18753449 on Build GRCh38
rs755542420 Chr.19: 18753500-18753500 on Build GRCh38
rs779653079 Chr.19: 18753476-18753478 on Build GRCh38
rs753331585 Chr.19: 18753491-18753491 on Build GRCh38
rs376781799 Chr.19: 18753485-18753485 on Build GRCh38
rs772569346 Chr.19: 18753516-18753516 on Build GRCh38
rs1023272533 Chr.19: 18753548-18753548 on Build GRCh38
rs766014696 Chr.19: 18753614-18753614 on Build GRCh38
rs781316204 Chr.19: 18753487-18753491 on Build GRCh38
rs868758733 Chr.19: 18753568-18753568 on Build GRCh38
rs956740303 Chr.19: 18753565-18753565 on Build GRCh38
rs767604546 Chr.19: 18753445-18753445 on Build GRCh38
rs567859889 Chr.19: 18753584-18753584 on Build GRCh38
rs779624303 Chr.19: 18753512-18753512 on Build GRCh38
rs1030801722 Chr.19: 18753555-18753555 on Build GRCh38
rs771097876 Chr.19: 18753545-18753545 on Build GRCh38
rs767364621 Chr.19: 18753610-18753610 on Build GRCh38
rs753389674 Chr.19: 18753621-18753621 on Build GRCh38

View Full Product Details