Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 36293571-36293838 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTHD1
Entrez Gene ID
401124
Gene Name
death domain containing 1
Gene Aliases
-
UniGene
Hs.363407
Gene Chromosome Location
Chr.4: 36281593-36347511 on Build GRCh38
Transcripts
NM_001136536.4 NM_001170700.2 XM_006714014.3 XM_011513693.2 XM_011513694.2 XM_011513695.2 XM_011513696.2 XM_017008191.1 XM_017008192.1 XM_017008193.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5000526 c.491C>A p.P164Q Chr.4: 36293668-36293668 on Build GRCh38
6047224 c.444C>T p.L148L Chr.4: 36293621-36293621 on Build GRCh38
1429499 c.964C>T p.R322* Chr.4: 36293646-36293646 on Build GRCh38
2845807 c.476C>A p.S159Y Chr.4: 36293653-36293653 on Build GRCh38
5680815 c.445G>C p.A149P Chr.4: 36293622-36293622 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368662128 Chr.4: 36293621-36293621 on Build GRCh38
rs370087938 Chr.4: 36293654-36293654 on Build GRCh38
rs928276724 Chr.4: 36293700-36293700 on Build GRCh38
rs938332725 Chr.4: 36293730-36293730 on Build GRCh38
rs908756205 Chr.4: 36293752-36293752 on Build GRCh38
rs780475018 Chr.4: 36293673-36293673 on Build GRCh38
rs542623785 Chr.4: 36293653-36293653 on Build GRCh38
rs1056021890 Chr.4: 36293617-36293617 on Build GRCh38
rs756535211 Chr.4: 36293668-36293668 on Build GRCh38
rs1020004462 Chr.4: 36293737-36293737 on Build GRCh38
rs182788481 Chr.4: 36293630-36293630 on Build GRCh38
rs754906230 Chr.4: 36293613-36293613 on Build GRCh38
rs530655283 Chr.4: 36293765-36293765 on Build GRCh38
rs970203447 Chr.4: 36293646-36293646 on Build GRCh38
rs1056752494 Chr.4: 36293735-36293735 on Build GRCh38
rs902856619 Chr.4: 36293763-36293763 on Build GRCh38
rs560693384 Chr.4: 36293664-36293664 on Build GRCh38
rs749810345 Chr.4: 36293755-36293755 on Build GRCh38
rs374158435 Chr.4: 36293618-36293618 on Build GRCh38
rs979831227 Chr.4: 36293647-36293647 on Build GRCh38
rs1011326370 Chr.4: 36293722-36293722 on Build GRCh38
rs778821651 Chr.4: 36293740-36293740 on Build GRCh38
rs1007870239 Chr.4: 36293798-36293798 on Build GRCh38
rs1038541711 Chr.4: 36293799-36293799 on Build GRCh38
rs372924892 Chr.4: 36293638-36293638 on Build GRCh38

View Full Product Details