Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 145680013-145680286 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4orf51
Entrez Gene ID
646603
Gene Name
chromosome 4 open reading frame 51
Gene Aliases
-
UniGene
Hs.452865
Gene Chromosome Location
Chr.4: 145680103-145792248 on Build GRCh38
Transcripts
NM_001080531.1 XM_006714289.3 XM_011532196.2 XM_011532197.2 XM_017008547.1 XM_017008548.1 XM_017008549.1 XM_017008550.1 XM_017008551.1 XM_017008552.1 XM_017008553.1 XM_017008554.1 XM_017008555.1 XM_017008556.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6327789 c.48C>T p.S16S Chr.4: 145680251-145680251 on Build GRCh38
5400567 c.83G>A p.R28H Chr.4: 145680286-145680286 on Build GRCh38
3825289 c.78C>A p.I26I Chr.4: 145680281-145680281 on Build GRCh38
5512073 c.12_14delCTT p.F5delF Chr.4: 145680215-145680217 on Build GRCh38
392513 c.52_55delCTTA p.T19fs*6 Chr.4: 145680255-145680258 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs181753963 Chr.4: 145680194-145680194 on Build GRCh38
rs774005719 Chr.4: 145680169-145680169 on Build GRCh38
rs749345999 Chr.4: 145680150-145680150 on Build GRCh38
rs199511104 Chr.4: 145680253-145680253 on Build GRCh38
rs758990313 Chr.4: 145680237-145680237 on Build GRCh38
rs1001920802 Chr.4: 145680174-145680174 on Build GRCh38
rs554039688 Chr.4: 145680260-145680260 on Build GRCh38
rs751687229 Chr.4: 145680256-145680256 on Build GRCh38
rs1047015015 Chr.4: 145680046-145680046 on Build GRCh38
rs771874649 Chr.4: 145680193-145680193 on Build GRCh38
rs1000881472 Chr.4: 145680225-145680225 on Build GRCh38
rs17020528 Chr.4: 145680197-145680197 on Build GRCh38
rs753429430 Chr.4: 145680221-145680221 on Build GRCh38
rs974940111 Chr.4: 145680241-145680241 on Build GRCh38
rs770796417 Chr.4: 145680162-145680162 on Build GRCh38
rs149073990 Chr.4: 145680228-145680228 on Build GRCh38
rs898412430 Chr.4: 145680097-145680097 on Build GRCh38
rs79903178 Chr.4: 145680086-145680086 on Build GRCh38
rs937886955 Chr.4: 145680099-145680099 on Build GRCh38
rs878915636 Chr.4: 145680211-145680211 on Build GRCh38
rs1021079385 Chr.4: 145680195-145680195 on Build GRCh38
rs763768595 Chr.4: 145680215-145680215 on Build GRCh38
rs1046112629 Chr.4: 145680108-145680108 on Build GRCh38
rs750721719 Chr.4: 145680233-145680233 on Build GRCh38
rs200707934 Chr.4: 145680198-145680198 on Build GRCh38
rs1054978039 Chr.4: 145680113-145680113 on Build GRCh38
rs367595571 Chr.4: 145680148-145680148 on Build GRCh38
rs373003529 Chr.4: 145680189-145680189 on Build GRCh38
rs1030528667 Chr.4: 145680243-145680243 on Build GRCh38
rs145104316 Chr.4: 145680118-145680118 on Build GRCh38
rs761442015 Chr.4: 145680227-145680227 on Build GRCh38

View Full Product Details