Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 195756196-195756421 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH7
Entrez Gene ID
56171
Gene Name
dynein axonemal heavy chain 7
Gene Aliases
-
UniGene
Hs.97403
Gene Chromosome Location
Chr.2: 195737604-196075188 on Build GRCh38
Transcripts
NM_018897.2 XM_011511487.2 XM_011511488.2 XM_011511489.2 XM_011511490.2 XM_011511497.2 XM_017004504.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4909887 c.11474G>A p.S3825N Chr.2: 195756245-195756245 on Build GRCh38
1014267 c.11470G>T p.V3824F Chr.2: 195756249-195756249 on Build GRCh38
1136548 c.11462A>C p.E3821A Chr.2: 195756257-195756257 on Build GRCh38
4903413 c.11434-1G>T p.? Chr.2: 195756286-195756286 on Build GRCh38
6646724 c.11449T>C p.S3817P Chr.2: 195756270-195756270 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756222394 Chr.2: 195756282-195756282 on Build GRCh38
rs754161080 Chr.2: 195756286-195756286 on Build GRCh38
rs199727636 Chr.2: 195756242-195756242 on Build GRCh38
rs748981089 Chr.2: 195756308-195756312 on Build GRCh38
rs912868412 Chr.2: 195756258-195756258 on Build GRCh38
rs556936934 Chr.2: 195756225-195756225 on Build GRCh38
rs944882735 Chr.2: 195756366-195756366 on Build GRCh38
rs986524185 Chr.2: 195756337-195756337 on Build GRCh38
rs372890943 Chr.2: 195756312-195756312 on Build GRCh38
rs761703315 Chr.2: 195756256-195756256 on Build GRCh38
rs758770710 Chr.2: 195756316-195756316 on Build GRCh38
rs751465302 Chr.2: 195756253-195756253 on Build GRCh38
rs755361830 Chr.2: 195756296-195756296 on Build GRCh38
rs779485073 Chr.2: 195756301-195756301 on Build GRCh38
rs148592943 Chr.2: 195756274-195756274 on Build GRCh38
rs541762433 Chr.2: 195756283-195756283 on Build GRCh38
rs989829084 Chr.2: 195756237-195756237 on Build GRCh38
rs778344883 Chr.2: 195756320-195756320 on Build GRCh38
rs945709041 Chr.2: 195756303-195756303 on Build GRCh38
rs368618634 Chr.2: 195756266-195756266 on Build GRCh38
rs80100699 Chr.2: 195756294-195756294 on Build GRCh38
rs745326510 Chr.2: 195756328-195756328 on Build GRCh38
rs545611794 Chr.2: 195756357-195756357 on Build GRCh38
rs558353410 Chr.2: 195756387-195756387 on Build GRCh38
rs767511688 Chr.2: 195756267-195756267 on Build GRCh38

View Full Product Details