Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 46799055-46799320 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4B1
Entrez Gene ID
1580
Gene Name
cytochrome P450 family 4 subfamily B member 1
Gene Aliases
CYPIVB1, P-450HP
UniGene
Hs.436317
Gene Chromosome Location
Chr.1: 46798995-46819349 on Build GRCh38
Transcripts
NM_000779.3 NM_001099772.1 NM_001319161.1 NM_001319162.1 NM_001319163.1 NR_135003.1 XM_011540832.1 XM_017000466.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5381145 c.174C>T p.A58A Chr.1: 46799255-46799255 on Build GRCh38
5949352 c.172G>T p.A58S Chr.1: 46799253-46799253 on Build GRCh38
5958204 c.10A>C p.S4R Chr.1: 46799091-46799091 on Build GRCh38
280273 c.151A>G p.T51A Chr.1: 46799232-46799232 on Build GRCh38
5521617 c.40G>A p.G14S Chr.1: 46799121-46799121 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773616192 Chr.1: 46799149-46799149 on Build GRCh38
rs746767776 Chr.1: 46799089-46799089 on Build GRCh38
rs755998147 Chr.1: 46799283-46799283 on Build GRCh38
rs377260959 Chr.1: 46799274-46799274 on Build GRCh38
rs779867012 Chr.1: 46799242-46799242 on Build GRCh38
rs553628563 Chr.1: 46799185-46799185 on Build GRCh38
rs111476033 Chr.1: 46799094-46799094 on Build GRCh38
rs954723152 Chr.1: 46799276-46799276 on Build GRCh38
rs751445615 Chr.1: 46799266-46799266 on Build GRCh38
rs200131677 Chr.1: 46799157-46799157 on Build GRCh38
rs773563294 Chr.1: 46799239-46799239 on Build GRCh38
rs45522235 Chr.1: 46799228-46799228 on Build GRCh38
rs45518538 Chr.1: 46799275-46799275 on Build GRCh38
rs958474459 Chr.1: 46799208-46799208 on Build GRCh38
rs760128967 Chr.1: 46799257-46799257 on Build GRCh38
rs954937628 Chr.1: 46799259-46799259 on Build GRCh38
rs761033950 Chr.1: 46799243-46799243 on Build GRCh38
rs745774872 Chr.1: 46799102-46799102 on Build GRCh38
rs777301820 Chr.1: 46799082-46799082 on Build GRCh38
rs758388131 Chr.1: 46799177-46799177 on Build GRCh38
rs766821883 Chr.1: 46799248-46799248 on Build GRCh38
rs762201149 Chr.1: 46799126-46799126 on Build GRCh38
rs910660201 Chr.1: 46799294-46799294 on Build GRCh38
rs751134907 Chr.1: 46799261-46799261 on Build GRCh38
rs754904125 Chr.1: 46799278-46799279 on Build GRCh38
rs555992529 Chr.1: 46799148-46799148 on Build GRCh38
rs769906289 Chr.1: 46799105-46799105 on Build GRCh38
rs34257324 Chr.1: 46799287-46799288 on Build GRCh38
rs752554919 Chr.1: 46799176-46799176 on Build GRCh38
rs745721776 Chr.1: 46799215-46799215 on Build GRCh38
rs761158779 Chr.1: 46799153-46799153 on Build GRCh38
rs781117115 Chr.1: 46799207-46799207 on Build GRCh38
rs367935135 Chr.1: 46799161-46799161 on Build GRCh38
rs1038525522 Chr.1: 46799227-46799227 on Build GRCh38
rs894906576 Chr.1: 46799195-46799195 on Build GRCh38
rs768653507 Chr.1: 46799233-46799233 on Build GRCh38
rs774463909 Chr.1: 46799122-46799122 on Build GRCh38
rs756979595 Chr.1: 46799196-46799196 on Build GRCh38
rs745623064 Chr.1: 46799116-46799116 on Build GRCh38
rs766522522 Chr.1: 46799174-46799174 on Build GRCh38
rs55642000 Chr.1: 46799258-46799258 on Build GRCh38
rs756045166 Chr.1: 46799222-46799222 on Build GRCh38
rs749329877 Chr.1: 46799114-46799114 on Build GRCh38
rs748297613 Chr.1: 46799234-46799234 on Build GRCh38
rs183371507 Chr.1: 46799154-46799154 on Build GRCh38
rs55672106 Chr.1: 46799184-46799184 on Build GRCh38
rs113351559 Chr.1: 46799093-46799093 on Build GRCh38
rs772338414 Chr.1: 46799236-46799236 on Build GRCh38
rs915276757 Chr.1: 46799085-46799085 on Build GRCh38
rs775704371 Chr.1: 46799113-46799113 on Build GRCh38
rs752143159 Chr.1: 46799155-46799155 on Build GRCh38
rs768867681 Chr.1: 46799120-46799120 on Build GRCh38
rs781172722 Chr.1: 46799097-46799097 on Build GRCh38
rs576666017 Chr.1: 46799220-46799220 on Build GRCh38
rs780156530 Chr.1: 46799223-46799223 on Build GRCh38
rs780103150 Chr.1: 46799293-46799293 on Build GRCh38
rs1013006446 Chr.1: 46799226-46799226 on Build GRCh38
rs1017692216 Chr.1: 46799284-46799284 on Build GRCh38
rs777033858 Chr.1: 46799251-46799251 on Build GRCh38

View Full Product Details