Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 75207145-75207370 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C15orf39
Entrez Gene ID
56905
Gene Name
chromosome 15 open reading frame 39
Gene Aliases
-
UniGene
Hs.17936
Gene Chromosome Location
Chr.15: 75201880-75212169 on Build GRCh38
Transcripts
NM_015492.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5156774 c.1320C>A p.T440T Chr.15: 75207368-75207368 on Build GRCh38
3734002 c.1231_1232delGG p.G411fs*45 Chr.15: 75207279-75207280 on Build GRCh38
6614399 c.1178G>A p.G393E Chr.15: 75207226-75207226 on Build GRCh38
6614398 c.1178G>A p.G393E Chr.15: 75207226-75207226 on Build GRCh38
5092513 c.1127_1128delTG p.L376fs*8 Chr.15: 75207175-75207176 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs182425183 Chr.15: 75207166-75207166 on Build GRCh38
rs761068022 Chr.15: 75207211-75207211 on Build GRCh38
rs199773722 Chr.15: 75207203-75207203 on Build GRCh38
rs532905421 Chr.15: 75207192-75207192 on Build GRCh38
rs769726152 Chr.15: 75207170-75207170 on Build GRCh38
rs762102303 Chr.15: 75207195-75207195 on Build GRCh38
rs530211941 Chr.15: 75207259-75207259 on Build GRCh38
rs375288255 Chr.15: 75207232-75207232 on Build GRCh38
rs970462896 Chr.15: 75207289-75207289 on Build GRCh38
rs951823939 Chr.15: 75207169-75207169 on Build GRCh38
rs768063957 Chr.15: 75207202-75207202 on Build GRCh38
rs780949284 Chr.15: 75207163-75207163 on Build GRCh38
rs772440778 Chr.15: 75207348-75207348 on Build GRCh38
rs754260564 Chr.15: 75207227-75207227 on Build GRCh38
rs746853077 Chr.15: 75207172-75207172 on Build GRCh38
rs997598155 Chr.15: 75207233-75207233 on Build GRCh38
rs146132044 Chr.15: 75207201-75207201 on Build GRCh38
rs916251928 Chr.15: 75207288-75207288 on Build GRCh38
rs139440190 Chr.15: 75207279-75207279 on Build GRCh38
rs774670548 Chr.15: 75207193-75207193 on Build GRCh38
rs537106521 Chr.15: 75207283-75207283 on Build GRCh38
rs757968007 Chr.15: 75207257-75207257 on Build GRCh38
rs769525322 Chr.15: 75207290-75207290 on Build GRCh38
rs564824944 Chr.15: 75207171-75207171 on Build GRCh38
rs990858823 Chr.15: 75207287-75207287 on Build GRCh38
rs191225583 Chr.15: 75207320-75207320 on Build GRCh38
rs969974963 Chr.15: 75207222-75207222 on Build GRCh38
rs868051066 Chr.15: 75207183-75207183 on Build GRCh38
rs11072531 Chr.15: 75207278-75207278 on Build GRCh38
rs780068598 Chr.15: 75207293-75207293 on Build GRCh38
rs12591506 Chr.15: 75207261-75207261 on Build GRCh38
rs745675466 Chr.15: 75207280-75207280 on Build GRCh38
rs149666825 Chr.15: 75207313-75207313 on Build GRCh38
rs1030116295 Chr.15: 75207244-75207244 on Build GRCh38
rs187815876 Chr.15: 75207210-75207210 on Build GRCh38
rs768826871 Chr.15: 75207314-75207314 on Build GRCh38
rs981766430 Chr.15: 75207240-75207240 on Build GRCh38
rs370447315 Chr.15: 75207301-75207301 on Build GRCh38
rs766931567 Chr.15: 75207213-75207213 on Build GRCh38
rs748175526 Chr.15: 75207342-75207342 on Build GRCh38
rs1052978932 Chr.15: 75207300-75207300 on Build GRCh38
rs769290532 Chr.15: 75207325-75207325 on Build GRCh38
rs759475579 Chr.15: 75207175-75207176 on Build GRCh38

View Full Product Details