Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 72916913-72917182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH1A1
Entrez Gene ID
216
Gene Name
aldehyde dehydrogenase 1 family member A1
Gene Aliases
ALDC, ALDH-E1, ALDH1, ALDH11, HEL-9, HEL-S-53e, HEL12, PUMB1, RALDH1
UniGene
Hs.76392
Gene Chromosome Location
Chr.9: 72900662-72953317 on Build GRCh38
Transcripts
NM_000689.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
73635 c.903G>A p.Q301Q Chr.9: 72917052-72917052 on Build GRCh38
1109995 c.961C>T p.R321* Chr.9: 72916994-72916994 on Build GRCh38
4394131 c.1030C>T p.P344S Chr.9: 72916925-72916925 on Build GRCh38
212077 c.881G>T p.G294V Chr.9: 72917074-72917074 on Build GRCh38
1109997 c.958G>A p.A320T Chr.9: 72917039-72917039 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757052269 Chr.9: 72917059-72917059 on Build GRCh38
rs772971855 Chr.9: 72917108-72917108 on Build GRCh38
rs1017668690 Chr.9: 72916963-72916963 on Build GRCh38
rs150455539 Chr.9: 72917076-72917076 on Build GRCh38
rs575056227 Chr.9: 72916978-72916978 on Build GRCh38
rs199745432 Chr.9: 72917094-72917094 on Build GRCh38
rs755094671 Chr.9: 72917130-72917130 on Build GRCh38
rs753670410 Chr.9: 72916937-72916937 on Build GRCh38
rs973936519 Chr.9: 72916988-72916988 on Build GRCh38
rs201706478 Chr.9: 72917151-72917151 on Build GRCh38
rs781731756 Chr.9: 72916993-72916993 on Build GRCh38
rs756675348 Chr.9: 72917141-72917141 on Build GRCh38
rs765078284 Chr.9: 72917103-72917103 on Build GRCh38
rs879030995 Chr.9: 72917106-72917106 on Build GRCh38
rs761738889 Chr.9: 72917100-72917100 on Build GRCh38
rs761610314 Chr.9: 72916965-72916965 on Build GRCh38
rs929327267 Chr.9: 72917034-72917034 on Build GRCh38
rs372219595 Chr.9: 72917026-72917026 on Build GRCh38
rs544303576 Chr.9: 72917040-72917040 on Build GRCh38
rs560871509 Chr.9: 72917137-72917137 on Build GRCh38
rs764950451 Chr.9: 72916969-72916969 on Build GRCh38
rs140513949 Chr.9: 72917046-72917046 on Build GRCh38
rs774967243 Chr.9: 72917072-72917072 on Build GRCh38
rs1020724997 Chr.9: 72916968-72916968 on Build GRCh38
rs751679589 Chr.9: 72917119-72917119 on Build GRCh38
rs1000735238 Chr.9: 72917093-72917093 on Build GRCh38
rs780044537 Chr.9: 72916979-72916979 on Build GRCh38
rs186527997 Chr.9: 72916972-72916972 on Build GRCh38
rs754968883 Chr.9: 72916989-72916989 on Build GRCh38
rs930459311 Chr.9: 72917074-72917074 on Build GRCh38
rs748607020 Chr.9: 72917020-72917020 on Build GRCh38
rs778276187 Chr.9: 72917039-72917039 on Build GRCh38
rs939153598 Chr.9: 72917097-72917097 on Build GRCh38
rs192586760 Chr.9: 72917132-72917132 on Build GRCh38
rs751623980 Chr.9: 72916986-72916986 on Build GRCh38
rs151269980 Chr.9: 72917018-72917018 on Build GRCh38
rs367577799 Chr.9: 72917118-72917118 on Build GRCh38
rs781034975 Chr.9: 72917131-72917148 on Build GRCh38
rs770285747 Chr.9: 72917022-72917022 on Build GRCh38
rs373948969 Chr.9: 72917142-72917142 on Build GRCh38
rs959328512 Chr.9: 72917099-72917099 on Build GRCh38
rs879059866 Chr.9: 72917105-72917105 on Build GRCh38
rs201857856 Chr.9: 72917109-72917109 on Build GRCh38
rs767993204 Chr.9: 72917077-72917077 on Build GRCh38

View Full Product Details