Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75001322-75001583 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDR2L
Entrez Gene ID
30850
Gene Name
cerebellar degeneration related protein 2 like
Gene Aliases
HUMPPA
UniGene
Hs.78358
Gene Chromosome Location
Chr.17: 74987632-75005800 on Build GRCh38
Transcripts
NM_014603.2 XM_006721852.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4671260 c.220C>T p.R74W Chr.17: 75001368-75001368 on Build GRCh38
4139810 c.275G>A p.R92Q Chr.17: 75001423-75001423 on Build GRCh38
6998869 c.221G>A p.R74Q Chr.17: 75001369-75001369 on Build GRCh38
6625779 c.226G>A p.V76M Chr.17: 75001374-75001374 on Build GRCh38
7144062 c.225C>T p.H75H Chr.17: 75001373-75001373 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1049093037 Chr.17: 75001537-75001537 on Build GRCh38
rs754477852 Chr.17: 75001526-75001526 on Build GRCh38
rs768544611 Chr.17: 75001355-75001355 on Build GRCh38
rs758799501 Chr.17: 75001521-75001521 on Build GRCh38
rs776372131 Chr.17: 75001454-75001454 on Build GRCh38
rs767689602 Chr.17: 75001488-75001488 on Build GRCh38
rs765581928 Chr.17: 75001517-75001517 on Build GRCh38
rs779409722 Chr.17: 75001430-75001430 on Build GRCh38
rs757570066 Chr.17: 75001403-75001403 on Build GRCh38
rs562790204 Chr.17: 75001511-75001511 on Build GRCh38
rs187641873 Chr.17: 75001549-75001549 on Build GRCh38
rs747967290 Chr.17: 75001467-75001467 on Build GRCh38
rs377006883 Chr.17: 75001367-75001367 on Build GRCh38
rs779407474 Chr.17: 75001404-75001404 on Build GRCh38
rs3765123 Chr.17: 75001553-75001553 on Build GRCh38
rs929398316 Chr.17: 75001534-75001534 on Build GRCh38
rs750996964 Chr.17: 75001518-75001518 on Build GRCh38
rs768453865 Chr.17: 75001436-75001436 on Build GRCh38
rs748195274 Chr.17: 75001506-75001506 on Build GRCh38
rs759123705 Chr.17: 75001539-75001539 on Build GRCh38
rs760771582 Chr.17: 75001494-75001494 on Build GRCh38
rs201693484 Chr.17: 75001363-75001363 on Build GRCh38
rs746714610 Chr.17: 75001433-75001433 on Build GRCh38
rs754235026 Chr.17: 75001389-75001389 on Build GRCh38
rs1014230705 Chr.17: 75001552-75001552 on Build GRCh38
rs545040544 Chr.17: 75001510-75001511 on Build GRCh38
rs780570234 Chr.17: 75001522-75001522 on Build GRCh38
rs746637864 Chr.17: 75001344-75001344 on Build GRCh38
rs147493425 Chr.17: 75001364-75001364 on Build GRCh38
rs953692555 Chr.17: 75001508-75001508 on Build GRCh38
rs762170939 Chr.17: 75001512-75001512 on Build GRCh38
rs770376578 Chr.17: 75001499-75001499 on Build GRCh38
rs761658844 Chr.17: 75001358-75001358 on Build GRCh38
rs541884053 Chr.17: 75001545-75001545 on Build GRCh38
rs572905728 Chr.17: 75001543-75001543 on Build GRCh38
rs1038549449 Chr.17: 75001560-75001560 on Build GRCh38
rs182923091 Chr.17: 75001531-75001531 on Build GRCh38
rs116611906 Chr.17: 75001562-75001562 on Build GRCh38
rs902644309 Chr.17: 75001556-75001556 on Build GRCh38
rs552991161 Chr.17: 75001538-75001538 on Build GRCh38
rs540291010 Chr.17: 75001476-75001476 on Build GRCh38
rs1055812460 Chr.17: 75001544-75001544 on Build GRCh38
rs775405116 Chr.17: 75001489-75001489 on Build GRCh38
rs557174385 Chr.17: 75001487-75001487 on Build GRCh38
rs371537062 Chr.17: 75001369-75001369 on Build GRCh38
rs773264530 Chr.17: 75001486-75001486 on Build GRCh38
rs368590177 Chr.17: 75001423-75001423 on Build GRCh38
rs924428499 Chr.17: 75001399-75001399 on Build GRCh38
rs764486501 Chr.17: 75001380-75001380 on Build GRCh38
rs367710498 Chr.17: 75001368-75001368 on Build GRCh38
rs776535452 Chr.17: 75001356-75001356 on Build GRCh38
rs776279725 Chr.17: 75001500-75001500 on Build GRCh38
rs746294692 Chr.17: 75001422-75001422 on Build GRCh38
rs191845112 Chr.17: 75001525-75001525 on Build GRCh38
rs769643628 Chr.17: 75001472-75001472 on Build GRCh38
rs756271868 Chr.17: 75001374-75001374 on Build GRCh38
rs60758921 Chr.17: 75001516-75001517 on Build GRCh38

View Full Product Details