Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 71942464-71942695 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PKD1L3
Entrez Gene ID
342372
Gene Name
polycystin 1 like 3, transient receptor potential channel interacting
Gene Aliases
-
UniGene
Hs.675286
Gene Chromosome Location
Chr.16: 71929538-72000401 on Build GRCh38
Transcripts
NM_181536.1 XM_017023201.1

SNP Detail

SNP ID Chromosome Location
rs1043023277 Chr.16: 71942614-71942614 on Build GRCh38
rs911705114 Chr.16: 71942555-71942555 on Build GRCh38
rs779980775 Chr.16: 71942513-71942513 on Build GRCh38
rs535377595 Chr.16: 71942661-71942661 on Build GRCh38
rs768524934 Chr.16: 71942641-71942641 on Build GRCh38
rs373251864 Chr.16: 71942596-71942596 on Build GRCh38
rs950429174 Chr.16: 71942508-71942508 on Build GRCh38
rs368635205 Chr.16: 71942632-71942632 on Build GRCh38
rs988356765 Chr.16: 71942525-71942525 on Build GRCh38
rs756819965 Chr.16: 71942527-71942527 on Build GRCh38
rs780514544 Chr.16: 71942543-71942543 on Build GRCh38
rs746905823 Chr.16: 71942515-71942515 on Build GRCh38
rs780037590 Chr.16: 71942609-71942609 on Build GRCh38
rs779026000 Chr.16: 71942656-71942656 on Build GRCh38
rs895932981 Chr.16: 71942665-71942665 on Build GRCh38
rs752009809 Chr.16: 71942516-71942516 on Build GRCh38
rs753910934 Chr.16: 71942603-71942603 on Build GRCh38
rs924740296 Chr.16: 71942626-71942626 on Build GRCh38
rs1045276136 Chr.16: 71942548-71942548 on Build GRCh38
rs750152507 Chr.16: 71942581-71942581 on Build GRCh38
rs375182382 Chr.16: 71942608-71942608 on Build GRCh38
rs192266774 Chr.16: 71942526-71942526 on Build GRCh38
rs765710486 Chr.16: 71942652-71942652 on Build GRCh38
rs144038021 Chr.16: 71942551-71942551 on Build GRCh38

View Full Product Details