Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 86028481-86028754 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLR1A
Entrez Gene ID
25885
Gene Name
RNA polymerase I subunit A
Gene Aliases
A190, AFDCIN, RPA1, RPA194, RPO1-4, RPO14
UniGene
Hs.531818
Gene Chromosome Location
Chr.2: 86026328-86106155 on Build GRCh38
Transcripts
NM_015425.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7420757 c.4837G>A p.G1613S Chr.2: 86028654-86028654 on Build GRCh38
6752235 c.4832C>T p.T1611M Chr.2: 86028659-86028659 on Build GRCh38
6443229 c.4808A>G p.N1603S Chr.2: 86028683-86028683 on Build GRCh38
1221490 c.4792C>T p.R1598C Chr.2: 86028699-86028699 on Build GRCh38
7190782 c.4843G>A p.E1615K Chr.2: 86028648-86028648 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763548965 Chr.2: 86028658-86028658 on Build GRCh38
rs769640833 Chr.2: 86028544-86028544 on Build GRCh38
rs188480389 Chr.2: 86028601-86028601 on Build GRCh38
rs778229105 Chr.2: 86028659-86028659 on Build GRCh38
rs748890003 Chr.2: 86028681-86028681 on Build GRCh38
rs368651607 Chr.2: 86028572-86028572 on Build GRCh38
rs753148901 Chr.2: 86028646-86028646 on Build GRCh38
rs762034368 Chr.2: 86028535-86028535 on Build GRCh38
rs763205466 Chr.2: 86028721-86028721 on Build GRCh38
rs747429086 Chr.2: 86028583-86028583 on Build GRCh38
rs537596117 Chr.2: 86028532-86028532 on Build GRCh38
rs775872023 Chr.2: 86028717-86028717 on Build GRCh38
rs988093964 Chr.2: 86028587-86028587 on Build GRCh38
rs142931486 Chr.2: 86028531-86028531 on Build GRCh38
rs766483569 Chr.2: 86028641-86028641 on Build GRCh38
rs183821257 Chr.2: 86028600-86028600 on Build GRCh38
rs759654510 Chr.2: 86028699-86028699 on Build GRCh38
rs766347826 Chr.2: 86028565-86028565 on Build GRCh38
rs181416565 Chr.2: 86028643-86028643 on Build GRCh38
rs763969242 Chr.2: 86028557-86028557 on Build GRCh38
rs775197336 Chr.2: 86028550-86028550 on Build GRCh38
rs201364942 Chr.2: 86028711-86028711 on Build GRCh38
rs368879644 Chr.2: 86028695-86028695 on Build GRCh38
rs759019818 Chr.2: 86028577-86028577 on Build GRCh38
rs780504459 Chr.2: 86028595-86028595 on Build GRCh38
rs776817775 Chr.2: 86028698-86028698 on Build GRCh38
rs560258003 Chr.2: 86028682-86028682 on Build GRCh38
rs778103646 Chr.2: 86028513-86028513 on Build GRCh38
rs773948595 Chr.2: 86028691-86028691 on Build GRCh38
rs558615565 Chr.2: 86028624-86028624 on Build GRCh38
rs780815472 Chr.2: 86028671-86028671 on Build GRCh38
rs35093541 Chr.2: 86028667-86028667 on Build GRCh38
rs760849405 Chr.2: 86028564-86028564 on Build GRCh38
rs770491584 Chr.2: 86028586-86028586 on Build GRCh38
rs1014643528 Chr.2: 86028520-86028520 on Build GRCh38
rs752114103 Chr.2: 86028666-86028666 on Build GRCh38
rs201345115 Chr.2: 86028672-86028672 on Build GRCh38
rs62147676 Chr.2: 86028627-86028627 on Build GRCh38
rs750533260 Chr.2: 86028561-86028561 on Build GRCh38
rs771090979 Chr.2: 86028673-86028673 on Build GRCh38
rs890105026 Chr.2: 86028503-86028503 on Build GRCh38
rs764589818 Chr.2: 86028726-86028726 on Build GRCh38
rs745602873 Chr.2: 86028598-86028598 on Build GRCh38
rs1057140820 Chr.2: 86028508-86028508 on Build GRCh38
rs367979854 Chr.2: 86028642-86028642 on Build GRCh38
rs778388664 Chr.2: 86028580-86028580 on Build GRCh38
rs537491902 Chr.2: 86028635-86028635 on Build GRCh38
rs202201209 Chr.2: 86028628-86028628 on Build GRCh38
rs779427751 Chr.2: 86028571-86028571 on Build GRCh38
rs755152866 Chr.2: 86028567-86028567 on Build GRCh38
rs114751086 Chr.2: 86028529-86028529 on Build GRCh38
rs762889888 Chr.2: 86028553-86028553 on Build GRCh38
rs904603520 Chr.2: 86028729-86028729 on Build GRCh38
rs924088334 Chr.2: 86028622-86028622 on Build GRCh38
rs200147381 Chr.2: 86028566-86028566 on Build GRCh38
rs747026297 Chr.2: 86028514-86028514 on Build GRCh38
rs1000360755 Chr.2: 86028537-86028537 on Build GRCh38
rs771887253 Chr.2: 86028696-86028696 on Build GRCh38

View Full Product Details