Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 16039466-16039697 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCOR1
Entrez Gene ID
9611
Gene Name
nuclear receptor corepressor 1
Gene Aliases
N-CoR, N-CoR1, PPP1R109, TRAC1, hN-CoR
UniGene
Hs.462323
Gene Chromosome Location
Chr.17: 16030094-16216746 on Build GRCh38
Transcripts
NM_001190440.1 NM_006311.3 XM_005256866.4 XM_005256868.4 XM_005256871.4 XM_005256872.4 XM_005256873.4 XM_005256874.4 XM_005256875.3 XM_006721601.3 XM_006721602.3 XM_006721603.3 XM_006721604.3 XM_011524083.2 XM_011524084.2 XM_011524085.2 XM_011524086.2 XM_017025396.1 XM_017025397.1 XM_017025398.1 XM_017025399.1 XM_017025400.1 XM_017025401.1 XM_017025402.1 XM_017025403.1 XM_017025404.1 XM_017025405.1 XM_017025406.1 XM_017025407.1 XM_017025408.1 XM_017025409.1 XM_017025410.1 XM_017025411.1 XM_017025412.1 XM_017025413.1 XM_017025414.1 XM_017025415.1 XM_017025416.1 XM_017025417.1 XM_017025418.1 XM_017025419.1 XM_017025420.1 XM_017024801.1 XM_017024802.1

Gene Symbol
TTC19
Entrez Gene ID
54902
Gene Name
tetratricopeptide repeat domain 19
Gene Aliases
2010204O13Rik, MC3DN2
UniGene
Hs.462316
Gene Chromosome Location
Chr.17: 15999380-16045428 on Build GRCh38
Transcripts
NM_001190440.1 NM_006311.3 XM_005256866.4 XM_005256868.4 XM_005256871.4 XM_005256872.4 XM_005256873.4 XM_005256874.4 XM_005256875.3 XM_006721601.3 XM_006721602.3 XM_006721603.3 XM_006721604.3 XM_011524083.2 XM_011524084.2 XM_011524085.2 XM_011524086.2 XM_017025396.1 XM_017025397.1 XM_017025398.1 XM_017025399.1 XM_017025400.1 XM_017025401.1 XM_017025402.1 XM_017025403.1 XM_017025404.1 XM_017025405.1 XM_017025406.1 XM_017025407.1 XM_017025408.1 XM_017025409.1 XM_017025410.1 XM_017025411.1 XM_017025412.1 XM_017025413.1 XM_017025414.1 XM_017025415.1 XM_017025416.1 XM_017025417.1 XM_017025418.1 XM_017025419.1 XM_017025420.1 XM_017024801.1 XM_017024802.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3795297 c.6813C>T p.L2271L Chr.17: 16039575-16039575 on Build GRCh38
6031549 c.6531_6533delGGA p.E2177delE Chr.17: 16039546-16039548 on Build GRCh38
4814121 c.6471T>A p.N2157K Chr.17: 16039608-16039608 on Build GRCh38
2738940 c.6758C>A p.S2253Y Chr.17: 16039630-16039630 on Build GRCh38
6966623 c.6428C>T p.S2143L Chr.17: 16039651-16039651 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199646563 Chr.17: 16039661-16039661 on Build GRCh38
rs751936193 Chr.17: 16039537-16039537 on Build GRCh38
rs761258580 Chr.17: 16039657-16039657 on Build GRCh38
rs778079269 Chr.17: 16039488-16039488 on Build GRCh38
rs374388689 Chr.17: 16039663-16039663 on Build GRCh38
rs61754986 Chr.17: 16039548-16039548 on Build GRCh38
rs562623179 Chr.17: 16039668-16039668 on Build GRCh38
rs371002943 Chr.17: 16039590-16039590 on Build GRCh38
rs149921069 Chr.17: 16039615-16039615 on Build GRCh38
rs373591408 Chr.17: 16039572-16039572 on Build GRCh38
rs371914465 Chr.17: 16039665-16039665 on Build GRCh38
rs774282901 Chr.17: 16039495-16039495 on Build GRCh38
rs766789395 Chr.17: 16039531-16039531 on Build GRCh38
rs771721292 Chr.17: 16039611-16039611 on Build GRCh38
rs753533227 Chr.17: 16039520-16039520 on Build GRCh38
rs544134448 Chr.17: 16039565-16039565 on Build GRCh38
rs984411549 Chr.17: 16039487-16039487 on Build GRCh38
rs898872808 Chr.17: 16039649-16039649 on Build GRCh38
rs772022058 Chr.17: 16039667-16039667 on Build GRCh38
rs770888727 Chr.17: 16039493-16039493 on Build GRCh38
rs745660317 Chr.17: 16039592-16039592 on Build GRCh38
rs61755986 Chr.17: 16039634-16039634 on Build GRCh38
rs768282882 Chr.17: 16039627-16039627 on Build GRCh38
rs139149781 Chr.17: 16039518-16039518 on Build GRCh38
rs958657707 Chr.17: 16039516-16039516 on Build GRCh38
rs760259611 Chr.17: 16039508-16039508 on Build GRCh38
rs564442571 Chr.17: 16039571-16039571 on Build GRCh38
rs556258444 Chr.17: 16039496-16039496 on Build GRCh38
rs924081269 Chr.17: 16039599-16039599 on Build GRCh38
rs753096531 Chr.17: 16039567-16039567 on Build GRCh38
rs764876927 Chr.17: 16039660-16039660 on Build GRCh38
rs761454179 Chr.17: 16039523-16039523 on Build GRCh38
rs3205089 Chr.17: 16039597-16039597 on Build GRCh38
rs200702771 Chr.17: 16039555-16039555 on Build GRCh38
rs909552730 Chr.17: 16039497-16039497 on Build GRCh38
rs949787616 Chr.17: 16039539-16039539 on Build GRCh38
rs749499082 Chr.17: 16039489-16039489 on Build GRCh38
rs1014686017 Chr.17: 16039647-16039647 on Build GRCh38
rs749587059 Chr.17: 16039574-16039574 on Build GRCh38
rs775220743 Chr.17: 16039507-16039507 on Build GRCh38
rs775515288 Chr.17: 16039612-16039612 on Build GRCh38

View Full Product Details