Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143727991-143728260 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM83H
Entrez Gene ID
286077
Gene Name
family with sequence similarity 83 member H
Gene Aliases
AI3
UniGene
Hs.67776
Gene Chromosome Location
Chr.8: 143723933-143739947 on Build GRCh38
Transcripts
NM_198488.3 XM_005250887.3 XM_005250888.3 XM_005250889.3 XM_011516980.2 XM_011516981.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1755608 c.1395G>T p.T465T Chr.8: 143728066-143728066 on Build GRCh38
4463713 c.1302C>T p.F434F Chr.8: 143728159-143728159 on Build GRCh38
6660280 c.1397C>T p.P466L Chr.8: 143728064-143728064 on Build GRCh38
6660279 c.1450G>A p.A484T Chr.8: 143728011-143728011 on Build GRCh38
7295038 c.1441G>A p.A481T Chr.8: 143728020-143728020 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782789374 Chr.8: 143728058-143728058 on Build GRCh38
rs371890336 Chr.8: 143728212-143728212 on Build GRCh38
rs782802376 Chr.8: 143728207-143728207 on Build GRCh38
rs782183035 Chr.8: 143728023-143728023 on Build GRCh38
rs781820772 Chr.8: 143728075-143728075 on Build GRCh38
rs782171766 Chr.8: 143728206-143728206 on Build GRCh38
rs782217934 Chr.8: 143728158-143728158 on Build GRCh38
rs530884669 Chr.8: 143728192-143728192 on Build GRCh38
rs782614978 Chr.8: 143728067-143728067 on Build GRCh38
rs781910966 Chr.8: 143728156-143728156 on Build GRCh38
rs781813332 Chr.8: 143728222-143728222 on Build GRCh38
rs782767961 Chr.8: 143728174-143728174 on Build GRCh38
rs372976146 Chr.8: 143728054-143728054 on Build GRCh38
rs201120639 Chr.8: 143728025-143728025 on Build GRCh38
rs369438416 Chr.8: 143728049-143728049 on Build GRCh38
rs144418186 Chr.8: 143728176-143728176 on Build GRCh38
rs782246523 Chr.8: 143728113-143728113 on Build GRCh38
rs200076617 Chr.8: 143728136-143728136 on Build GRCh38
rs782609652 Chr.8: 143728142-143728142 on Build GRCh38
rs782154528 Chr.8: 143728056-143728056 on Build GRCh38
rs782237404 Chr.8: 143728104-143728104 on Build GRCh38
rs781937260 Chr.8: 143728165-143728165 on Build GRCh38
rs561916419 Chr.8: 143728162-143728162 on Build GRCh38
rs782048543 Chr.8: 143728215-143728215 on Build GRCh38
rs550491841 Chr.8: 143728171-143728171 on Build GRCh38
rs137854444 Chr.8: 143728082-143728082 on Build GRCh38
rs782570872 Chr.8: 143728097-143728097 on Build GRCh38
rs782084500 Chr.8: 143728128-143728128 on Build GRCh38
rs781972229 Chr.8: 143728041-143728041 on Build GRCh38
rs782436270 Chr.8: 143728026-143728026 on Build GRCh38
rs782372579 Chr.8: 143728187-143728187 on Build GRCh38
rs779195206 Chr.8: 143728032-143728032 on Build GRCh38
rs782354163 Chr.8: 143728024-143728024 on Build GRCh38
rs370721345 Chr.8: 143728163-143728163 on Build GRCh38
rs782114804 Chr.8: 143728050-143728050 on Build GRCh38
rs369685277 Chr.8: 143728064-143728064 on Build GRCh38
rs781971560 Chr.8: 143728127-143728127 on Build GRCh38
rs377700279 Chr.8: 143728105-143728105 on Build GRCh38
rs782089115 Chr.8: 143728042-143728042 on Build GRCh38
rs369006906 Chr.8: 143728234-143728234 on Build GRCh38
rs782512164 Chr.8: 143728233-143728233 on Build GRCh38
rs781852665 Chr.8: 143728135-143728135 on Build GRCh38
rs782053539 Chr.8: 143728169-143728169 on Build GRCh38
rs782081943 Chr.8: 143728194-143728194 on Build GRCh38
rs28573699 Chr.8: 143728152-143728152 on Build GRCh38
rs368072973 Chr.8: 143728189-143728189 on Build GRCh38
rs377309350 Chr.8: 143728061-143728061 on Build GRCh38
rs782627919 Chr.8: 143728009-143728009 on Build GRCh38
rs782489938 Chr.8: 143728180-143728180 on Build GRCh38
rs782535275 Chr.8: 143728241-143728241 on Build GRCh38
rs782021978 Chr.8: 143728029-143728029 on Build GRCh38
rs529093146 Chr.8: 143728164-143728164 on Build GRCh38
rs782508351 Chr.8: 143728149-143728149 on Build GRCh38
rs387907056 Chr.8: 143728095-143728095 on Build GRCh38
rs782487421 Chr.8: 143728141-143728141 on Build GRCh38
rs782272372 Chr.8: 143728146-143728146 on Build GRCh38
rs782530895 Chr.8: 143728010-143728010 on Build GRCh38
rs781991113 Chr.8: 143728204-143728204 on Build GRCh38
rs137854441 Chr.8: 143728053-143728053 on Build GRCh38
rs782263850 Chr.8: 143728182-143728182 on Build GRCh38
rs201186432 Chr.8: 143728175-143728175 on Build GRCh38
rs137854439 Chr.8: 143728081-143728081 on Build GRCh38
rs782268701 Chr.8: 143728150-143728150 on Build GRCh38
rs781900601 Chr.8: 143728236-143728236 on Build GRCh38
rs782428075 Chr.8: 143728120-143728120 on Build GRCh38
rs782579656 Chr.8: 143728018-143728018 on Build GRCh38
rs782059434 Chr.8: 143728123-143728123 on Build GRCh38
rs137854437 Chr.8: 143728218-143728218 on Build GRCh38
rs374749010 Chr.8: 143728030-143728030 on Build GRCh38
rs782525455 Chr.8: 143728077-143728077 on Build GRCh38
rs782399988 Chr.8: 143728196-143728196 on Build GRCh38
rs374210320 Chr.8: 143728078-143728078 on Build GRCh38
rs782276220 Chr.8: 143728080-143728080 on Build GRCh38
rs782591910 Chr.8: 143728181-143728181 on Build GRCh38
rs782005930 Chr.8: 143728108-143728108 on Build GRCh38
rs782693124 Chr.8: 143728126-143728126 on Build GRCh38
rs782732391 Chr.8: 143728137-143728137 on Build GRCh38
rs371289649 Chr.8: 143728122-143728122 on Build GRCh38

View Full Product Details