Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 71648818-71649047 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHLPP2
Entrez Gene ID
23035
Gene Name
PH domain and leucine rich repeat protein phosphatase 2
Gene Aliases
PHLPPL, PPM3B
UniGene
Hs.709458
Gene Chromosome Location
Chr.16: 71644924-71724701 on Build GRCh38
Transcripts
NM_001289003.1 NM_015020.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4566493 c.3946_3947CC>GT p.P1316V Chr.16: 71648915-71648916 on Build GRCh38
472061 c.3915T>C p.P1305P Chr.16: 71648947-71648947 on Build GRCh38
704223 c.3868G>C p.E1290Q Chr.16: 71648994-71648994 on Build GRCh38
3795102 c.3966A>T p.A1322A Chr.16: 71648896-71648896 on Build GRCh38
273107 c.3964G>T p.A1322S Chr.16: 71648898-71648898 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748258512 Chr.16: 71648952-71648952 on Build GRCh38
rs747492423 Chr.16: 71648993-71648993 on Build GRCh38
rs371565551 Chr.16: 71648883-71648883 on Build GRCh38
rs115575792 Chr.16: 71649018-71649018 on Build GRCh38
rs61733124 Chr.16: 71648927-71648927 on Build GRCh38
rs147202140 Chr.16: 71648953-71648953 on Build GRCh38
rs755865525 Chr.16: 71649023-71649023 on Build GRCh38
rs751206937 Chr.16: 71648917-71648917 on Build GRCh38
rs149705576 Chr.16: 71649008-71649008 on Build GRCh38
rs765770499 Chr.16: 71648908-71648908 on Build GRCh38
rs146034837 Chr.16: 71648956-71648956 on Build GRCh38
rs753212273 Chr.16: 71648997-71648997 on Build GRCh38
rs371375607 Chr.16: 71648899-71648899 on Build GRCh38
rs753711947 Chr.16: 71648991-71648993 on Build GRCh38
rs139626439 Chr.16: 71648923-71648923 on Build GRCh38
rs777760011 Chr.16: 71648972-71648973 on Build GRCh38
rs760864609 Chr.16: 71648958-71648958 on Build GRCh38
rs752265941 Chr.16: 71648909-71648909 on Build GRCh38
rs192113290 Chr.16: 71648957-71648957 on Build GRCh38
rs141296681 Chr.16: 71648915-71648915 on Build GRCh38
rs61733125 Chr.16: 71648893-71648893 on Build GRCh38
rs777229982 Chr.16: 71648847-71648847 on Build GRCh38
rs147433033 Chr.16: 71648922-71648922 on Build GRCh38
rs765865741 Chr.16: 71648995-71648995 on Build GRCh38
rs139511750 Chr.16: 71649005-71649005 on Build GRCh38
rs201990639 Chr.16: 71648887-71648887 on Build GRCh38
rs770587173 Chr.16: 71648853-71648853 on Build GRCh38
rs756867990 Chr.16: 71649013-71649013 on Build GRCh38
rs760184822 Chr.16: 71648905-71648905 on Build GRCh38
rs746839361 Chr.16: 71648849-71648849 on Build GRCh38
rs780842467 Chr.16: 71649014-71649014 on Build GRCh38
rs756924434 Chr.16: 71648919-71648919 on Build GRCh38
rs1023953103 Chr.16: 71648994-71648994 on Build GRCh38
rs759096889 Chr.16: 71648883-71648884 on Build GRCh38
rs777138639 Chr.16: 71648985-71648985 on Build GRCh38
rs201211632 Chr.16: 71648901-71648901 on Build GRCh38
rs201141453 Chr.16: 71648900-71648900 on Build GRCh38
rs977504865 Chr.16: 71648932-71648932 on Build GRCh38
rs557846154 Chr.16: 71648872-71648873 on Build GRCh38
rs961802089 Chr.16: 71648869-71648869 on Build GRCh38
rs376338553 Chr.16: 71648976-71648976 on Build GRCh38
rs745867400 Chr.16: 71648880-71648880 on Build GRCh38
rs752747649 Chr.16: 71648899-71648902 on Build GRCh38
rs758126134 Chr.16: 71648845-71648845 on Build GRCh38
rs144133911 Chr.16: 71648914-71648914 on Build GRCh38
rs200189816 Chr.16: 71648904-71648904 on Build GRCh38
rs534044390 Chr.16: 71648839-71648839 on Build GRCh38
rs773267489 Chr.16: 71648898-71648898 on Build GRCh38
rs756088786 Chr.16: 71648905-71648907 on Build GRCh38
rs751079826 Chr.16: 71649011-71649011 on Build GRCh38
rs776468659 Chr.16: 71648872-71648872 on Build GRCh38
rs779920332 Chr.16: 71648928-71648928 on Build GRCh38
rs771178310 Chr.16: 71648968-71648968 on Build GRCh38
rs759919040 Chr.16: 71648986-71648986 on Build GRCh38
rs766925624 Chr.16: 71648885-71648886 on Build GRCh38

View Full Product Details