Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154000797-154001024 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUP210L
Entrez Gene ID
91181
Gene Name
nucleoporin 210 like
Gene Aliases
-
UniGene
Hs.67639
Gene Chromosome Location
Chr.1: 153992582-154155116 on Build GRCh38
Transcripts
NM_001159484.1 NM_207308.2 XM_011510122.1 XM_011510123.1 XM_011510124.1 XM_017002788.1 XM_017002789.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5570826 c.5327T>C p.V1776A Chr.1: 154000915-154000915 on Build GRCh38
5377755 c.5303C>T p.P1768L Chr.1: 154000939-154000939 on Build GRCh38
4771145 c.5371G>C p.D1791H Chr.1: 154000871-154000871 on Build GRCh38
7286072 c.5379G>T p.L1793F Chr.1: 154000863-154000863 on Build GRCh38
3474574 c.5321C>T p.S1774F Chr.1: 154000921-154000921 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772702324 Chr.1: 154000991-154000991 on Build GRCh38
rs763539130 Chr.1: 154000953-154000953 on Build GRCh38
rs374684568 Chr.1: 154000916-154000916 on Build GRCh38
rs754880817 Chr.1: 154000904-154000904 on Build GRCh38
rs770366491 Chr.1: 154000946-154000946 on Build GRCh38
rs186494072 Chr.1: 154000969-154000969 on Build GRCh38
rs747998705 Chr.1: 154000909-154000909 on Build GRCh38
rs751466649 Chr.1: 154000903-154000903 on Build GRCh38
rs753298593 Chr.1: 154000994-154000994 on Build GRCh38
rs769559025 Chr.1: 154000913-154000913 on Build GRCh38
rs754089610 Chr.1: 154000880-154000880 on Build GRCh38
rs765712918 Chr.1: 154000895-154000895 on Build GRCh38
rs759964165 Chr.1: 154000976-154000976 on Build GRCh38
rs757444945 Chr.1: 154000884-154000884 on Build GRCh38
rs868361673 Chr.1: 154000855-154000855 on Build GRCh38
rs181789514 Chr.1: 154000872-154000872 on Build GRCh38
rs775846838 Chr.1: 154000993-154000993 on Build GRCh38
rs199644738 Chr.1: 154000859-154000859 on Build GRCh38
rs79204435 Chr.1: 154000831-154000831 on Build GRCh38
rs748896050 Chr.1: 154000919-154000919 on Build GRCh38
rs370385865 Chr.1: 154000907-154000907 on Build GRCh38
rs1029134090 Chr.1: 154000870-154000870 on Build GRCh38
rs978297279 Chr.1: 154000972-154000972 on Build GRCh38
rs775793631 Chr.1: 154000842-154000842 on Build GRCh38
rs1021236896 Chr.1: 154000925-154000925 on Build GRCh38
rs777439302 Chr.1: 154000914-154000914 on Build GRCh38

View Full Product Details