Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 20224520-20224791 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR11H6
Entrez Gene ID
122748
Gene Name
olfactory receptor family 11 subfamily H member 6
Gene Aliases
-
UniGene
Hs.553575
Gene Chromosome Location
Chr.14: 20223710-20224702 on Build GRCh38
Transcripts
NM_001004480.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4563393 c.971G>A p.G324E Chr.14: 20224680-20224680 on Build GRCh38
6041597 c.890C>T p.T297I Chr.14: 20224599-20224599 on Build GRCh38
137608 c.932G>A p.R311Q Chr.14: 20224641-20224641 on Build GRCh38
6044624 c.986delA p.N330fs? Chr.14: 20224695-20224695 on Build GRCh38
7236388 c.881T>C p.L294P Chr.14: 20224590-20224590 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142134785 Chr.14: 20224624-20224624 on Build GRCh38
rs774626358 Chr.14: 20224549-20224549 on Build GRCh38
rs973934440 Chr.14: 20224765-20224765 on Build GRCh38
rs745732995 Chr.14: 20224735-20224735 on Build GRCh38
rs771668601 Chr.14: 20224740-20224740 on Build GRCh38
rs777404045 Chr.14: 20224749-20224749 on Build GRCh38
rs191411878 Chr.14: 20224753-20224753 on Build GRCh38
rs760092591 Chr.14: 20224562-20224562 on Build GRCh38
rs777594802 Chr.14: 20224570-20224570 on Build GRCh38
rs760891553 Chr.14: 20224572-20224572 on Build GRCh38
rs866042152 Chr.14: 20224625-20224625 on Build GRCh38
rs777094033 Chr.14: 20224716-20224716 on Build GRCh38
rs775984165 Chr.14: 20224569-20224569 on Build GRCh38
rs139056260 Chr.14: 20224583-20224583 on Build GRCh38
rs948767118 Chr.14: 20224689-20224689 on Build GRCh38
rs768878380 Chr.14: 20224713-20224713 on Build GRCh38
rs967644040 Chr.14: 20224676-20224676 on Build GRCh38
rs757005751 Chr.14: 20224687-20224690 on Build GRCh38
rs541140880 Chr.14: 20224637-20224637 on Build GRCh38
rs768206646 Chr.14: 20224565-20224565 on Build GRCh38
rs751134055 Chr.14: 20224622-20224622 on Build GRCh38
rs201972533 Chr.14: 20224692-20224692 on Build GRCh38
rs753736834 Chr.14: 20224751-20224751 on Build GRCh38
rs749453506 Chr.14: 20224645-20224645 on Build GRCh38
rs912041283 Chr.14: 20224603-20224603 on Build GRCh38
rs752530530 Chr.14: 20224740-20224740 on Build GRCh38
rs111837455 Chr.14: 20224640-20224640 on Build GRCh38
rs372466993 Chr.14: 20224546-20224546 on Build GRCh38
rs765843456 Chr.14: 20224615-20224615 on Build GRCh38
rs756128338 Chr.14: 20224746-20224746 on Build GRCh38
rs749993984 Chr.14: 20224595-20224595 on Build GRCh38
rs751222147 Chr.14: 20224733-20224733 on Build GRCh38
rs149054978 Chr.14: 20224599-20224599 on Build GRCh38
rs141044530 Chr.14: 20224708-20224708 on Build GRCh38
rs749103668 Chr.14: 20224655-20224655 on Build GRCh38
rs778824524 Chr.14: 20224692-20224692 on Build GRCh38
rs774569319 Chr.14: 20224678-20224678 on Build GRCh38
rs915944955 Chr.14: 20224685-20224685 on Build GRCh38
rs957028395 Chr.14: 20224548-20224548 on Build GRCh38
rs143225754 Chr.14: 20224641-20224641 on Build GRCh38
rs371339272 Chr.14: 20224737-20224737 on Build GRCh38
rs771229185 Chr.14: 20224669-20224669 on Build GRCh38
rs373017030 Chr.14: 20224674-20224674 on Build GRCh38
rs746003730 Chr.14: 20224681-20224681 on Build GRCh38
rs910978348 Chr.14: 20224741-20224741 on Build GRCh38

View Full Product Details