Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 56963343-56963576 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOA1
Entrez Gene ID
84273
Gene Name
nitric oxide associated 1
Gene Aliases
C4orf14, MTG3, hAtNOS1, hNOA1, mAtNOS1
UniGene
Hs.8715
Gene Chromosome Location
Chr.4: 56963344-56977660 on Build GRCh38
Transcripts
NM_032313.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3775913 c.2073G>T p.K691N Chr.4: 56963474-56963474 on Build GRCh38
4434332 c.1998C>T p.I666I Chr.4: 56963549-56963549 on Build GRCh38
6727519 c.2008C>T p.R670C Chr.4: 56963539-56963539 on Build GRCh38
1056350 c.2007G>A p.Q669Q Chr.4: 56963540-56963540 on Build GRCh38
5355732 c.1972_1973delCC p.P658fs*7 Chr.4: 56963574-56963575 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753787678 Chr.4: 56963374-56963374 on Build GRCh38
rs747111511 Chr.4: 56963517-56963517 on Build GRCh38
rs377687660 Chr.4: 56963449-56963449 on Build GRCh38
rs756758028 Chr.4: 56963416-56963416 on Build GRCh38
rs758111185 Chr.4: 56963413-56963414 on Build GRCh38
rs769192251 Chr.4: 56963539-56963539 on Build GRCh38
rs781337181 Chr.4: 56963525-56963525 on Build GRCh38
rs777938102 Chr.4: 56963445-56963445 on Build GRCh38
rs201263284 Chr.4: 56963490-56963490 on Build GRCh38
rs1041203595 Chr.4: 56963540-56963540 on Build GRCh38
rs373594983 Chr.4: 56963485-56963485 on Build GRCh38
rs750105041 Chr.4: 56963502-56963502 on Build GRCh38
rs202110056 Chr.4: 56963446-56963446 on Build GRCh38
rs201910058 Chr.4: 56963538-56963538 on Build GRCh38
rs780755808 Chr.4: 56963406-56963406 on Build GRCh38
rs563583487 Chr.4: 56963400-56963400 on Build GRCh38
rs61747375 Chr.4: 56963503-56963503 on Build GRCh38
rs61747378 Chr.4: 56963483-56963483 on Build GRCh38
rs771504430 Chr.4: 56963467-56963470 on Build GRCh38
rs746112009 Chr.4: 56963526-56963526 on Build GRCh38
rs756040659 Chr.4: 56963420-56963420 on Build GRCh38
rs190470042 Chr.4: 56963482-56963482 on Build GRCh38
rs770924191 Chr.4: 56963522-56963522 on Build GRCh38
rs764151864 Chr.4: 56963373-56963373 on Build GRCh38
rs759832067 Chr.4: 56963457-56963457 on Build GRCh38
rs775802734 Chr.4: 56963529-56963529 on Build GRCh38
rs6835933 Chr.4: 56963509-56963509 on Build GRCh38
rs780066118 Chr.4: 56963423-56963423 on Build GRCh38
rs767194752 Chr.4: 56963501-56963501 on Build GRCh38
rs776788258 Chr.4: 56963455-56963455 on Build GRCh38
rs940154739 Chr.4: 56963519-56963519 on Build GRCh38
rs531624777 Chr.4: 56963534-56963534 on Build GRCh38
rs966905903 Chr.4: 56963392-56963392 on Build GRCh38
rs976295009 Chr.4: 56963451-56963451 on Build GRCh38
rs574022841 Chr.4: 56963432-56963432 on Build GRCh38
rs749247295 Chr.4: 56963425-56963425 on Build GRCh38
rs951711611 Chr.4: 56963397-56963397 on Build GRCh38
rs564797177 Chr.4: 56963484-56963484 on Build GRCh38
rs770104365 Chr.4: 56963527-56963527 on Build GRCh38
rs559327089 Chr.4: 56963461-56963461 on Build GRCh38
rs146945780 Chr.4: 56963516-56963516 on Build GRCh38

View Full Product Details