Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 190160757-190161026 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRINP3
Entrez Gene ID
339479
Gene Name
BMP/retinoic acid inducible neural specific 3
Gene Aliases
DBCCR1L, DBCCR1L1, FAM5C
UniGene
Hs.65765
Gene Chromosome Location
Chr.1: 190097662-190477997 on Build GRCh38
Transcripts
NM_001317188.1 NM_199051.2 XM_011509472.2 XM_011509475.2 XM_011509476.2 XM_017001125.1 XM_017001126.1 XM_017001127.1 XM_017001128.1 XM_017001129.1 XM_017001130.1 XM_017001131.1 XM_017001132.1 XM_017001133.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1206405 c.1058G>A p.R353H Chr.1: 190160794-190160794 on Build GRCh38
7227925 c.979A>C p.M327L Chr.1: 190160873-190160873 on Build GRCh38
3369474 c.1049A>C p.N350T Chr.1: 190160803-190160803 on Build GRCh38
7135717 c.996G>T p.M332I Chr.1: 190160856-190160856 on Build GRCh38
333351 c.1080C>A p.N360K Chr.1: 190160772-190160772 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768964414 Chr.1: 190160868-190160868 on Build GRCh38
rs752388166 Chr.1: 190160814-190160814 on Build GRCh38
rs561378221 Chr.1: 190160883-190160883 on Build GRCh38
rs371286556 Chr.1: 190160900-190160900 on Build GRCh38
rs1008081308 Chr.1: 190160819-190160819 on Build GRCh38
rs532630443 Chr.1: 190160980-190160980 on Build GRCh38
rs974547537 Chr.1: 190160956-190160956 on Build GRCh38
rs111689030 Chr.1: 190160899-190160899 on Build GRCh38
rs1035268903 Chr.1: 190160894-190160894 on Build GRCh38
rs776501266 Chr.1: 190160904-190160904 on Build GRCh38
rs754542304 Chr.1: 190160794-190160794 on Build GRCh38
rs199998815 Chr.1: 190160850-190160850 on Build GRCh38
rs752163219 Chr.1: 190160916-190160916 on Build GRCh38
rs761656846 Chr.1: 190160866-190160867 on Build GRCh38
rs560759270 Chr.1: 190160953-190160953 on Build GRCh38
rs772838461 Chr.1: 190160783-190160783 on Build GRCh38
rs759437768 Chr.1: 190160907-190160907 on Build GRCh38
rs199724740 Chr.1: 190160823-190160823 on Build GRCh38
rs112829253 Chr.1: 190160890-190160890 on Build GRCh38
rs772505697 Chr.1: 190160817-190160817 on Build GRCh38
rs746379368 Chr.1: 190160837-190160837 on Build GRCh38
rs778586472 Chr.1: 190160791-190160791 on Build GRCh38
rs1002577984 Chr.1: 190160893-190160893 on Build GRCh38
rs765514781 Chr.1: 190160915-190160917 on Build GRCh38
rs770029165 Chr.1: 190160842-190160842 on Build GRCh38
rs112426635 Chr.1: 190160930-190160930 on Build GRCh38
rs775685772 Chr.1: 190160858-190160858 on Build GRCh38
rs757794234 Chr.1: 190160828-190160828 on Build GRCh38
rs865951009 Chr.1: 190160798-190160798 on Build GRCh38
rs751208269 Chr.1: 190160940-190160940 on Build GRCh38
rs760332336 Chr.1: 190160892-190160892 on Build GRCh38
rs530432467 Chr.1: 190160908-190160908 on Build GRCh38
rs781605000 Chr.1: 190160831-190160831 on Build GRCh38
rs147790033 Chr.1: 190160795-190160795 on Build GRCh38
rs759423552 Chr.1: 190160977-190160977 on Build GRCh38
rs952105344 Chr.1: 190160954-190160954 on Build GRCh38
rs758048297 Chr.1: 190160918-190160918 on Build GRCh38
rs201551652 Chr.1: 190160824-190160824 on Build GRCh38
rs200210126 Chr.1: 190160792-190160792 on Build GRCh38
rs749547925 Chr.1: 190160864-190160864 on Build GRCh38

View Full Product Details