Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 17772245-17772482 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNC1
Entrez Gene ID
3746
Gene Name
potassium voltage-gated channel subfamily C member 1
Gene Aliases
EPM7, KV3.1, KV4, NGK2
UniGene
Hs.552896
Gene Chromosome Location
Chr.11: 17734810-17783055 on Build GRCh38
Transcripts
NM_001112741.1 NM_004976.4 XM_011520080.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6690587 c.1171T>C p.F391L Chr.11: 17772265-17772265 on Build GRCh38
3445914 c.1386G>A p.R462R Chr.11: 17772480-17772480 on Build GRCh38
6690585 c.1344T>C p.A448A Chr.11: 17772438-17772438 on Build GRCh38
3445912 c.1245G>A p.L415L Chr.11: 17772339-17772339 on Build GRCh38
4031874 c.1303A>G p.I435V Chr.11: 17772397-17772397 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113423862 Chr.11: 17772391-17772391 on Build GRCh38
rs776064757 Chr.11: 17772411-17772411 on Build GRCh38
rs73424033 Chr.11: 17772330-17772330 on Build GRCh38
rs758320453 Chr.11: 17772315-17772315 on Build GRCh38
rs545364229 Chr.11: 17772464-17772464 on Build GRCh38
rs754890050 Chr.11: 17772337-17772337 on Build GRCh38
rs760728034 Chr.11: 17772420-17772420 on Build GRCh38
rs779961591 Chr.11: 17772318-17772318 on Build GRCh38
rs150682385 Chr.11: 17772363-17772363 on Build GRCh38
rs148132957 Chr.11: 17772276-17772276 on Build GRCh38
rs374371136 Chr.11: 17772303-17772303 on Build GRCh38
rs139804857 Chr.11: 17772381-17772381 on Build GRCh38
rs769341904 Chr.11: 17772372-17772372 on Build GRCh38
rs777458040 Chr.11: 17772378-17772378 on Build GRCh38
rs908674741 Chr.11: 17772399-17772399 on Build GRCh38
rs754101721 Chr.11: 17772300-17772300 on Build GRCh38
rs747615166 Chr.11: 17772366-17772366 on Build GRCh38
rs577711585 Chr.11: 17772387-17772387 on Build GRCh38
rs202218342 Chr.11: 17772393-17772393 on Build GRCh38
rs574378895 Chr.11: 17772291-17772291 on Build GRCh38
rs915600389 Chr.11: 17772357-17772357 on Build GRCh38

View Full Product Details