Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 46931098-46931325 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOSR2
Entrez Gene ID
9570
Gene Name
golgi SNAP receptor complex member 2
Gene Aliases
Bos1, EPM6, GS27
UniGene
Hs.463278
Gene Chromosome Location
Chr.17: 46923086-46975519 on Build GRCh38
Transcripts
NM_001012511.2 NM_001321133.1 NM_001321134.1 NM_004287.4 NM_054022.3 XM_005257844.3 XM_006722190.3 XM_011525501.2 XM_011525502.2 XM_017025378.1 XM_017025379.1 XM_017025380.1 XM_017025381.1 XM_017025382.1 XM_017025383.1 XM_017025384.1 XM_017025385.1 XM_017025386.1 XM_017025387.1 XM_017025388.1 XM_017025389.1 XM_017025390.1 XM_017025391.1 XM_017025392.1 XM_017025393.1 XM_017025394.1 XM_017025395.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7191188 c.179C>A p.P60H Chr.17: 46931183-46931183 on Build GRCh38
7150300 c.135C>T p.F45F Chr.17: 46931139-46931139 on Build GRCh38
1325009 c.170A>G p.K57R Chr.17: 46931174-46931174 on Build GRCh38
7150303 c.135C>T p.F45F Chr.17: 46931139-46931139 on Build GRCh38
3755609 c.200G>A p.R67K Chr.17: 46931204-46931204 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754535137 Chr.17: 46931203-46931203 on Build GRCh38
rs762867555 Chr.17: 46931162-46931162 on Build GRCh38
rs779696333 Chr.17: 46931192-46931192 on Build GRCh38
rs768756655 Chr.17: 46931244-46931244 on Build GRCh38
rs770063190 Chr.17: 46931229-46931229 on Build GRCh38
rs775702857 Chr.17: 46931233-46931233 on Build GRCh38
rs747328256 Chr.17: 46931143-46931143 on Build GRCh38
rs775000160 Chr.17: 46931157-46931157 on Build GRCh38
rs534119817 Chr.17: 46931140-46931140 on Build GRCh38
rs775618697 Chr.17: 46931160-46931160 on Build GRCh38
rs755897636 Chr.17: 46931190-46931190 on Build GRCh38
rs771307162 Chr.17: 46931212-46931212 on Build GRCh38
rs197922 Chr.17: 46931204-46931204 on Build GRCh38
rs377765254 Chr.17: 46931153-46931153 on Build GRCh38
rs772763319 Chr.17: 46931132-46931132 on Build GRCh38
rs772523876 Chr.17: 46931188-46931188 on Build GRCh38
rs373232844 Chr.17: 46931254-46931254 on Build GRCh38
rs34093958 Chr.17: 46931176-46931177 on Build GRCh38
rs774297006 Chr.17: 46931251-46931251 on Build GRCh38
rs771228855 Chr.17: 46931144-46931144 on Build GRCh38
rs748660197 Chr.17: 46931127-46931127 on Build GRCh38
rs538712056 Chr.17: 46931166-46931166 on Build GRCh38
rs761695109 Chr.17: 46931253-46931253 on Build GRCh38
rs535191147 Chr.17: 46931178-46931178 on Build GRCh38
rs925915075 Chr.17: 46931259-46931259 on Build GRCh38
rs751458272 Chr.17: 46931171-46931171 on Build GRCh38
rs762955002 Chr.17: 46931234-46931234 on Build GRCh38
rs193091114 Chr.17: 46931270-46931270 on Build GRCh38
rs750058795 Chr.17: 46931182-46931182 on Build GRCh38
rs746089403 Chr.17: 46931228-46931228 on Build GRCh38
rs201423456 Chr.17: 46931210-46931210 on Build GRCh38
rs753336865 Chr.17: 46931193-46931193 on Build GRCh38
rs143754727 Chr.17: 46931152-46931152 on Build GRCh38
rs776861115 Chr.17: 46931148-46931148 on Build GRCh38
rs746358802 Chr.17: 46931175-46931175 on Build GRCh38
rs773712776 Chr.17: 46931239-46931239 on Build GRCh38
rs775911834 Chr.17: 46931218-46931219 on Build GRCh38

View Full Product Details