Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 125101130-125101381 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM218
Entrez Gene ID
219854
Gene Name
transmembrane protein 218
Gene Aliases
-
UniGene
Hs.731899
Gene Chromosome Location
Chr.11: 125094370-125111763 on Build GRCh38
Transcripts
NM_001080546.2 NM_001258238.1 NM_001258239.1 NM_001258240.1 NM_001258241.1 NM_001258242.1 NM_001258243.1 NM_001258244.1 NM_001258245.1 NM_001258246.1 NM_001258247.1 NR_047586.1 NR_047587.1 NR_047588.1 NR_047589.1 NR_047590.1 NR_047591.1 NR_047592.1 XM_006718779.3 XM_006718780.3 XM_006718781.3 XM_006718783.3 XM_017017322.1 XM_017017323.1 XM_017017324.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4019020 c.133T>G p.F45V Chr.11: 125101281-125101281 on Build GRCh38
1352358 c.141T>C p.A47A Chr.11: 125101273-125101273 on Build GRCh38
3444943 c.190C>T p.P64S Chr.11: 125101224-125101224 on Build GRCh38
1352359 c.116C>T p.S39F Chr.11: 125101298-125101298 on Build GRCh38
4940991 c.111-2A>T p.? Chr.11: 125101305-125101305 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768089994 Chr.11: 125101269-125101269 on Build GRCh38
rs764217473 Chr.11: 125101342-125101342 on Build GRCh38
rs369008819 Chr.11: 125101213-125101213 on Build GRCh38
rs369336433 Chr.11: 125101348-125101348 on Build GRCh38
rs572010506 Chr.11: 125101281-125101281 on Build GRCh38
rs776335247 Chr.11: 125101153-125101153 on Build GRCh38
rs747752756 Chr.11: 125101277-125101277 on Build GRCh38
rs756328213 Chr.11: 125101200-125101200 on Build GRCh38
rs77509816 Chr.11: 125101156-125101156 on Build GRCh38
rs768288239 Chr.11: 125101151-125101151 on Build GRCh38
rs762863673 Chr.11: 125101300-125101300 on Build GRCh38
rs377432538 Chr.11: 125101210-125101210 on Build GRCh38
rs776133777 Chr.11: 125101276-125101276 on Build GRCh38
rs754054558 Chr.11: 125101211-125101211 on Build GRCh38
rs759558592 Chr.11: 125101336-125101336 on Build GRCh38
rs758860898 Chr.11: 125101196-125101196 on Build GRCh38
rs777278582 Chr.11: 125101194-125101194 on Build GRCh38
rs757386185 Chr.11: 125101354-125101354 on Build GRCh38
rs774362607 Chr.11: 125101331-125101331 on Build GRCh38
rs201696124 Chr.11: 125101183-125101183 on Build GRCh38
rs766222848 Chr.11: 125101316-125101316 on Build GRCh38
rs752705898 Chr.11: 125101192-125101192 on Build GRCh38
rs376065497 Chr.11: 125101278-125101278 on Build GRCh38
rs138480376 Chr.11: 125101227-125101227 on Build GRCh38
rs552099293 Chr.11: 125101279-125101279 on Build GRCh38
rs780681509 Chr.11: 125101263-125101265 on Build GRCh38
rs1056061916 Chr.11: 125101252-125101252 on Build GRCh38
rs746522788 Chr.11: 125101240-125101240 on Build GRCh38
rs750256788 Chr.11: 125101180-125101180 on Build GRCh38
rs747596049 Chr.11: 125101352-125101352 on Build GRCh38
rs201187750 Chr.11: 125101220-125101220 on Build GRCh38
rs758805438 Chr.11: 125101239-125101239 on Build GRCh38
rs762759481 Chr.11: 125101182-125101182 on Build GRCh38
rs78488446 Chr.11: 125101167-125101167 on Build GRCh38
rs960305658 Chr.11: 125101341-125101341 on Build GRCh38
rs34985373 Chr.11: 125101179-125101179 on Build GRCh38
rs372444659 Chr.11: 125101238-125101238 on Build GRCh38

View Full Product Details