Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 2061407-2061660 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMG6
Entrez Gene ID
23293
Gene Name
SMG6, nonsense mediated mRNA decay factor
Gene Aliases
C17orf31, EST1A, SMG-6, hSMG5/7a
UniGene
Hs.448342
Gene Chromosome Location
Chr.17: 2059839-2303836 on Build GRCh38
Transcripts
NM_001256827.1 NM_001256828.1 NM_001282326.1 NM_017575.4 XM_005256569.3 XM_005256571.4 XM_011523769.2 XM_011523775.2 XM_017024399.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6390820 c.4184T>C p.L1395P Chr.17: 2061568-2061568 on Build GRCh38
976456 c.4153G>T p.E1385* Chr.17: 2061599-2061599 on Build GRCh38
976458 c.4141C>T p.R1381W Chr.17: 2061611-2061611 on Build GRCh38
3387743 c.4138A>G p.I1380V Chr.17: 2061614-2061614 on Build GRCh38
4064676 c.4186C>T p.R1396C Chr.17: 2061566-2061566 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1027491780 Chr.17: 2061537-2061537 on Build GRCh38
rs943586326 Chr.17: 2061426-2061426 on Build GRCh38
rs771725003 Chr.17: 2061466-2061466 on Build GRCh38
rs746325036 Chr.17: 2061519-2061519 on Build GRCh38
rs757197344 Chr.17: 2061476-2061476 on Build GRCh38
rs749994929 Chr.17: 2061458-2061458 on Build GRCh38
rs762549957 Chr.17: 2061463-2061463 on Build GRCh38
rs932843791 Chr.17: 2061473-2061474 on Build GRCh38
rs36097261 Chr.17: 2061472-2061473 on Build GRCh38
rs951841523 Chr.17: 2061632-2061632 on Build GRCh38
rs774357112 Chr.17: 2061468-2061469 on Build GRCh38
rs775769121 Chr.17: 2061525-2061525 on Build GRCh38
rs866312155 Chr.17: 2061602-2061602 on Build GRCh38
rs752643842 Chr.17: 2061505-2061505 on Build GRCh38
rs770234882 Chr.17: 2061466-2061467 on Build GRCh38
rs751395020 Chr.17: 2061467-2061467 on Build GRCh38
rs776762033 Chr.17: 2061555-2061555 on Build GRCh38
rs761092700 Chr.17: 2061528-2061528 on Build GRCh38
rs144129895 Chr.17: 2061582-2061582 on Build GRCh38
rs756376437 Chr.17: 2061474-2061474 on Build GRCh38
rs762566596 Chr.17: 2061534-2061534 on Build GRCh38
rs567513304 Chr.17: 2061464-2061465 on Build GRCh38
rs777936944 Chr.17: 2061476-2061476 on Build GRCh38
rs371827551 Chr.17: 2061511-2061511 on Build GRCh38
rs752984487 Chr.17: 2061469-2061469 on Build GRCh38
rs749434185 Chr.17: 2061491-2061491 on Build GRCh38
rs776381239 Chr.17: 2061443-2061443 on Build GRCh38
rs761326971 Chr.17: 2061454-2061454 on Build GRCh38
rs894970159 Chr.17: 2061531-2061531 on Build GRCh38
rs777798090 Chr.17: 2061463-2061464 on Build GRCh38
rs866566302 Chr.17: 2061618-2061618 on Build GRCh38
rs772512575 Chr.17: 2061524-2061524 on Build GRCh38
rs773744379 Chr.17: 2061467-2061468 on Build GRCh38
rs765919281 Chr.17: 2061539-2061539 on Build GRCh38
rs145926620 Chr.17: 2061465-2061466 on Build GRCh38
rs368411080 Chr.17: 2061503-2061503 on Build GRCh38
rs769639379 Chr.17: 2061529-2061529 on Build GRCh38
rs77194529 Chr.17: 2061457-2061457 on Build GRCh38
rs760570965 Chr.17: 2061470-2061471 on Build GRCh38
rs867343563 Chr.17: 2061621-2061621 on Build GRCh38
rs757362746 Chr.17: 2061640-2061640 on Build GRCh38
rs867472768 Chr.17: 2061601-2061601 on Build GRCh38
rs766483955 Chr.17: 2061466-2061466 on Build GRCh38
rs770980971 Chr.17: 2061510-2061510 on Build GRCh38
rs775900116 Chr.17: 2061624-2061625 on Build GRCh38
rs753809183 Chr.17: 2061474-2061475 on Build GRCh38
rs867547820 Chr.17: 2061546-2061546 on Build GRCh38
rs373285460 Chr.17: 2061611-2061611 on Build GRCh38
rs141226599 Chr.17: 2061532-2061532 on Build GRCh38
rs965723260 Chr.17: 2061606-2061606 on Build GRCh38
rs369323853 Chr.17: 2061565-2061565 on Build GRCh38
rs755977968 Chr.17: 2061614-2061614 on Build GRCh38
rs960617013 Chr.17: 2061588-2061588 on Build GRCh38
rs753998015 Chr.17: 2061639-2061639 on Build GRCh38
rs572844711 Chr.17: 2061437-2061437 on Build GRCh38
rs558385917 Chr.17: 2061468-2061468 on Build GRCh38
rs867280691 Chr.17: 2061566-2061566 on Build GRCh38
rs376525214 Chr.17: 2061521-2061521 on Build GRCh38
rs760294697 Chr.17: 2061438-2061438 on Build GRCh38
rs550810571 Chr.17: 2061436-2061436 on Build GRCh38
rs397730291 Chr.17: 2061475-2061476 on Build GRCh38
rs751547450 Chr.17: 2061554-2061554 on Build GRCh38
rs200494730 Chr.17: 2061564-2061564 on Build GRCh38
rs199666681 Chr.17: 2061469-2061470 on Build GRCh38
rs772262968 Chr.17: 2061442-2061442 on Build GRCh38

View Full Product Details