Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43416450-43416721 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SZT2
Entrez Gene ID
23334
Gene Name
seizure threshold 2 homolog (mouse)
Gene Aliases
C1orf84, EIEE18, KIAA0467, SZT2A, SZT2B
UniGene
Hs.643560
Gene Chromosome Location
Chr.1: 43389884-43454247 on Build GRCh38
Transcripts
NM_015284.3 XM_005270686.3 XM_006710501.3 XM_011541106.2 XM_011541107.2 XM_017000819.1 XM_017000820.1 XM_017000821.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3369775 c.871T>G p.F291V Chr.1: 43416633-43416633 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs746758732 Chr.1: 43416580-43416580 on Build GRCh38
rs774024938 Chr.1: 43416567-43416567 on Build GRCh38
rs545791977 Chr.1: 43416543-43416543 on Build GRCh38
rs779077199 Chr.1: 43416544-43416544 on Build GRCh38
rs750501691 Chr.1: 43416689-43416689 on Build GRCh38
rs36004858 Chr.1: 43416654-43416654 on Build GRCh38
rs1051327530 Chr.1: 43416564-43416564 on Build GRCh38
rs762701931 Chr.1: 43416664-43416664 on Build GRCh38
rs762521533 Chr.1: 43416505-43416505 on Build GRCh38
rs370390171 Chr.1: 43416578-43416578 on Build GRCh38
rs750697937 Chr.1: 43416550-43416550 on Build GRCh38
rs763660997 Chr.1: 43416525-43416525 on Build GRCh38
rs925316889 Chr.1: 43416519-43416519 on Build GRCh38
rs373897662 Chr.1: 43416597-43416597 on Build GRCh38
rs760942511 Chr.1: 43416674-43416674 on Build GRCh38
rs764450888 Chr.1: 43416676-43416676 on Build GRCh38
rs769312692 Chr.1: 43416633-43416633 on Build GRCh38
rs752969635 Chr.1: 43416503-43416503 on Build GRCh38
rs762362086 Chr.1: 43416684-43416684 on Build GRCh38
rs772666294 Chr.1: 43416663-43416663 on Build GRCh38
rs375794037 Chr.1: 43416542-43416542 on Build GRCh38
rs770464311 Chr.1: 43416671-43416671 on Build GRCh38
rs896985784 Chr.1: 43416540-43416540 on Build GRCh38
rs748015088 Chr.1: 43416610-43416610 on Build GRCh38
rs764494467 Chr.1: 43416507-43416507 on Build GRCh38
rs1055041289 Chr.1: 43416472-43416472 on Build GRCh38
rs768700110 Chr.1: 43416594-43416594 on Build GRCh38
rs765441147 Chr.1: 43416685-43416685 on Build GRCh38
rs889448726 Chr.1: 43416575-43416575 on Build GRCh38
rs775979348 Chr.1: 43416672-43416672 on Build GRCh38
rs909831106 Chr.1: 43416499-43416499 on Build GRCh38
rs559039208 Chr.1: 43416682-43416682 on Build GRCh38

View Full Product Details