Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17283910-17284161 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKLE1
Entrez Gene ID
126549
Gene Name
ankyrin repeat and LEM domain containing 1
Gene Aliases
ANKRD41, LEM3, LEMD6
UniGene
Hs.721610
Gene Chromosome Location
Chr.19: 17281645-17287646 on Build GRCh38
Transcripts
NM_001278443.1 NM_001278444.1 NM_001278445.1 NM_152363.5 NR_103530.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1391426 c.1113T>C p.A371A Chr.19: 17284123-17284123 on Build GRCh38
1182824 c.1243C>T p.R415W Chr.19: 17284133-17284133 on Build GRCh38
4140368 c.1113T>A p.A371A Chr.19: 17284123-17284123 on Build GRCh38
6885089 c.1193C>T p.A398V Chr.19: 17283957-17283957 on Build GRCh38
6600136 c.1105G>T p.E369* Chr.19: 17284115-17284115 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768503971 Chr.19: 17284129-17284129 on Build GRCh38
rs866029345 Chr.19: 17284096-17284096 on Build GRCh38
rs775379907 Chr.19: 17284011-17284011 on Build GRCh38
rs778991017 Chr.19: 17284087-17284087 on Build GRCh38
rs756524552 Chr.19: 17284065-17284065 on Build GRCh38
rs781643970 Chr.19: 17283957-17283957 on Build GRCh38
rs750761050 Chr.19: 17284118-17284118 on Build GRCh38
rs756712231 Chr.19: 17284130-17284130 on Build GRCh38
rs759355662 Chr.19: 17284004-17284004 on Build GRCh38
rs751047354 Chr.19: 17284048-17284048 on Build GRCh38
rs772238259 Chr.19: 17284093-17284093 on Build GRCh38
rs749809111 Chr.19: 17283976-17283976 on Build GRCh38
rs768767390 Chr.19: 17283979-17283979 on Build GRCh38
rs775271496 Chr.19: 17284121-17284121 on Build GRCh38
rs902212697 Chr.19: 17284033-17284033 on Build GRCh38
rs374482694 Chr.19: 17284056-17284056 on Build GRCh38
rs778925239 Chr.19: 17283980-17283980 on Build GRCh38
rs755444858 Chr.19: 17284079-17284079 on Build GRCh38
rs998940544 Chr.19: 17284088-17284088 on Build GRCh38
rs941010100 Chr.19: 17284013-17284013 on Build GRCh38
rs369819688 Chr.19: 17283983-17283983 on Build GRCh38
rs139466646 Chr.19: 17283982-17283982 on Build GRCh38
rs910806903 Chr.19: 17283948-17283948 on Build GRCh38
rs766782760 Chr.19: 17284138-17284138 on Build GRCh38
rs748197557 Chr.19: 17284092-17284092 on Build GRCh38
rs66753001 Chr.19: 17284030-17284030 on Build GRCh38
rs767186144 Chr.19: 17284134-17284134 on Build GRCh38
rs543893832 Chr.19: 17284094-17284094 on Build GRCh38
rs780624435 Chr.19: 17284071-17284071 on Build GRCh38
rs758117979 Chr.19: 17284032-17284032 on Build GRCh38
rs756868016 Chr.19: 17283951-17283951 on Build GRCh38
rs201075128 Chr.19: 17283961-17283961 on Build GRCh38
rs200745328 Chr.19: 17284012-17284012 on Build GRCh38
rs760470646 Chr.19: 17284137-17284137 on Build GRCh38
rs750142045 Chr.19: 17284062-17284062 on Build GRCh38
rs754364374 Chr.19: 17284072-17284072 on Build GRCh38
rs762892589 Chr.19: 17284128-17284128 on Build GRCh38
rs769612263 Chr.19: 17284010-17284010 on Build GRCh38
rs750088569 Chr.19: 17284135-17284135 on Build GRCh38
rs373197020 Chr.19: 17284014-17284014 on Build GRCh38
rs769560798 Chr.19: 17284110-17284110 on Build GRCh38
rs200487914 Chr.19: 17284045-17284045 on Build GRCh38
rs780675948 Chr.19: 17283970-17283970 on Build GRCh38
rs756650852 Chr.19: 17283966-17283966 on Build GRCh38
rs776358412 Chr.19: 17283991-17283991 on Build GRCh38
rs151076335 Chr.19: 17284133-17284133 on Build GRCh38
rs999281963 Chr.19: 17284038-17284038 on Build GRCh38
rs375871327 Chr.19: 17284054-17284054 on Build GRCh38
rs976287836 Chr.19: 17283963-17283963 on Build GRCh38
rs894405724 Chr.19: 17284031-17284031 on Build GRCh38
rs751224470 Chr.19: 17283945-17283945 on Build GRCh38
rs367974930 Chr.19: 17284046-17284046 on Build GRCh38

View Full Product Details