Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 19303273-19303524 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUGP1
Entrez Gene ID
57794
Gene Name
SURP and G-patch domain containing 1
Gene Aliases
F23858, RBP, SF4
UniGene
Hs.515274
Gene Chromosome Location
Chr.19: 19276513-19320512 on Build GRCh38
Transcripts
NM_172231.3 XM_005260002.2 XM_011528155.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6792843 c.673G>A p.D225N Chr.19: 19303438-19303438 on Build GRCh38
1325379 c.702C>A p.Y234* Chr.19: 19303409-19303409 on Build GRCh38
6150008 c.730G>C p.E244Q Chr.19: 19303381-19303381 on Build GRCh38
7259679 c.744G>A p.S248S Chr.19: 19303367-19303367 on Build GRCh38
6264644 c.759G>A p.Q253Q Chr.19: 19303352-19303352 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371260349 Chr.19: 19303340-19303340 on Build GRCh38
rs769033669 Chr.19: 19303403-19303403 on Build GRCh38
rs369252834 Chr.19: 19303302-19303302 on Build GRCh38
rs755593835 Chr.19: 19303388-19303388 on Build GRCh38
rs745392386 Chr.19: 19303383-19303383 on Build GRCh38
rs758151949 Chr.19: 19303298-19303298 on Build GRCh38
rs7508644 Chr.19: 19303415-19303415 on Build GRCh38
rs914332860 Chr.19: 19303423-19303423 on Build GRCh38
rs377357759 Chr.19: 19303333-19303333 on Build GRCh38
rs776559099 Chr.19: 19303457-19303457 on Build GRCh38
rs749107561 Chr.19: 19303459-19303459 on Build GRCh38
rs767292710 Chr.19: 19303480-19303480 on Build GRCh38
rs564501602 Chr.19: 19303353-19303353 on Build GRCh38
rs371344364 Chr.19: 19303470-19303470 on Build GRCh38
rs769000483 Chr.19: 19303327-19303327 on Build GRCh38
rs780432892 Chr.19: 19303389-19303389 on Build GRCh38
rs761825051 Chr.19: 19303472-19303473 on Build GRCh38
rs750027929 Chr.19: 19303486-19303486 on Build GRCh38
rs776017903 Chr.19: 19303316-19303316 on Build GRCh38
rs1004591228 Chr.19: 19303292-19303292 on Build GRCh38
rs373329475 Chr.19: 19303332-19303332 on Build GRCh38
rs768830006 Chr.19: 19303460-19303469 on Build GRCh38
rs146659898 Chr.19: 19303367-19303367 on Build GRCh38
rs765097277 Chr.19: 19303350-19303350 on Build GRCh38
rs200227561 Chr.19: 19303492-19303492 on Build GRCh38
rs932888304 Chr.19: 19303493-19303493 on Build GRCh38
rs981380888 Chr.19: 19303348-19303348 on Build GRCh38
rs770235516 Chr.19: 19303315-19303315 on Build GRCh38
rs980750977 Chr.19: 19303365-19303365 on Build GRCh38
rs553195068 Chr.19: 19303439-19303439 on Build GRCh38
rs779676090 Chr.19: 19303491-19303491 on Build GRCh38
rs774696589 Chr.19: 19303330-19303330 on Build GRCh38
rs762996479 Chr.19: 19303460-19303460 on Build GRCh38
rs751432162 Chr.19: 19303464-19303464 on Build GRCh38
rs770694106 Chr.19: 19303307-19303307 on Build GRCh38
rs181184094 Chr.19: 19303434-19303434 on Build GRCh38
rs773362606 Chr.19: 19303427-19303427 on Build GRCh38
rs767708960 Chr.19: 19303337-19303337 on Build GRCh38
rs772299597 Chr.19: 19303413-19303413 on Build GRCh38
rs774751475 Chr.19: 19303409-19303409 on Build GRCh38
rs201089916 Chr.19: 19303410-19303410 on Build GRCh38
rs757069779 Chr.19: 19303467-19303467 on Build GRCh38
rs376230640 Chr.19: 19303297-19303297 on Build GRCh38
rs374307015 Chr.19: 19303428-19303428 on Build GRCh38
rs376933215 Chr.19: 19303466-19303466 on Build GRCh38
rs774563504 Chr.19: 19303471-19303472 on Build GRCh38
rs780898434 Chr.19: 19303308-19303308 on Build GRCh38
rs368137482 Chr.19: 19303479-19303479 on Build GRCh38
rs573356647 Chr.19: 19303454-19303454 on Build GRCh38
rs755719169 Chr.19: 19303490-19303490 on Build GRCh38
rs749876599 Chr.19: 19303402-19303402 on Build GRCh38
rs758261723 Chr.19: 19303361-19303361 on Build GRCh38
rs748468708 Chr.19: 19303411-19303411 on Build GRCh38
rs750529216 Chr.19: 19303338-19303338 on Build GRCh38
rs745340165 Chr.19: 19303313-19303313 on Build GRCh38
rs764037415 Chr.19: 19303362-19303362 on Build GRCh38
rs764092542 Chr.19: 19303463-19303463 on Build GRCh38
rs746724288 Chr.19: 19303303-19303303 on Build GRCh38
rs755523644 Chr.19: 19303498-19303498 on Build GRCh38
rs763242431 Chr.19: 19303324-19303324 on Build GRCh38
rs1978144 Chr.19: 19303368-19303368 on Build GRCh38

View Full Product Details