Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 110632263-110632536 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCTN1
Entrez Gene ID
79600
Gene Name
tectonic family member 1
Gene Aliases
JBTS13, TECT1
UniGene
Hs.211511
Gene Chromosome Location
Chr.12: 110614027-110649130 on Build GRCh38
Transcripts
NM_001082537.2 NM_001082538.2 NM_001173975.2 NM_001173976.1 NM_001319680.1 NM_001319681.1 NM_024549.5 NR_135088.1 XM_005253934.3 XM_005253935.3 XM_006719594.2 XM_006719595.2 XM_006719596.2 XM_006719597.3 XM_006719598.2 XM_006719599.2 XM_006719600.2 XM_011538733.2 XM_011538734.2 XM_011538735.1 XM_011538737.2 XM_011538738.2 XM_017019964.1 XM_017019965.1 XM_017019966.1 XM_017019967.1 XM_017019968.1 XM_017019969.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5909307 c.653C>T p.S218L Chr.12: 110632500-110632500 on Build GRCh38
6798926 c.637C>G p.L213V Chr.12: 110632484-110632484 on Build GRCh38
5909308 c.653C>T p.S218L Chr.12: 110632500-110632500 on Build GRCh38
5567704 c.654G>A p.S218S Chr.12: 110632501-110632501 on Build GRCh38
7428789 c.635_636delCT p.L213fs*20 Chr.12: 110632482-110632483 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779157923 Chr.12: 110632432-110632432 on Build GRCh38
rs781253687 Chr.12: 110632439-110632439 on Build GRCh38
rs755146349 Chr.12: 110632416-110632416 on Build GRCh38
rs987734321 Chr.12: 110632355-110632355 on Build GRCh38
rs943520943 Chr.12: 110632342-110632342 on Build GRCh38
rs777545986 Chr.12: 110632461-110632461 on Build GRCh38
rs573994317 Chr.12: 110632349-110632349 on Build GRCh38
rs748019534 Chr.12: 110632444-110632444 on Build GRCh38
rs971915314 Chr.12: 110632304-110632304 on Build GRCh38
rs575159541 Chr.12: 110632407-110632407 on Build GRCh38
rs754413948 Chr.12: 110632423-110632423 on Build GRCh38
rs779901439 Chr.12: 110632456-110632458 on Build GRCh38
rs545613207 Chr.12: 110632491-110632491 on Build GRCh38
rs757932264 Chr.12: 110632428-110632432 on Build GRCh38
rs756019255 Chr.12: 110632447-110632447 on Build GRCh38
rs889172120 Chr.12: 110632339-110632339 on Build GRCh38
rs763992416 Chr.12: 110632500-110632500 on Build GRCh38
rs201805622 Chr.12: 110632473-110632473 on Build GRCh38
rs760316119 Chr.12: 110632369-110632369 on Build GRCh38
rs1026548377 Chr.12: 110632388-110632388 on Build GRCh38
rs1042632342 Chr.12: 110632443-110632443 on Build GRCh38
rs1043867530 Chr.12: 110632334-110632334 on Build GRCh38
rs970228 Chr.12: 110632364-110632364 on Build GRCh38
rs780221979 Chr.12: 110632478-110632478 on Build GRCh38
rs770297289 Chr.12: 110632514-110632514 on Build GRCh38
rs886812040 Chr.12: 110632441-110632441 on Build GRCh38
rs772332249 Chr.12: 110632476-110632476 on Build GRCh38
rs12228683 Chr.12: 110632357-110632357 on Build GRCh38
rs776715588 Chr.12: 110632501-110632501 on Build GRCh38
rs191374494 Chr.12: 110632377-110632377 on Build GRCh38

View Full Product Details