Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 74427072-74427339 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RTKN
Entrez Gene ID
6242
Gene Name
rhotekin
Gene Aliases
-
UniGene
Hs.192854
Gene Chromosome Location
Chr.2: 74425861-74441933 on Build GRCh38
Transcripts
NM_001015055.1 NM_001015056.1 NM_033046.2 XM_005264478.2 XM_005264479.4 XM_017004635.1 XM_017004636.1 XM_017004637.1 XM_017004638.1 XM_017004639.1 XM_017004640.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1669075 c.1255G>T p.E419* Chr.2: 74427235-74427235 on Build GRCh38
7131789 c.1315G>A p.E439K Chr.2: 74427175-74427175 on Build GRCh38
6158846 c.1240G>A p.E414K Chr.2: 74427250-74427250 on Build GRCh38
5538421 c.1338G>A p.Q446Q Chr.2: 74427191-74427191 on Build GRCh38
5429359 c.1270C>T p.R424W Chr.2: 74427220-74427220 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745518022 Chr.2: 74427295-74427295 on Build GRCh38
rs141388964 Chr.2: 74427242-74427242 on Build GRCh38
rs750649068 Chr.2: 74427245-74427245 on Build GRCh38
rs758277869 Chr.2: 74427269-74427269 on Build GRCh38
rs760847376 Chr.2: 74427249-74427249 on Build GRCh38
rs774861501 Chr.2: 74427225-74427225 on Build GRCh38
rs759768701 Chr.2: 74427112-74427112 on Build GRCh38
rs780865315 Chr.2: 74427287-74427287 on Build GRCh38
rs758668222 Chr.2: 74427286-74427286 on Build GRCh38
rs34890445 Chr.2: 74427219-74427220 on Build GRCh38
rs202054055 Chr.2: 74427251-74427251 on Build GRCh38
rs1008530297 Chr.2: 74427149-74427149 on Build GRCh38
rs776132648 Chr.2: 74427220-74427220 on Build GRCh38
rs779605866 Chr.2: 74427294-74427295 on Build GRCh38
rs140028641 Chr.2: 74427219-74427219 on Build GRCh38
rs770605083 Chr.2: 74427142-74427142 on Build GRCh38
rs967501742 Chr.2: 74427297-74427298 on Build GRCh38
rs746743946 Chr.2: 74427132-74427132 on Build GRCh38
rs200885542 Chr.2: 74427284-74427284 on Build GRCh38
rs769405113 Chr.2: 74427307-74427307 on Build GRCh38
rs966755203 Chr.2: 74427120-74427120 on Build GRCh38
rs777414293 Chr.2: 74427119-74427119 on Build GRCh38
rs72905476 Chr.2: 74427282-74427282 on Build GRCh38
rs1018609772 Chr.2: 74427192-74427192 on Build GRCh38
rs763357914 Chr.2: 74427221-74427221 on Build GRCh38
rs564002053 Chr.2: 74427107-74427107 on Build GRCh38
rs778036244 Chr.2: 74427236-74427236 on Build GRCh38
rs901858282 Chr.2: 74427127-74427127 on Build GRCh38
rs757819819 Chr.2: 74427163-74427166 on Build GRCh38
rs776220788 Chr.2: 74427148-74427148 on Build GRCh38
rs745434630 Chr.2: 74427177-74427177 on Build GRCh38

View Full Product Details