Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61481761-61482030 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP1R32
Entrez Gene ID
220004
Gene Name
protein phosphatase 1 regulatory subunit 32
Gene Aliases
C11orf66, IIIG9
UniGene
Hs.729003
Gene Chromosome Location
Chr.11: 61481113-61490928 on Build GRCh38
Transcripts
NM_001170753.1 NM_145017.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
259706 c.53C>T p.S18L Chr.11: 61481862-61481862 on Build GRCh38
399642 c.83A>T p.Y28F Chr.11: 61481892-61481892 on Build GRCh38
6014451 c.33C>T p.S11S Chr.11: 61481842-61481842 on Build GRCh38
6754055 c.54delG p.G20fs*6 Chr.11: 61481863-61481863 on Build GRCh38
3450707 c.32C>T p.S11F Chr.11: 61481841-61481841 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866744849 Chr.11: 61481895-61481895 on Build GRCh38
rs993704179 Chr.11: 61481827-61481827 on Build GRCh38
rs776603876 Chr.11: 61481797-61481797 on Build GRCh38
rs751684876 Chr.11: 61481991-61481991 on Build GRCh38
rs754622311 Chr.11: 61481832-61481832 on Build GRCh38
rs774058760 Chr.11: 61481896-61481896 on Build GRCh38
rs200163017 Chr.11: 61481954-61481954 on Build GRCh38
rs759356584 Chr.11: 61481898-61481898 on Build GRCh38
rs748417367 Chr.11: 61481950-61481950 on Build GRCh38
rs369794949 Chr.11: 61481823-61481823 on Build GRCh38
rs201787588 Chr.11: 61481864-61481864 on Build GRCh38
rs373643914 Chr.11: 61481935-61481935 on Build GRCh38
rs763007461 Chr.11: 61481877-61481877 on Build GRCh38
rs779460276 Chr.11: 61481841-61481841 on Build GRCh38
rs765325127 Chr.11: 61481803-61481803 on Build GRCh38
rs760819783 Chr.11: 61481782-61481782 on Build GRCh38
rs773369172 Chr.11: 61481876-61481876 on Build GRCh38
rs776889574 Chr.11: 61481868-61481868 on Build GRCh38
rs759361095 Chr.11: 61481949-61481950 on Build GRCh38
rs781345578 Chr.11: 61481949-61481949 on Build GRCh38
rs755634463 Chr.11: 61481838-61481838 on Build GRCh38
rs761919694 Chr.11: 61481801-61481801 on Build GRCh38
rs992449644 Chr.11: 61481875-61481875 on Build GRCh38
rs776339792 Chr.11: 61481830-61481830 on Build GRCh38
rs763603391 Chr.11: 61481917-61481917 on Build GRCh38
rs773459561 Chr.11: 61481961-61481961 on Build GRCh38
rs746433929 Chr.11: 61481812-61481812 on Build GRCh38
rs755366471 Chr.11: 61481948-61481948 on Build GRCh38
rs140605406 Chr.11: 61481899-61481899 on Build GRCh38
rs373043146 Chr.11: 61481793-61481793 on Build GRCh38
rs73496203 Chr.11: 61481934-61481934 on Build GRCh38
rs768873743 Chr.11: 61481784-61481784 on Build GRCh38
rs925023352 Chr.11: 61481919-61481919 on Build GRCh38
rs747358982 Chr.11: 61481865-61481865 on Build GRCh38
rs751354493 Chr.11: 61481830-61481830 on Build GRCh38
rs746374003 Chr.11: 61481862-61481862 on Build GRCh38
rs372889299 Chr.11: 61481894-61481894 on Build GRCh38
rs909575641 Chr.11: 61481937-61481937 on Build GRCh38
rs376721598 Chr.11: 61481879-61481879 on Build GRCh38
rs137981780 Chr.11: 61481918-61481918 on Build GRCh38
rs375085615 Chr.11: 61481904-61481904 on Build GRCh38
rs3019200 Chr.11: 61481911-61481911 on Build GRCh38
rs1003622266 Chr.11: 61481808-61481808 on Build GRCh38
rs145779108 Chr.11: 61481874-61481874 on Build GRCh38
rs752201169 Chr.11: 61481837-61481837 on Build GRCh38
rs770867347 Chr.11: 61481823-61481823 on Build GRCh38
rs199669821 Chr.11: 61481836-61481836 on Build GRCh38
rs202015048 Chr.11: 61481863-61481863 on Build GRCh38
rs761866873 Chr.11: 61481871-61481871 on Build GRCh38
rs762917760 Chr.11: 61481806-61481806 on Build GRCh38
rs747381909 Chr.11: 61481943-61481943 on Build GRCh38
rs375465905 Chr.11: 61481804-61481804 on Build GRCh38

View Full Product Details