Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 21505649-21505894 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF429
Entrez Gene ID
353088
Gene Name
zinc finger protein 429
Gene Aliases
-
UniGene
Hs.572567
Gene Chromosome Location
Chr.19: 21505257-21541157 on Build GRCh38
Transcripts
NM_001001415.2 XM_017026748.1 XM_017026750.1 XM_017026751.1 XM_017026753.1 XM_017026754.1 XM_017026756.1 XM_017026757.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
993691 c.3+2T>C p.? Chr.19: 21505776-21505776 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs767083475 Chr.19: 21505771-21505771 on Build GRCh38
rs201745214 Chr.19: 21505778-21505778 on Build GRCh38
rs912414342 Chr.19: 21505718-21505718 on Build GRCh38
rs563480450 Chr.19: 21505700-21505700 on Build GRCh38
rs977285808 Chr.19: 21505686-21505686 on Build GRCh38
rs750047124 Chr.19: 21505775-21505775 on Build GRCh38
rs757734245 Chr.19: 21505812-21505812 on Build GRCh38
rs763388208 Chr.19: 21505815-21505815 on Build GRCh38
rs1046658160 Chr.19: 21505860-21505860 on Build GRCh38
rs906317770 Chr.19: 21505861-21505861 on Build GRCh38
rs1002349659 Chr.19: 21505876-21505876 on Build GRCh38
rs73017722 Chr.19: 21505693-21505693 on Build GRCh38
rs778977141 Chr.19: 21505797-21505797 on Build GRCh38
rs943882534 Chr.19: 21505729-21505729 on Build GRCh38
rs781466970 Chr.19: 21505806-21505806 on Build GRCh38
rs373886181 Chr.19: 21505821-21505821 on Build GRCh38
rs10420016 Chr.19: 21505800-21505800 on Build GRCh38
rs932725481 Chr.19: 21505696-21505696 on Build GRCh38
rs575546349 Chr.19: 21505739-21505739 on Build GRCh38
rs775650520 Chr.19: 21505813-21505813 on Build GRCh38
rs374436198 Chr.19: 21505755-21505755 on Build GRCh38
rs376832656 Chr.19: 21505787-21505787 on Build GRCh38
rs764665760 Chr.19: 21505756-21505756 on Build GRCh38
rs746026837 Chr.19: 21505807-21505807 on Build GRCh38
rs759875819 Chr.19: 21505748-21505748 on Build GRCh38
rs200587037 Chr.19: 21505785-21505785 on Build GRCh38
rs777728076 Chr.19: 21505800-21505801 on Build GRCh38
rs776985132 Chr.19: 21505743-21505743 on Build GRCh38
rs368655398 Chr.19: 21505774-21505774 on Build GRCh38
rs62108288 Chr.19: 21505671-21505671 on Build GRCh38
rs777446990 Chr.19: 21505709-21505709 on Build GRCh38
rs926952369 Chr.19: 21505875-21505875 on Build GRCh38
rs920014937 Chr.19: 21505695-21505695 on Build GRCh38
rs766993645 Chr.19: 21505826-21505826 on Build GRCh38
rs985267650 Chr.19: 21505697-21505697 on Build GRCh38
rs201367777 Chr.19: 21505777-21505777 on Build GRCh38
rs191177521 Chr.19: 21505768-21505768 on Build GRCh38
rs747094151 Chr.19: 21505737-21505737 on Build GRCh38
rs182604444 Chr.19: 21505788-21505788 on Build GRCh38
rs759536955 Chr.19: 21505805-21505805 on Build GRCh38
rs372665002 Chr.19: 21505757-21505757 on Build GRCh38
rs765494665 Chr.19: 21505751-21505751 on Build GRCh38
rs758491548 Chr.19: 21505759-21505760 on Build GRCh38
rs770084523 Chr.19: 21505812-21505812 on Build GRCh38
rs768960157 Chr.19: 21505819-21505819 on Build GRCh38
rs779668266 Chr.19: 21505779-21505779 on Build GRCh38
rs985505063 Chr.19: 21505694-21505694 on Build GRCh38
rs922749499 Chr.19: 21505690-21505690 on Build GRCh38
rs775944318 Chr.19: 21505754-21505754 on Build GRCh38
rs532609408 Chr.19: 21505840-21505840 on Build GRCh38
rs771101076 Chr.19: 21505805-21505805 on Build GRCh38

View Full Product Details