Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 75286541-75286800 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSHZ1
Entrez Gene ID
10194
Gene Name
teashirt zinc finger homeobox 1
Gene Aliases
CAA, NY-CO-33, SDCCAG33, TSH1
UniGene
Hs.284217
Gene Chromosome Location
Chr.18: 75210755-75289950 on Build GRCh38
Transcripts
NM_001308210.1 NM_005786.5 XM_005266641.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7172220 c.1384G>A p.E462K Chr.18: 75286791-75286791 on Build GRCh38
3718005 c.1107G>A p.Q369Q Chr.18: 75286649-75286649 on Build GRCh38
5474021 c.1162G>A p.A388T Chr.18: 75286704-75286704 on Build GRCh38
6388112 c.1308G>A p.M436I Chr.18: 75286715-75286715 on Build GRCh38
4073297 c.1238C>T p.A413V Chr.18: 75286645-75286645 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200266026 Chr.18: 75286569-75286569 on Build GRCh38
rs761292589 Chr.18: 75286727-75286727 on Build GRCh38
rs777836164 Chr.18: 75286691-75286691 on Build GRCh38
rs758861199 Chr.18: 75286690-75286690 on Build GRCh38
rs200472754 Chr.18: 75286705-75286705 on Build GRCh38
rs34016339 Chr.18: 75286604-75286605 on Build GRCh38
rs139004973 Chr.18: 75286766-75286766 on Build GRCh38
rs34095753 Chr.18: 75286571-75286571 on Build GRCh38
rs202060462 Chr.18: 75286748-75286748 on Build GRCh38
rs373193244 Chr.18: 75286646-75286646 on Build GRCh38
rs774327671 Chr.18: 75286726-75286726 on Build GRCh38
rs754088993 Chr.18: 75286579-75286579 on Build GRCh38
rs762964879 Chr.18: 75286613-75286613 on Build GRCh38
rs34416873 Chr.18: 75286580-75286580 on Build GRCh38
rs35028697 Chr.18: 75286611-75286611 on Build GRCh38
rs750082958 Chr.18: 75286675-75286675 on Build GRCh38
rs776499718 Chr.18: 75286606-75286606 on Build GRCh38
rs747287680 Chr.18: 75286598-75286598 on Build GRCh38
rs757347385 Chr.18: 75286703-75286703 on Build GRCh38
rs73484103 Chr.18: 75286721-75286721 on Build GRCh38
rs768654001 Chr.18: 75286722-75286722 on Build GRCh38
rs373817259 Chr.18: 75286704-75286704 on Build GRCh38
rs956928502 Chr.18: 75286590-75286590 on Build GRCh38
rs201275095 Chr.18: 75286776-75286776 on Build GRCh38
rs770993914 Chr.18: 75286601-75286601 on Build GRCh38
rs374001632 Chr.18: 75286643-75286643 on Build GRCh38
rs779462219 Chr.18: 75286583-75286583 on Build GRCh38
rs568803492 Chr.18: 75286747-75286747 on Build GRCh38
rs767021398 Chr.18: 75286734-75286734 on Build GRCh38
rs145008445 Chr.18: 75286575-75286575 on Build GRCh38
rs765027625 Chr.18: 75286563-75286563 on Build GRCh38
rs746102569 Chr.18: 75286775-75286775 on Build GRCh38
rs571038490 Chr.18: 75286763-75286763 on Build GRCh38
rs376781399 Chr.18: 75286574-75286574 on Build GRCh38
rs1007375979 Chr.18: 75286564-75286564 on Build GRCh38
rs997870682 Chr.18: 75286674-75286674 on Build GRCh38
rs369060962 Chr.18: 75286687-75286687 on Build GRCh38
rs376372721 Chr.18: 75286682-75286682 on Build GRCh38
rs774635790 Chr.18: 75286688-75286690 on Build GRCh38
rs143712115 Chr.18: 75286688-75286688 on Build GRCh38
rs762622920 Chr.18: 75286570-75286570 on Build GRCh38
rs759308271 Chr.18: 75286607-75286607 on Build GRCh38
rs148126168 Chr.18: 75286712-75286712 on Build GRCh38
rs748622384 Chr.18: 75286591-75286591 on Build GRCh38
rs777780760 Chr.18: 75286596-75286596 on Build GRCh38
rs772789282 Chr.18: 75286741-75286741 on Build GRCh38
rs765563523 Chr.18: 75286686-75286686 on Build GRCh38
rs1025783310 Chr.18: 75286683-75286683 on Build GRCh38
rs913098293 Chr.18: 75286779-75286779 on Build GRCh38
rs145021024 Chr.18: 75286756-75286756 on Build GRCh38
rs769711329 Chr.18: 75286610-75286610 on Build GRCh38
rs772186940 Chr.18: 75286592-75286592 on Build GRCh38
rs550963209 Chr.18: 75286754-75286754 on Build GRCh38
rs747005684 Chr.18: 75286702-75286702 on Build GRCh38
rs757324946 Chr.18: 75286765-75286765 on Build GRCh38
rs780468421 Chr.18: 75286576-75286576 on Build GRCh38
rs767129921 Chr.18: 75286655-75286655 on Build GRCh38
rs1018481672 Chr.18: 75286565-75286565 on Build GRCh38
rs148944812 Chr.18: 75286638-75286638 on Build GRCh38

View Full Product Details