Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4048074-4048347 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZBTB7A
Entrez Gene ID
51341
Gene Name
zinc finger and BTB domain containing 7A
Gene Aliases
FBI-1, FBI1, LRF, TIP21, ZBTB7, ZNF857A, pokemon
UniGene
Hs.591384
Gene Chromosome Location
Chr.19: 4043304-4066945 on Build GRCh38
Transcripts
NM_001317990.1 NM_015898.3 XM_005259570.4 XM_005259571.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6825465 c.1405G>A p.D469N Chr.19: 4048102-4048102 on Build GRCh38
3771072 c.1372C>T p.R458C Chr.19: 4048135-4048135 on Build GRCh38
6825464 c.1331G>C p.G444A Chr.19: 4048176-4048176 on Build GRCh38
474730 c.1272G>T p.K424N Chr.19: 4048235-4048235 on Build GRCh38
3787667 c.1366C>T p.H456Y Chr.19: 4048141-4048141 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375386703 Chr.19: 4048286-4048286 on Build GRCh38
rs867835425 Chr.19: 4048228-4048228 on Build GRCh38
rs759013962 Chr.19: 4048279-4048279 on Build GRCh38
rs745338351 Chr.19: 4048252-4048252 on Build GRCh38
rs951017837 Chr.19: 4048316-4048316 on Build GRCh38
rs770961493 Chr.19: 4048103-4048103 on Build GRCh38
rs773100200 Chr.19: 4048092-4048092 on Build GRCh38
rs762241835 Chr.19: 4048264-4048264 on Build GRCh38
rs997250304 Chr.19: 4048303-4048303 on Build GRCh38
rs760365409 Chr.19: 4048148-4048148 on Build GRCh38
rs770709830 Chr.19: 4048241-4048241 on Build GRCh38
rs747144020 Chr.19: 4048289-4048289 on Build GRCh38
rs777418830 Chr.19: 4048233-4048233 on Build GRCh38
rs192146606 Chr.19: 4048106-4048106 on Build GRCh38
rs937626503 Chr.19: 4048133-4048133 on Build GRCh38
rs771545015 Chr.19: 4048253-4048253 on Build GRCh38
rs775150177 Chr.19: 4048139-4048139 on Build GRCh38
rs757689355 Chr.19: 4048175-4048175 on Build GRCh38
rs758581721 Chr.19: 4048196-4048196 on Build GRCh38
rs772031516 Chr.19: 4048252-4048253 on Build GRCh38
rs1010453260 Chr.19: 4048194-4048194 on Build GRCh38
rs765621953 Chr.19: 4048267-4048267 on Build GRCh38
rs764330841 Chr.19: 4048150-4048150 on Build GRCh38
rs754292739 Chr.19: 4048166-4048166 on Build GRCh38
rs747204568 Chr.19: 4048208-4048208 on Build GRCh38
rs774306774 Chr.19: 4048243-4048243 on Build GRCh38
rs763863211 Chr.19: 4048259-4048259 on Build GRCh38
rs1014400556 Chr.19: 4048311-4048311 on Build GRCh38
rs760314143 Chr.19: 4048256-4048256 on Build GRCh38
rs932030722 Chr.19: 4048165-4048165 on Build GRCh38
rs982734923 Chr.19: 4048317-4048317 on Build GRCh38
rs781483584 Chr.19: 4048293-4048293 on Build GRCh38
rs549149431 Chr.19: 4048202-4048202 on Build GRCh38
rs1023982459 Chr.19: 4048313-4048313 on Build GRCh38
rs765785189 Chr.19: 4048178-4048178 on Build GRCh38
rs1051733315 Chr.19: 4048174-4048174 on Build GRCh38
rs199761446 Chr.19: 4048226-4048226 on Build GRCh38
rs759055137 Chr.19: 4048124-4048124 on Build GRCh38
rs958497232 Chr.19: 4048308-4048308 on Build GRCh38
rs374870281 Chr.19: 4048288-4048288 on Build GRCh38
rs750989184 Chr.19: 4048276-4048276 on Build GRCh38
rs1000352683 Chr.19: 4048300-4048300 on Build GRCh38
rs777803398 Chr.19: 4048278-4048278 on Build GRCh38
rs529208407 Chr.19: 4048312-4048312 on Build GRCh38
rs1034544725 Chr.19: 4048304-4048304 on Build GRCh38
rs749178819 Chr.19: 4048094-4048094 on Build GRCh38
rs767120363 Chr.19: 4048127-4048127 on Build GRCh38
rs144598692 Chr.19: 4048190-4048190 on Build GRCh38
rs766563768 Chr.19: 4048285-4048285 on Build GRCh38
rs749123726 Chr.19: 4048239-4048239 on Build GRCh38
rs751747728 Chr.19: 4048287-4048287 on Build GRCh38
rs1041870564 Chr.19: 4048205-4048205 on Build GRCh38

View Full Product Details