Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 680547-680772 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRKAR1B
Entrez Gene ID
5575
Gene Name
protein kinase cAMP-dependent type I regulatory subunit beta
Gene Aliases
PRKAR1
UniGene
Hs.520851
Gene Chromosome Location
Chr.7: 549185-727676 on Build GRCh38
Transcripts
NM_001164758.1 NM_001164759.1 NM_001164760.1 NM_001164761.1 NM_001164762.1 NM_002735.2 XM_011515446.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
291067 c.337T>C p.Y113H Chr.7: 680567-680567 on Build GRCh38
6755709 c.203G>A p.R68Q Chr.7: 680701-680701 on Build GRCh38
5098298 c.307G>A p.E103K Chr.7: 680597-680597 on Build GRCh38
3785759 c.286C>T p.R96W Chr.7: 680618-680618 on Build GRCh38
6755706 c.244G>A p.V82M Chr.7: 680660-680660 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764069617 Chr.7: 680643-680644 on Build GRCh38
rs770716715 Chr.7: 680644-680644 on Build GRCh38
rs774298132 Chr.7: 680656-680656 on Build GRCh38
rs1026143498 Chr.7: 680625-680625 on Build GRCh38
rs370909605 Chr.7: 680702-680702 on Build GRCh38
rs116198569 Chr.7: 680729-680729 on Build GRCh38
rs367822885 Chr.7: 680671-680671 on Build GRCh38
rs776128488 Chr.7: 680652-680652 on Build GRCh38
rs200458521 Chr.7: 680637-680637 on Build GRCh38
rs199568828 Chr.7: 680646-680646 on Build GRCh38
rs762351800 Chr.7: 680624-680624 on Build GRCh38
rs778958450 Chr.7: 680623-680623 on Build GRCh38
rs376698663 Chr.7: 680734-680734 on Build GRCh38
rs947912556 Chr.7: 680716-680716 on Build GRCh38
rs372164270 Chr.7: 680732-680732 on Build GRCh38
rs906632600 Chr.7: 680589-680589 on Build GRCh38
rs140110814 Chr.7: 680701-680701 on Build GRCh38
rs767658145 Chr.7: 680617-680617 on Build GRCh38
rs200749428 Chr.7: 680653-680653 on Build GRCh38
rs1053414449 Chr.7: 680683-680683 on Build GRCh38
rs1005927520 Chr.7: 680642-680643 on Build GRCh38
rs964235668 Chr.7: 680644-680644 on Build GRCh38
rs1018426691 Chr.7: 680654-680654 on Build GRCh38
rs142537452 Chr.7: 680655-680655 on Build GRCh38
rs530700943 Chr.7: 680710-680710 on Build GRCh38
rs144462255 Chr.7: 680620-680620 on Build GRCh38
rs781027465 Chr.7: 680691-680691 on Build GRCh38
rs369835648 Chr.7: 680568-680568 on Build GRCh38
rs369695106 Chr.7: 680740-680740 on Build GRCh38
rs1021923442 Chr.7: 680670-680670 on Build GRCh38
rs758284378 Chr.7: 680680-680680 on Build GRCh38
rs1047146509 Chr.7: 680730-680730 on Build GRCh38
rs1044891046 Chr.7: 680575-680575 on Build GRCh38
rs374177156 Chr.7: 680607-680607 on Build GRCh38
rs777702342 Chr.7: 680687-680687 on Build GRCh38
rs770020244 Chr.7: 680650-680650 on Build GRCh38
rs142687236 Chr.7: 680586-680586 on Build GRCh38
rs148427628 Chr.7: 680639-680639 on Build GRCh38
rs745507312 Chr.7: 680697-680697 on Build GRCh38
rs761841270 Chr.7: 680658-680658 on Build GRCh38
rs757483222 Chr.7: 680609-680609 on Build GRCh38
rs1026074487 Chr.7: 680634-680634 on Build GRCh38
rs763982944 Chr.7: 680735-680735 on Build GRCh38
rs538195666 Chr.7: 680574-680574 on Build GRCh38
rs764939348 Chr.7: 680679-680679 on Build GRCh38
rs199799634 Chr.7: 680645-680645 on Build GRCh38
rs751507796 Chr.7: 680739-680739 on Build GRCh38
rs772109340 Chr.7: 680714-680714 on Build GRCh38
rs1003652118 Chr.7: 680613-680613 on Build GRCh38
rs756892034 Chr.7: 680743-680743 on Build GRCh38
rs780833197 Chr.7: 680749-680749 on Build GRCh38
rs371160585 Chr.7: 680719-680719 on Build GRCh38
rs372454246 Chr.7: 680621-680621 on Build GRCh38
rs764074623 Chr.7: 680606-680606 on Build GRCh38
rs147480983 Chr.7: 680703-680703 on Build GRCh38
rs374729888 Chr.7: 680721-680721 on Build GRCh38
rs547133945 Chr.7: 680744-680745 on Build GRCh38
rs558009253 Chr.7: 680577-680577 on Build GRCh38
rs183383238 Chr.7: 680704-680704 on Build GRCh38
rs774195498 Chr.7: 680739-680739 on Build GRCh38
rs369593324 Chr.7: 680747-680748 on Build GRCh38
rs545804273 Chr.7: 680643-680643 on Build GRCh38
rs369017795 Chr.7: 680677-680677 on Build GRCh38
rs201442077 Chr.7: 680689-680689 on Build GRCh38
rs192066279 Chr.7: 680565-680565 on Build GRCh38
rs143819086 Chr.7: 680712-680712 on Build GRCh38
rs763130825 Chr.7: 680598-680598 on Build GRCh38
rs150538055 Chr.7: 680694-680694 on Build GRCh38

View Full Product Details