Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 102317117-102317378 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SH2B2
Entrez Gene ID
10603
Gene Name
SH2B adaptor protein 2
Gene Aliases
APS
UniGene
Hs.489448
Gene Chromosome Location
Chr.7: 102284526-102321711 on Build GRCh38
Transcripts
NM_020979.4 XM_005276975.4 XM_005276976.3 XM_005276977.4 XM_005276979.3 XM_011515740.2 XM_017011680.1 XM_017011681.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5744955 c.1345C>T p.L449L Chr.7: 102317234-102317234 on Build GRCh38
3083370 c.1297delG p.R436fs*7 Chr.7: 102317300-102317300 on Build GRCh38
6175352 c.1354G>A p.E452K Chr.7: 102317357-102317357 on Build GRCh38
6175354 c.1333C>T p.Q445* Chr.7: 102317336-102317336 on Build GRCh38
4949499 c.1342G>C p.E448Q Chr.7: 102317231-102317231 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781947704 Chr.7: 102317270-102317270 on Build GRCh38
rs782256668 Chr.7: 102317306-102317306 on Build GRCh38
rs782717144 Chr.7: 102317235-102317235 on Build GRCh38
rs782447117 Chr.7: 102317303-102317303 on Build GRCh38
rs782398817 Chr.7: 102317309-102317309 on Build GRCh38
rs782395940 Chr.7: 102317329-102317329 on Build GRCh38
rs369927479 Chr.7: 102317161-102317161 on Build GRCh38
rs782323572 Chr.7: 102317268-102317268 on Build GRCh38
rs574609362 Chr.7: 102317206-102317206 on Build GRCh38
rs577028578 Chr.7: 102317173-102317173 on Build GRCh38
rs782181858 Chr.7: 102317311-102317311 on Build GRCh38
rs9691917 Chr.7: 102317151-102317151 on Build GRCh38
rs921762851 Chr.7: 102317354-102317354 on Build GRCh38
rs782751129 Chr.7: 102317298-102317298 on Build GRCh38
rs200961263 Chr.7: 102317205-102317205 on Build GRCh38
rs782085314 Chr.7: 102317297-102317297 on Build GRCh38
rs781953796 Chr.7: 102317305-102317306 on Build GRCh38
rs782143454 Chr.7: 102317282-102317282 on Build GRCh38
rs189154913 Chr.7: 102317349-102317349 on Build GRCh38
rs781891011 Chr.7: 102317224-102317224 on Build GRCh38
rs781996415 Chr.7: 102317279-102317279 on Build GRCh38
rs13244392 Chr.7: 102317333-102317333 on Build GRCh38
rs782809706 Chr.7: 102317286-102317286 on Build GRCh38
rs782076378 Chr.7: 102317342-102317342 on Build GRCh38
rs373079904 Chr.7: 102317169-102317169 on Build GRCh38
rs782777792 Chr.7: 102317213-102317213 on Build GRCh38
rs781822209 Chr.7: 102317239-102317239 on Build GRCh38
rs782579624 Chr.7: 102317304-102317304 on Build GRCh38
rs782109422 Chr.7: 102317211-102317211 on Build GRCh38
rs782289208 Chr.7: 102317191-102317191 on Build GRCh38
rs557329329 Chr.7: 102317240-102317240 on Build GRCh38
rs781922810 Chr.7: 102317318-102317318 on Build GRCh38
rs782628402 Chr.7: 102317244-102317244 on Build GRCh38
rs782264863 Chr.7: 102317248-102317248 on Build GRCh38
rs781849345 Chr.7: 102317352-102317352 on Build GRCh38
rs782120964 Chr.7: 102317327-102317327 on Build GRCh38
rs534773193 Chr.7: 102317198-102317198 on Build GRCh38
rs781983972 Chr.7: 102317340-102317340 on Build GRCh38
rs782191132 Chr.7: 102317262-102317262 on Build GRCh38
rs782410817 Chr.7: 102317150-102317151 on Build GRCh38
rs1056554988 Chr.7: 102317219-102317219 on Build GRCh38
rs782613440 Chr.7: 102317152-102317152 on Build GRCh38
rs781935715 Chr.7: 102317291-102317291 on Build GRCh38
rs782484238 Chr.7: 102317305-102317305 on Build GRCh38
rs781856825 Chr.7: 102317299-102317299 on Build GRCh38
rs369220336 Chr.7: 102317225-102317225 on Build GRCh38
rs782696384 Chr.7: 102317334-102317334 on Build GRCh38
rs781908952 Chr.7: 102317302-102317302 on Build GRCh38
rs373360709 Chr.7: 102317271-102317271 on Build GRCh38
rs546518657 Chr.7: 102317317-102317317 on Build GRCh38
rs370458582 Chr.7: 102317310-102317310 on Build GRCh38
rs782690995 Chr.7: 102317258-102317258 on Build GRCh38
rs782364656 Chr.7: 102317299-102317300 on Build GRCh38
rs955674274 Chr.7: 102317280-102317280 on Build GRCh38
rs782541583 Chr.7: 102317257-102317257 on Build GRCh38
rs376043619 Chr.7: 102317301-102317301 on Build GRCh38
rs782326052 Chr.7: 102317277-102317277 on Build GRCh38
rs781811152 Chr.7: 102317339-102317339 on Build GRCh38
rs781964919 Chr.7: 102317209-102317209 on Build GRCh38

View Full Product Details