Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 140335137-140335388 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC37A3
Entrez Gene ID
84255
Gene Name
solute carrier family 37 member 3
Gene Aliases
-
UniGene
Hs.446021
Gene Chromosome Location
Chr.7: 140333752-140404352 on Build GRCh38
Transcripts
NM_001287498.1 NM_032295.3 NM_207113.2 XM_011516626.2 XM_011516627.2 XM_017012712.1 XM_017012713.1 XM_017012714.1 XM_017012715.1 XM_017012716.1 XM_017012717.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
315315 c.1298C>T p.A433V Chr.7: 140335297-140335297 on Build GRCh38
7241566 c.1243G>T p.V415F Chr.7: 140335352-140335352 on Build GRCh38
1086209 c.1229C>T p.P410L Chr.7: 140335366-140335366 on Build GRCh38
5406666 c.1240C>T p.R414* Chr.7: 140335355-140335355 on Build GRCh38
6471689 c.1276A>C p.S426R Chr.7: 140335319-140335319 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765446872 Chr.7: 140335343-140335344 on Build GRCh38
rs754332025 Chr.7: 140335328-140335328 on Build GRCh38
rs147157364 Chr.7: 140335324-140335324 on Build GRCh38
rs772414538 Chr.7: 140335352-140335352 on Build GRCh38
rs894024002 Chr.7: 140335169-140335169 on Build GRCh38
rs372795507 Chr.7: 140335355-140335355 on Build GRCh38
rs1011123926 Chr.7: 140335182-140335182 on Build GRCh38
rs759829828 Chr.7: 140335251-140335257 on Build GRCh38
rs768941551 Chr.7: 140335233-140335233 on Build GRCh38
rs755669142 Chr.7: 140335301-140335301 on Build GRCh38
rs927907116 Chr.7: 140335275-140335275 on Build GRCh38
rs761460612 Chr.7: 140335297-140335297 on Build GRCh38
rs540545215 Chr.7: 140335224-140335224 on Build GRCh38
rs772217351 Chr.7: 140335249-140335249 on Build GRCh38
rs749641753 Chr.7: 140335232-140335232 on Build GRCh38
rs983422087 Chr.7: 140335326-140335326 on Build GRCh38
rs966442739 Chr.7: 140335201-140335201 on Build GRCh38
rs951908074 Chr.7: 140335325-140335325 on Build GRCh38
rs749922145 Chr.7: 140335300-140335300 on Build GRCh38
rs762771474 Chr.7: 140335287-140335287 on Build GRCh38
rs543710643 Chr.7: 140335162-140335162 on Build GRCh38
rs868268355 Chr.7: 140335342-140335343 on Build GRCh38
rs747057672 Chr.7: 140335354-140335354 on Build GRCh38
rs868723789 Chr.7: 140335168-140335168 on Build GRCh38
rs140410401 Chr.7: 140335345-140335345 on Build GRCh38
rs371842286 Chr.7: 140335265-140335265 on Build GRCh38
rs763819539 Chr.7: 140335290-140335290 on Build GRCh38
rs767224731 Chr.7: 140335298-140335298 on Build GRCh38
rs982020740 Chr.7: 140335267-140335267 on Build GRCh38
rs756609802 Chr.7: 140335230-140335230 on Build GRCh38
rs551341534 Chr.7: 140335319-140335319 on Build GRCh38
rs890064837 Chr.7: 140335366-140335366 on Build GRCh38
rs1021034877 Chr.7: 140335192-140335192 on Build GRCh38
rs769836773 Chr.7: 140335279-140335279 on Build GRCh38
rs528778661 Chr.7: 140335184-140335184 on Build GRCh38
rs773426595 Chr.7: 140335261-140335261 on Build GRCh38
rs533055947 Chr.7: 140335318-140335318 on Build GRCh38
rs143648898 Chr.7: 140335296-140335296 on Build GRCh38
rs779156476 Chr.7: 140335336-140335336 on Build GRCh38
rs187867019 Chr.7: 140335231-140335231 on Build GRCh38
rs749199198 Chr.7: 140335365-140335365 on Build GRCh38
rs748338680 Chr.7: 140335245-140335245 on Build GRCh38
rs559964305 Chr.7: 140335333-140335333 on Build GRCh38
rs775333555 Chr.7: 140335284-140335284 on Build GRCh38
rs199678966 Chr.7: 140335236-140335236 on Build GRCh38
rs775901769 Chr.7: 140335213-140335213 on Build GRCh38
rs775451723 Chr.7: 140335361-140335361 on Build GRCh38

View Full Product Details