Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 64156022-64156257 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPIB
Entrez Gene ID
5479
Gene Name
peptidylprolyl isomerase B
Gene Aliases
CYP-S1, CYPB, HEL-S-39, OI9, SCYLP
UniGene
Hs.434937
Gene Chromosome Location
Chr.15: 64155815-64163155 on Build GRCh38
Transcripts
NM_000942.4 NM_024798.2 NR_073534.1 XM_017022581.1

Gene Symbol
SNX22
Entrez Gene ID
79856
Gene Name
sorting nexin 22
Gene Aliases
-
UniGene
Hs.744250
Gene Chromosome Location
Chr.15: 64151698-64157481 on Build GRCh38
Transcripts
NM_000942.4 NM_024798.2 NR_073534.1 XM_017022581.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
963914 c.569G>A p.R190Q Chr.15: 64156105-64156105 on Build GRCh38
1659637 c.630C>A p.P210P Chr.15: 64156044-64156044 on Build GRCh38
5971495 c.640G>A p.A214T Chr.15: 64156034-64156034 on Build GRCh38
7245487 c.612G>C p.K204N Chr.15: 64156062-64156062 on Build GRCh38
470950 c.605G>A p.C202Y Chr.15: 64156069-64156069 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760065852 Chr.15: 64156135-64156135 on Build GRCh38
rs779373821 Chr.15: 64156197-64156197 on Build GRCh38
rs749670269 Chr.15: 64156108-64156108 on Build GRCh38
rs754080129 Chr.15: 64156165-64156165 on Build GRCh38
rs746706260 Chr.15: 64156100-64156100 on Build GRCh38
rs756245364 Chr.15: 64156048-64156048 on Build GRCh38
rs200864554 Chr.15: 64156105-64156105 on Build GRCh38
rs751075971 Chr.15: 64156177-64156177 on Build GRCh38
rs1804427 Chr.15: 64156054-64156054 on Build GRCh38
rs779934719 Chr.15: 64156176-64156176 on Build GRCh38
rs143287832 Chr.15: 64156068-64156068 on Build GRCh38
rs767732148 Chr.15: 64156136-64156136 on Build GRCh38
rs758291391 Chr.15: 64156175-64156175 on Build GRCh38
rs369181494 Chr.15: 64156196-64156196 on Build GRCh38
rs757054086 Chr.15: 64156067-64156067 on Build GRCh38
rs989724311 Chr.15: 64156159-64156159 on Build GRCh38
rs369889089 Chr.15: 64156077-64156077 on Build GRCh38
rs750380543 Chr.15: 64156172-64156172 on Build GRCh38
rs775675512 Chr.15: 64156138-64156138 on Build GRCh38
rs779488612 Chr.15: 64156095-64156095 on Build GRCh38
rs554627987 Chr.15: 64156209-64156209 on Build GRCh38
rs948001783 Chr.15: 64156168-64156168 on Build GRCh38
rs752983598 Chr.15: 64156040-64156040 on Build GRCh38
rs377032173 Chr.15: 64156169-64156169 on Build GRCh38
rs530126062 Chr.15: 64156094-64156094 on Build GRCh38
rs886972785 Chr.15: 64156233-64156233 on Build GRCh38
rs776320521 Chr.15: 64156106-64156106 on Build GRCh38
rs191357754 Chr.15: 64156218-64156218 on Build GRCh38
rs1029326377 Chr.15: 64156070-64156070 on Build GRCh38
rs1007167118 Chr.15: 64156072-64156072 on Build GRCh38
rs775716090 Chr.15: 64156049-64156049 on Build GRCh38
rs780772293 Chr.15: 64156193-64156193 on Build GRCh38
rs909768328 Chr.15: 64156203-64156203 on Build GRCh38
rs764273426 Chr.15: 64156164-64156164 on Build GRCh38
rs2253557 Chr.15: 64156166-64156166 on Build GRCh38
rs879243616 Chr.15: 64156149-64156149 on Build GRCh38
rs137853869 Chr.15: 64156115-64156118 on Build GRCh38
rs398122834 Chr.15: 64156108-64156111 on Build GRCh38
rs112393084 Chr.15: 64156147-64156147 on Build GRCh38
rs763983999 Chr.15: 64156058-64156058 on Build GRCh38
rs760777976 Chr.15: 64156145-64156145 on Build GRCh38
rs754493288 Chr.15: 64156191-64156191 on Build GRCh38
rs369952028 Chr.15: 64156126-64156126 on Build GRCh38
rs775006610 Chr.15: 64156131-64156131 on Build GRCh38
rs878958952 Chr.15: 64156146-64156147 on Build GRCh38
rs200904948 Chr.15: 64156046-64156046 on Build GRCh38
rs753616327 Chr.15: 64156060-64156060 on Build GRCh38

View Full Product Details