Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 107070196-107070443 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRKAR2B
Entrez Gene ID
5577
Gene Name
protein kinase cAMP-dependent type II regulatory subunit beta
Gene Aliases
PRKAR2, RII-BETA
UniGene
Hs.433068
Gene Chromosome Location
Chr.7: 107044728-107161811 on Build GRCh38
Transcripts
NM_002736.2 XM_011516398.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3922436 c.323G>A p.R108Q Chr.7: 107070296-107070296 on Build GRCh38
3254640 c.343G>T p.V115L Chr.7: 107070316-107070316 on Build GRCh38
288685 c.335G>A p.R112H Chr.7: 107070308-107070308 on Build GRCh38
7424285 c.317T>C p.I106T Chr.7: 107070290-107070290 on Build GRCh38
3631756 c.325T>C p.F109L Chr.7: 107070298-107070298 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772833439 Chr.7: 107070277-107070277 on Build GRCh38
rs952109955 Chr.7: 107070223-107070223 on Build GRCh38
rs937479213 Chr.7: 107070383-107070383 on Build GRCh38
rs75861058 Chr.7: 107070394-107070394 on Build GRCh38
rs777106107 Chr.7: 107070329-107070329 on Build GRCh38
rs765365068 Chr.7: 107070348-107070348 on Build GRCh38
rs766361113 Chr.7: 107070367-107070367 on Build GRCh38
rs201718858 Chr.7: 107070256-107070256 on Build GRCh38
rs750529814 Chr.7: 107070363-107070363 on Build GRCh38
rs141271607 Chr.7: 107070389-107070389 on Build GRCh38
rs1056079582 Chr.7: 107070385-107070385 on Build GRCh38
rs746851645 Chr.7: 107070257-107070257 on Build GRCh38
rs985179902 Chr.7: 107070239-107070239 on Build GRCh38
rs368481575 Chr.7: 107070260-107070260 on Build GRCh38
rs769429297 Chr.7: 107070274-107070274 on Build GRCh38
rs1041769351 Chr.7: 107070405-107070405 on Build GRCh38
rs3729869 Chr.7: 107070321-107070321 on Build GRCh38
rs542129628 Chr.7: 107070336-107070336 on Build GRCh38
rs780941061 Chr.7: 107070265-107070265 on Build GRCh38
rs748684280 Chr.7: 107070289-107070289 on Build GRCh38
rs747836733 Chr.7: 107070268-107070268 on Build GRCh38
rs928823801 Chr.7: 107070334-107070334 on Build GRCh38
rs572666860 Chr.7: 107070324-107070324 on Build GRCh38
rs1013457097 Chr.7: 107070231-107070231 on Build GRCh38
rs1022957525 Chr.7: 107070241-107070241 on Build GRCh38
rs772633149 Chr.7: 107070291-107070291 on Build GRCh38
rs758369918 Chr.7: 107070364-107070364 on Build GRCh38
rs758420032 Chr.7: 107070247-107070247 on Build GRCh38
rs751988693 Chr.7: 107070363-107070363 on Build GRCh38
rs775824400 Chr.7: 107070296-107070296 on Build GRCh38
rs1034952052 Chr.7: 107070302-107070302 on Build GRCh38

View Full Product Details