Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 42864467-42864710 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACKR2
Entrez Gene ID
1238
Gene Name
atypical chemokine receptor 2
Gene Aliases
CCBP2, CCR10, CCR9, CMKBR9, D6, hD6
UniGene
Hs.146346
Gene Chromosome Location
Chr.3: 42809472-42867283 on Build GRCh38
Transcripts
NM_001296.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3365182 c.63C>G p.S21R Chr.3: 42864565-42864565 on Build GRCh38
3736319 c.122T>C p.V41A Chr.3: 42864624-42864624 on Build GRCh38
420004 c.31G>A p.A11T Chr.3: 42864533-42864533 on Build GRCh38
7230979 c.27A>T p.P9P Chr.3: 42864529-42864529 on Build GRCh38
3594095 c.115G>A p.D39N Chr.3: 42864617-42864617 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760762059 Chr.3: 42864495-42864495 on Build GRCh38
rs564198715 Chr.3: 42864683-42864683 on Build GRCh38
rs780915000 Chr.3: 42864529-42864529 on Build GRCh38
rs538042992 Chr.3: 42864526-42864526 on Build GRCh38
rs770661964 Chr.3: 42864580-42864580 on Build GRCh38
rs574627315 Chr.3: 42864653-42864653 on Build GRCh38
rs764204521 Chr.3: 42864501-42864501 on Build GRCh38
rs753888314 Chr.3: 42864504-42864504 on Build GRCh38
rs777309929 Chr.3: 42864689-42864689 on Build GRCh38
rs755607839 Chr.3: 42864688-42864688 on Build GRCh38
rs761897179 Chr.3: 42864613-42864613 on Build GRCh38
rs750619942 Chr.3: 42864631-42864631 on Build GRCh38
rs747470333 Chr.3: 42864523-42864523 on Build GRCh38
rs376019929 Chr.3: 42864649-42864649 on Build GRCh38
rs755600354 Chr.3: 42864660-42864660 on Build GRCh38
rs942599460 Chr.3: 42864525-42864525 on Build GRCh38
rs550370044 Chr.3: 42864524-42864524 on Build GRCh38
rs2228467 Chr.3: 42864624-42864624 on Build GRCh38
rs762684097 Chr.3: 42864559-42864559 on Build GRCh38
rs781740560 Chr.3: 42864664-42864664 on Build GRCh38
rs1048207322 Chr.3: 42864498-42864498 on Build GRCh38
rs747681282 Chr.3: 42864538-42864538 on Build GRCh38
rs373009278 Chr.3: 42864645-42864645 on Build GRCh38
rs750769083 Chr.3: 42864517-42864517 on Build GRCh38
rs748608775 Chr.3: 42864599-42864599 on Build GRCh38
rs769342379 Chr.3: 42864547-42864547 on Build GRCh38
rs758723301 Chr.3: 42864518-42864518 on Build GRCh38
rs774106382 Chr.3: 42864590-42864590 on Build GRCh38
rs757492481 Chr.3: 42864509-42864509 on Build GRCh38
rs940106677 Chr.3: 42864628-42864628 on Build GRCh38
rs780739221 Chr.3: 42864522-42864522 on Build GRCh38
rs772853632 Chr.3: 42864555-42864555 on Build GRCh38
rs779184291 Chr.3: 42864514-42864514 on Build GRCh38
rs930751389 Chr.3: 42864488-42864488 on Build GRCh38
rs758713198 Chr.3: 42864637-42864637 on Build GRCh38
rs113132811 Chr.3: 42864597-42864597 on Build GRCh38

View Full Product Details