Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 31397521-31397780 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGAD
Entrez Gene ID
3681
Gene Name
integrin subunit alpha D
Gene Aliases
ADB2, CD11D
UniGene
Hs.679163
Gene Chromosome Location
Chr.16: 31393224-31426513 on Build GRCh38
Transcripts
NM_001318185.1 NM_005353.2 XM_011545835.2 XM_011545836.2 XM_011545837.2 XM_011545838.2 XM_011545839.2 XM_011545841.2 XM_011545842.2 XM_011545843.2 XM_011545844.2 XM_011545845.2 XM_011545846.2 XM_011545847.2 XM_011545848.2 XM_017023215.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
557672 c.245G>A p.R82H Chr.16: 31397599-31397599 on Build GRCh38
6688040 c.256G>A p.V86M Chr.16: 31397610-31397610 on Build GRCh38
4579000 c.296A>G p.N99S Chr.16: 31397650-31397650 on Build GRCh38
1740268 c.242-1G>A p.? Chr.16: 31397595-31397595 on Build GRCh38
2918096 c.286G>A p.A96T Chr.16: 31397640-31397640 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147897710 Chr.16: 31397633-31397633 on Build GRCh38
rs375108274 Chr.16: 31397717-31397717 on Build GRCh38
rs898411953 Chr.16: 31397757-31397757 on Build GRCh38
rs746752888 Chr.16: 31397627-31397627 on Build GRCh38
rs976487968 Chr.16: 31397597-31397597 on Build GRCh38
rs768454274 Chr.16: 31397632-31397632 on Build GRCh38
rs575585724 Chr.16: 31397703-31397703 on Build GRCh38
rs754064352 Chr.16: 31397659-31397659 on Build GRCh38
rs776228323 Chr.16: 31397574-31397574 on Build GRCh38
rs780099407 Chr.16: 31397687-31397687 on Build GRCh38
rs200546283 Chr.16: 31397577-31397577 on Build GRCh38
rs915173260 Chr.16: 31397622-31397622 on Build GRCh38
rs763954283 Chr.16: 31397693-31397696 on Build GRCh38
rs766915137 Chr.16: 31397576-31397576 on Build GRCh38
rs979678571 Chr.16: 31397586-31397586 on Build GRCh38
rs772049300 Chr.16: 31397547-31397547 on Build GRCh38
rs777928308 Chr.16: 31397598-31397598 on Build GRCh38
rs775473424 Chr.16: 31397621-31397621 on Build GRCh38
rs112720706 Chr.16: 31397573-31397573 on Build GRCh38
rs772777944 Chr.16: 31397640-31397640 on Build GRCh38
rs888718269 Chr.16: 31397726-31397726 on Build GRCh38
rs752186760 Chr.16: 31397689-31397689 on Build GRCh38
rs367685485 Chr.16: 31397636-31397636 on Build GRCh38
rs761400469 Chr.16: 31397634-31397634 on Build GRCh38
rs34475127 Chr.16: 31397688-31397689 on Build GRCh38
rs762406018 Chr.16: 31397696-31397696 on Build GRCh38
rs745937843 Chr.16: 31397676-31397676 on Build GRCh38
rs756401257 Chr.16: 31397653-31397653 on Build GRCh38
rs749387931 Chr.16: 31397599-31397599 on Build GRCh38
rs144758242 Chr.16: 31397615-31397615 on Build GRCh38
rs763816898 Chr.16: 31397569-31397569 on Build GRCh38
rs753764970 Chr.16: 31397696-31397697 on Build GRCh38
rs759241127 Chr.16: 31397687-31397688 on Build GRCh38
rs371949817 Chr.16: 31397658-31397658 on Build GRCh38
rs772568237 Chr.16: 31397701-31397701 on Build GRCh38
rs541187532 Chr.16: 31397706-31397706 on Build GRCh38
rs555808092 Chr.16: 31397691-31397691 on Build GRCh38
rs201719522 Chr.16: 31397650-31397650 on Build GRCh38
rs760079439 Chr.16: 31397645-31397645 on Build GRCh38
rs370572657 Chr.16: 31397561-31397561 on Build GRCh38
rs772650486 Chr.16: 31397694-31397694 on Build GRCh38
rs945403853 Chr.16: 31397580-31397580 on Build GRCh38
rs375873553 Chr.16: 31397556-31397556 on Build GRCh38
rs961456140 Chr.16: 31397596-31397596 on Build GRCh38
rs549367422 Chr.16: 31397552-31397552 on Build GRCh38
rs374878446 Chr.16: 31397590-31397590 on Build GRCh38
rs772104513 Chr.16: 31397616-31397616 on Build GRCh38
rs1005797647 Chr.16: 31397735-31397735 on Build GRCh38
rs750563076 Chr.16: 31397758-31397758 on Build GRCh38
rs975673541 Chr.16: 31397555-31397555 on Build GRCh38
rs767940485 Chr.16: 31397578-31397578 on Build GRCh38
rs113303483 Chr.16: 31397584-31397584 on Build GRCh38
rs560275698 Chr.16: 31397737-31397737 on Build GRCh38
rs765450729 Chr.16: 31397713-31397713 on Build GRCh38
rs202059105 Chr.16: 31397693-31397693 on Build GRCh38
rs371570127 Chr.16: 31397651-31397651 on Build GRCh38
rs754198525 Chr.16: 31397712-31397712 on Build GRCh38
rs1055709506 Chr.16: 31397609-31397609 on Build GRCh38
rs778900238 Chr.16: 31397672-31397672 on Build GRCh38
rs199843774 Chr.16: 31397579-31397579 on Build GRCh38
rs757495174 Chr.16: 31397660-31397660 on Build GRCh38
rs71151484 Chr.16: 31397695-31397696 on Build GRCh38
rs373163507 Chr.16: 31397546-31397546 on Build GRCh38
rs566170081 Chr.16: 31397610-31397610 on Build GRCh38
rs537397657 Chr.16: 31397679-31397679 on Build GRCh38
rs746806292 Chr.16: 31397689-31397689 on Build GRCh38
rs764431196 Chr.16: 31397707-31397707 on Build GRCh38

View Full Product Details