Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 132485524-132485795 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRF1
Entrez Gene ID
3659
Gene Name
interferon regulatory factor 1
Gene Aliases
IRF-1, MAR
UniGene
Hs.436061
Gene Chromosome Location
Chr.5: 132481609-132490789 on Build GRCh38
Transcripts
NM_002198.2 XM_011543378.2 XM_011543379.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3365817 c.687G>A p.E229E Chr.5: 132485697-132485697 on Build GRCh38
3409729 c.674C>T p.T225I Chr.5: 132485710-132485710 on Build GRCh38
5867257 c.692G>A p.G231E Chr.5: 132485692-132485692 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778994328 Chr.5: 132485649-132485649 on Build GRCh38
rs749154498 Chr.5: 132485648-132485662 on Build GRCh38
rs530764359 Chr.5: 132485749-132485749 on Build GRCh38
rs544839641 Chr.5: 132485620-132485620 on Build GRCh38
rs750734988 Chr.5: 132485768-132485768 on Build GRCh38
rs564717504 Chr.5: 132485741-132485741 on Build GRCh38
rs759283313 Chr.5: 132485591-132485591 on Build GRCh38
rs547088541 Chr.5: 132485734-132485735 on Build GRCh38
rs746967501 Chr.5: 132485729-132485729 on Build GRCh38
rs10068129 Chr.5: 132485554-132485554 on Build GRCh38
rs768507040 Chr.5: 132485720-132485721 on Build GRCh38
rs757604645 Chr.5: 132485606-132485606 on Build GRCh38
rs775957119 Chr.5: 132485585-132485585 on Build GRCh38
rs375551867 Chr.5: 132485628-132485628 on Build GRCh38
rs767811304 Chr.5: 132485619-132485619 on Build GRCh38
rs773104978 Chr.5: 132485696-132485696 on Build GRCh38
rs749303285 Chr.5: 132485629-132485629 on Build GRCh38
rs767899455 Chr.5: 132485756-132485756 on Build GRCh38
rs772000390 Chr.5: 132485666-132485666 on Build GRCh38
rs368236878 Chr.5: 132485745-132485745 on Build GRCh38
rs755264134 Chr.5: 132485637-132485637 on Build GRCh38
rs925271879 Chr.5: 132485690-132485690 on Build GRCh38
rs748326725 Chr.5: 132485662-132485662 on Build GRCh38
rs572717408 Chr.5: 132485603-132485603 on Build GRCh38
rs762524402 Chr.5: 132485755-132485755 on Build GRCh38
rs369379107 Chr.5: 132485738-132485739 on Build GRCh38
rs776245057 Chr.5: 132485740-132485740 on Build GRCh38
rs770752737 Chr.5: 132485732-132485732 on Build GRCh38
rs1033096335 Chr.5: 132485568-132485568 on Build GRCh38
rs111857237 Chr.5: 132485596-132485596 on Build GRCh38

View Full Product Details