Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 65197271-65197510 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CILP
Entrez Gene ID
8483
Gene Name
cartilage intermediate layer protein
Gene Aliases
CILP-1, HsT18872
UniGene
Hs.442180
Gene Chromosome Location
Chr.15: 65195999-65211608 on Build GRCh38
Transcripts
NM_003613.3 XM_017022678.1 XM_017022679.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6629261 c.2808C>T p.S936S Chr.15: 65197478-65197478 on Build GRCh38
7224246 c.3009T>C p.A1003A Chr.15: 65197277-65197277 on Build GRCh38
6142511 c.2983G>A p.D995N Chr.15: 65197303-65197303 on Build GRCh38
5363110 c.2819A>T p.D940V Chr.15: 65197467-65197467 on Build GRCh38
4621604 c.2791G>A p.E931K Chr.15: 65197495-65197495 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369696446 Chr.15: 65197375-65197375 on Build GRCh38
rs149741206 Chr.15: 65197292-65197292 on Build GRCh38
rs143163077 Chr.15: 65197305-65197305 on Build GRCh38
rs770457575 Chr.15: 65197415-65197415 on Build GRCh38
rs774012195 Chr.15: 65197362-65197362 on Build GRCh38
rs145652225 Chr.15: 65197294-65197294 on Build GRCh38
rs757160279 Chr.15: 65197363-65197363 on Build GRCh38
rs1001759106 Chr.15: 65197364-65197364 on Build GRCh38
rs375320311 Chr.15: 65197367-65197367 on Build GRCh38
rs764392452 Chr.15: 65197295-65197295 on Build GRCh38
rs777211471 Chr.15: 65197351-65197351 on Build GRCh38
rs765770685 Chr.15: 65197374-65197374 on Build GRCh38
rs751671698 Chr.15: 65197462-65197462 on Build GRCh38
rs371441372 Chr.15: 65197427-65197427 on Build GRCh38
rs531290271 Chr.15: 65197400-65197400 on Build GRCh38
rs149684578 Chr.15: 65197352-65197352 on Build GRCh38
rs747172066 Chr.15: 65197323-65197323 on Build GRCh38
rs769047039 Chr.15: 65197365-65197365 on Build GRCh38
rs148815769 Chr.15: 65197313-65197313 on Build GRCh38
rs759900331 Chr.15: 65197463-65197463 on Build GRCh38
rs748079200 Chr.15: 65197331-65197331 on Build GRCh38
rs770958657 Chr.15: 65197340-65197340 on Build GRCh38
rs763140331 Chr.15: 65197380-65197380 on Build GRCh38
rs998424675 Chr.15: 65197296-65197296 on Build GRCh38
rs771888858 Chr.15: 65197438-65197438 on Build GRCh38
rs1055174406 Chr.15: 65197478-65197478 on Build GRCh38
rs762283998 Chr.15: 65197368-65197368 on Build GRCh38
rs867559583 Chr.15: 65197451-65197451 on Build GRCh38
rs752800287 Chr.15: 65197465-65197465 on Build GRCh38
rs755224607 Chr.15: 65197398-65197398 on Build GRCh38
rs373252930 Chr.15: 65197353-65197353 on Build GRCh38
rs377336076 Chr.15: 65197444-65197444 on Build GRCh38
rs778362259 Chr.15: 65197422-65197422 on Build GRCh38
rs751865393 Chr.15: 65197388-65197388 on Build GRCh38
rs754332108 Chr.15: 65197297-65197297 on Build GRCh38
rs771698636 Chr.15: 65197440-65197440 on Build GRCh38
rs755956176 Chr.15: 65197475-65197475 on Build GRCh38
rs374644355 Chr.15: 65197314-65197314 on Build GRCh38
rs763516453 Chr.15: 65197453-65197453 on Build GRCh38
rs777389416 Chr.15: 65197479-65197479 on Build GRCh38
rs559616243 Chr.15: 65197324-65197324 on Build GRCh38
rs770367551 Chr.15: 65197354-65197354 on Build GRCh38
rs750503262 Chr.15: 65197306-65197306 on Build GRCh38
rs766609086 Chr.15: 65197456-65197456 on Build GRCh38
rs745719027 Chr.15: 65197343-65197344 on Build GRCh38
rs369015415 Chr.15: 65197321-65197321 on Build GRCh38
rs144790648 Chr.15: 65197407-65197407 on Build GRCh38
rs755794574 Chr.15: 65197402-65197402 on Build GRCh38
rs781096900 Chr.15: 65197326-65197326 on Build GRCh38
rs2679117 Chr.15: 65197350-65197350 on Build GRCh38
rs907510533 Chr.15: 65197328-65197328 on Build GRCh38
rs572631157 Chr.15: 65197301-65197301 on Build GRCh38
rs370254537 Chr.15: 65197464-65197464 on Build GRCh38
rs752528671 Chr.15: 65197401-65197401 on Build GRCh38
rs777776022 Chr.15: 65197405-65197405 on Build GRCh38
rs760366605 Chr.15: 65197442-65197442 on Build GRCh38
rs150946463 Chr.15: 65197381-65197381 on Build GRCh38
rs753812699 Chr.15: 65197486-65197486 on Build GRCh38

View Full Product Details