Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 97229070-97229343 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP19
Entrez Gene ID
84986
Gene Name
Rho GTPase activating protein 19
Gene Aliases
-
UniGene
Hs.80305
Gene Chromosome Location
Chr.10: 97222173-97292673 on Build GRCh38
Transcripts
NM_001204300.1 NM_001256423.1 NM_032900.5 NR_037909.1

Gene Symbol
ARHGAP19-SLIT1
Entrez Gene ID
100533184
Gene Name
ARHGAP19-SLIT1 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.80305
Gene Chromosome Location
Chr.10: 97153042-97292673 on Build GRCh38
Transcripts
NM_001204300.1 NM_001256423.1 NM_032900.5 NR_037909.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5950564 c.1387+10G>A p.? Chr.10: 97229137-97229137 on Build GRCh38
5673057 c.1350A>T p.R450S Chr.10: 97229184-97229184 on Build GRCh38
3770716 c.1463A>C p.E488A Chr.10: 97229158-97229158 on Build GRCh38
5673056 c.1437A>T p.R479S Chr.10: 97229184-97229184 on Build GRCh38
6250406 c.1419C>A p.V473V Chr.10: 97229202-97229202 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376418052 Chr.10: 97229183-97229183 on Build GRCh38
rs538002721 Chr.10: 97229196-97229196 on Build GRCh38
rs370179433 Chr.10: 97229199-97229199 on Build GRCh38
rs781645924 Chr.10: 97229124-97229124 on Build GRCh38
rs767306060 Chr.10: 97229160-97229160 on Build GRCh38
rs370679610 Chr.10: 97229137-97229137 on Build GRCh38
rs201909455 Chr.10: 97229269-97229269 on Build GRCh38
rs114848942 Chr.10: 97229267-97229267 on Build GRCh38
rs577019086 Chr.10: 97229109-97229109 on Build GRCh38
rs778403116 Chr.10: 97229204-97229204 on Build GRCh38
rs764635769 Chr.10: 97229157-97229158 on Build GRCh38
rs747528775 Chr.10: 97229117-97229117 on Build GRCh38
rs752655720 Chr.10: 97229165-97229165 on Build GRCh38
rs759548911 Chr.10: 97229152-97229152 on Build GRCh38
rs200672097 Chr.10: 97229126-97229126 on Build GRCh38
rs370231559 Chr.10: 97229240-97229240 on Build GRCh38
rs972650198 Chr.10: 97229236-97229236 on Build GRCh38
rs910944835 Chr.10: 97229262-97229262 on Build GRCh38
rs57251887 Chr.10: 97229242-97229242 on Build GRCh38
rs866437571 Chr.10: 97229152-97229153 on Build GRCh38
rs749899616 Chr.10: 97229206-97229206 on Build GRCh38
rs774505706 Chr.10: 97229144-97229144 on Build GRCh38
rs781601284 Chr.10: 97229231-97229231 on Build GRCh38
rs984001520 Chr.10: 97229257-97229257 on Build GRCh38
rs758732488 Chr.10: 97229153-97229154 on Build GRCh38
rs368136378 Chr.10: 97229265-97229265 on Build GRCh38
rs768982242 Chr.10: 97229118-97229118 on Build GRCh38
rs748681111 Chr.10: 97229235-97229235 on Build GRCh38
rs748416097 Chr.10: 97229125-97229125 on Build GRCh38
rs186616763 Chr.10: 97229310-97229310 on Build GRCh38
rs756387431 Chr.10: 97229243-97229243 on Build GRCh38
rs1000260952 Chr.10: 97229140-97229140 on Build GRCh38
rs756829731 Chr.10: 97229100-97229100 on Build GRCh38
rs761173048 Chr.10: 97229186-97229188 on Build GRCh38
rs756846192 Chr.10: 97229200-97229200 on Build GRCh38
rs748910200 Chr.10: 97229098-97229098 on Build GRCh38
rs1014650543 Chr.10: 97229191-97229191 on Build GRCh38
rs77340648 Chr.10: 97229209-97229209 on Build GRCh38
rs904700417 Chr.10: 97229139-97229139 on Build GRCh38
rs901431027 Chr.10: 97229106-97229106 on Build GRCh38
rs866532664 Chr.10: 97229179-97229179 on Build GRCh38
rs755533213 Chr.10: 97229227-97229227 on Build GRCh38
rs773633122 Chr.10: 97229131-97229131 on Build GRCh38
rs981551325 Chr.10: 97229205-97229205 on Build GRCh38
rs201485424 Chr.10: 97229130-97229130 on Build GRCh38
rs757333169 Chr.10: 97229190-97229190 on Build GRCh38

View Full Product Details