Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 181715227-181715500 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNA1E
Entrez Gene ID
777
Gene Name
calcium voltage-gated channel subunit alpha1 E
Gene Aliases
BII, CACH6, CACNL1A6, Cav2.3, gm139
UniGene
Hs.437444
Gene Chromosome Location
Chr.1: 181317712-181808084 on Build GRCh38
Transcripts
NM_000721.3 NM_001205293.1 NM_001205294.1 XM_017002243.1 XM_017002244.1 XM_017002245.1 XM_017002246.1 XM_017002247.1 XM_017002248.1 XM_017002249.1 XM_017002250.1 XM_017002251.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6996368 c.1182G>T p.M394I Chr.1: 181715348-181715348 on Build GRCh38
5227694 c.1039G>T p.A347S Chr.1: 181715352-181715352 on Build GRCh38
4226871 c.13G>T p.E5* Chr.1: 181715358-181715358 on Build GRCh38
4025760 c.1216G>A p.A406T Chr.1: 181715382-181715382 on Build GRCh38
3479535 c.6C>T p.L2L Chr.1: 181715351-181715351 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767087787 Chr.1: 181715402-181715402 on Build GRCh38
rs778550852 Chr.1: 181715302-181715302 on Build GRCh38
rs765802112 Chr.1: 181715418-181715418 on Build GRCh38
rs777265667 Chr.1: 181715352-181715352 on Build GRCh38
rs748406317 Chr.1: 181715306-181715306 on Build GRCh38
rs370112728 Chr.1: 181715443-181715443 on Build GRCh38
rs778007393 Chr.1: 181715334-181715334 on Build GRCh38
rs191210738 Chr.1: 181715374-181715374 on Build GRCh38
rs931390363 Chr.1: 181715249-181715249 on Build GRCh38
rs959798575 Chr.1: 181715467-181715467 on Build GRCh38
rs746749659 Chr.1: 181715405-181715407 on Build GRCh38
rs758655123 Chr.1: 181715319-181715319 on Build GRCh38
rs12407563 Chr.1: 181715469-181715469 on Build GRCh38
rs113071809 Chr.1: 181715278-181715278 on Build GRCh38
rs746325499 Chr.1: 181715368-181715368 on Build GRCh38
rs770239808 Chr.1: 181715370-181715370 on Build GRCh38
rs35265801 Chr.1: 181715381-181715381 on Build GRCh38
rs1010079884 Chr.1: 181715348-181715348 on Build GRCh38
rs367948281 Chr.1: 181715297-181715297 on Build GRCh38
rs1053828086 Chr.1: 181715268-181715268 on Build GRCh38
rs186789666 Chr.1: 181715351-181715351 on Build GRCh38
rs866297372 Chr.1: 181715397-181715397 on Build GRCh38
rs555358370 Chr.1: 181715436-181715436 on Build GRCh38
rs746420654 Chr.1: 181715346-181715346 on Build GRCh38
rs943899420 Chr.1: 181715296-181715296 on Build GRCh38
rs760290208 Chr.1: 181715413-181715413 on Build GRCh38
rs753862668 Chr.1: 181715432-181715432 on Build GRCh38
rs894157921 Chr.1: 181715441-181715441 on Build GRCh38
rs144146925 Chr.1: 181715345-181715345 on Build GRCh38
rs1016062571 Chr.1: 181715447-181715447 on Build GRCh38
rs375416527 Chr.1: 181715442-181715442 on Build GRCh38
rs375021294 Chr.1: 181715292-181715292 on Build GRCh38
rs772721347 Chr.1: 181715411-181715411 on Build GRCh38

View Full Product Details