Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 51653564-51653831 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA10
Entrez Gene ID
56934
Gene Name
carbonic anhydrase 10
Gene Aliases
CA-RPX, CARPX, HUCEP-15
UniGene
Hs.463466
Gene Chromosome Location
Chr.17: 51630313-52160017 on Build GRCh38
Transcripts
NM_001082533.1 NM_001082534.1 NM_020178.4 XM_017024878.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
981405 c.501G>A p.T167T Chr.17: 51653701-51653701 on Build GRCh38
4858922 c.495C>A p.I165I Chr.17: 51653725-51653725 on Build GRCh38
3519521 c.476T>G p.I159S Chr.17: 51653726-51653726 on Build GRCh38
3519522 c.494T>G p.I165S Chr.17: 51653726-51653726 on Build GRCh38
981404 c.506T>A p.V169D Chr.17: 51653696-51653696 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751202380 Chr.17: 51653604-51653604 on Build GRCh38
rs747305523 Chr.17: 51653684-51653684 on Build GRCh38
rs769181729 Chr.17: 51653701-51653701 on Build GRCh38
rs770444593 Chr.17: 51653744-51653744 on Build GRCh38
rs772376578 Chr.17: 51653656-51653656 on Build GRCh38
rs773944128 Chr.17: 51653751-51653751 on Build GRCh38
rs746095046 Chr.17: 51653651-51653651 on Build GRCh38
rs1051602446 Chr.17: 51653601-51653601 on Build GRCh38
rs759219984 Chr.17: 51653766-51653766 on Build GRCh38
rs767209024 Chr.17: 51653783-51653783 on Build GRCh38
rs774875297 Chr.17: 51653755-51653757 on Build GRCh38
rs780407663 Chr.17: 51653667-51653667 on Build GRCh38
rs773770613 Chr.17: 51653652-51653652 on Build GRCh38
rs765969311 Chr.17: 51653600-51653600 on Build GRCh38
rs374627558 Chr.17: 51653628-51653628 on Build GRCh38
rs777074047 Chr.17: 51653702-51653702 on Build GRCh38
rs754771474 Chr.17: 51653608-51653608 on Build GRCh38
rs1039271575 Chr.17: 51653747-51653747 on Build GRCh38
rs754007380 Chr.17: 51653631-51653631 on Build GRCh38
rs868585458 Chr.17: 51653798-51653798 on Build GRCh38
rs565415114 Chr.17: 51653632-51653632 on Build GRCh38
rs756136172 Chr.17: 51653624-51653624 on Build GRCh38
rs762418935 Chr.17: 51653713-51653713 on Build GRCh38
rs752742333 Chr.17: 51653619-51653619 on Build GRCh38
rs371904327 Chr.17: 51653786-51653786 on Build GRCh38
rs768348175 Chr.17: 51653634-51653634 on Build GRCh38
rs915981371 Chr.17: 51653598-51653598 on Build GRCh38
rs114531802 Chr.17: 51653593-51653593 on Build GRCh38
rs1045726415 Chr.17: 51653637-51653637 on Build GRCh38
rs368843762 Chr.17: 51653649-51653649 on Build GRCh38
rs766964950 Chr.17: 51653621-51653621 on Build GRCh38
rs767342083 Chr.17: 51653615-51653615 on Build GRCh38
rs762313239 Chr.17: 51653765-51653766 on Build GRCh38

View Full Product Details