Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 92461134-92461359 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IDE
Entrez Gene ID
3416
Gene Name
insulin degrading enzyme
Gene Aliases
INSULYSIN
UniGene
Hs.500546
Gene Chromosome Location
Chr.10: 92451684-92574484 on Build GRCh38
Transcripts
NM_001165946.1 NM_001322793.1 NM_001322794.1 NM_001322795.1 NM_001322796.1 NM_001322797.1 NM_004969.3 NR_136399.1 XM_017016187.1 XM_017016188.1 XM_017016189.1 XM_017016190.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3441527 c.2794A>T p.T932S Chr.10: 92461220-92461220 on Build GRCh38
4016625 c.2769T>C p.T923T Chr.10: 92461245-92461245 on Build GRCh38
3383087 c.2813A>C p.K938T Chr.10: 92461201-92461201 on Build GRCh38
3441526 c.2817C>T p.F939F Chr.10: 92461197-92461197 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1056592053 Chr.10: 92461323-92461323 on Build GRCh38
rs781508503 Chr.10: 92461203-92461203 on Build GRCh38
rs764635158 Chr.10: 92461265-92461265 on Build GRCh38
rs749319197 Chr.10: 92461209-92461209 on Build GRCh38
rs755423509 Chr.10: 92461201-92461201 on Build GRCh38
rs140729995 Chr.10: 92461246-92461246 on Build GRCh38
rs778588284 Chr.10: 92461193-92461193 on Build GRCh38
rs372503165 Chr.10: 92461295-92461295 on Build GRCh38
rs780368441 Chr.10: 92461289-92461289 on Build GRCh38
rs773290496 Chr.10: 92461208-92461208 on Build GRCh38
rs369930618 Chr.10: 92461286-92461286 on Build GRCh38
rs771089800 Chr.10: 92461235-92461235 on Build GRCh38
rs775413987 Chr.10: 92461243-92461243 on Build GRCh38
rs951087436 Chr.10: 92461175-92461175 on Build GRCh38
rs145220987 Chr.10: 92461207-92461207 on Build GRCh38
rs771090586 Chr.10: 92461301-92461301 on Build GRCh38
rs750134020 Chr.10: 92461270-92461270 on Build GRCh38
rs982413812 Chr.10: 92461177-92461177 on Build GRCh38
rs758044157 Chr.10: 92461278-92461278 on Build GRCh38
rs377425045 Chr.10: 92461170-92461170 on Build GRCh38
rs549856782 Chr.10: 92461236-92461236 on Build GRCh38
rs764057888 Chr.10: 92461255-92461255 on Build GRCh38
rs939431211 Chr.10: 92461314-92461314 on Build GRCh38
rs756746552 Chr.10: 92461262-92461262 on Build GRCh38
rs201354702 Chr.10: 92461261-92461261 on Build GRCh38
rs748718341 Chr.10: 92461205-92461205 on Build GRCh38
rs558948942 Chr.10: 92461298-92461298 on Build GRCh38
rs756865947 Chr.10: 92461186-92461186 on Build GRCh38
rs1027002948 Chr.10: 92461324-92461324 on Build GRCh38
rs974110299 Chr.10: 92461273-92461273 on Build GRCh38
rs1029293674 Chr.10: 92461171-92461171 on Build GRCh38
rs150947060 Chr.10: 92461238-92461238 on Build GRCh38
rs747359695 Chr.10: 92461198-92461198 on Build GRCh38

View Full Product Details