Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27444347-27444620 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFT172
Entrez Gene ID
26160
Gene Name
intraflagellar transport 172
Gene Aliases
BBS20, NPHP17, RP71, SLB, SRTD10, osm-1, wim
UniGene
Hs.127401
Gene Chromosome Location
Chr.2: 27444373-27489819 on Build GRCh38
Transcripts
NM_015662.2 XM_005264254.1 XM_006711986.3 XM_006711987.1 XM_011532757.2 XM_011532759.2 XM_011532760.2 XM_017003790.1 XM_017003791.1 XM_017003793.1 XM_017003794.1 XM_017003795.1 NM_001168364.1

Gene Symbol
KRTCAP3
Entrez Gene ID
200634
Gene Name
keratinocyte associated protein 3
Gene Aliases
KCP3
UniGene
Hs.59509
Gene Chromosome Location
Chr.2: 27442366-27446481 on Build GRCh38
Transcripts
NM_015662.2 XM_005264254.1 XM_006711986.3 XM_006711987.1 XM_011532757.2 XM_011532759.2 XM_011532760.2 XM_017003790.1 XM_017003791.1 XM_017003793.1 XM_017003794.1 XM_017003795.1 NM_001168364.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6026331 c.5180G>A p.C1727Y Chr.2: 27444502-27444502 on Build GRCh38
442747 c.5247G>T p.Q1749H Chr.2: 27444435-27444435 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs148800421 Chr.2: 27444512-27444512 on Build GRCh38
rs779019745 Chr.2: 27444532-27444532 on Build GRCh38
rs755653071 Chr.2: 27444390-27444391 on Build GRCh38
rs769331570 Chr.2: 27444450-27444450 on Build GRCh38
rs779787513 Chr.2: 27444571-27444571 on Build GRCh38
rs372020267 Chr.2: 27444573-27444573 on Build GRCh38
rs772224250 Chr.2: 27444420-27444420 on Build GRCh38
rs768451674 Chr.2: 27444555-27444555 on Build GRCh38
rs776552648 Chr.2: 27444435-27444435 on Build GRCh38
rs879266526 Chr.2: 27444491-27444491 on Build GRCh38
rs761254391 Chr.2: 27444447-27444447 on Build GRCh38
rs985755052 Chr.2: 27444494-27444494 on Build GRCh38
rs779876638 Chr.2: 27444428-27444428 on Build GRCh38
rs375251963 Chr.2: 27444383-27444383 on Build GRCh38
rs746737744 Chr.2: 27444544-27444544 on Build GRCh38
rs946599676 Chr.2: 27444564-27444564 on Build GRCh38
rs753075654 Chr.2: 27444500-27444500 on Build GRCh38
rs768540015 Chr.2: 27444434-27444434 on Build GRCh38
rs746100848 Chr.2: 27444412-27444412 on Build GRCh38
rs778920690 Chr.2: 27444391-27444391 on Build GRCh38
rs764640478 Chr.2: 27444511-27444511 on Build GRCh38
rs4803 Chr.2: 27444430-27444430 on Build GRCh38
rs894890263 Chr.2: 27444418-27444418 on Build GRCh38
rs987906613 Chr.2: 27444527-27444527 on Build GRCh38
rs757772988 Chr.2: 27444528-27444528 on Build GRCh38
rs149614625 Chr.2: 27444503-27444503 on Build GRCh38
rs267599318 Chr.2: 27444457-27444457 on Build GRCh38
rs375519807 Chr.2: 27444453-27444453 on Build GRCh38
rs372154058 Chr.2: 27444541-27444541 on Build GRCh38
rs143671920 Chr.2: 27444495-27444495 on Build GRCh38
rs765993455 Chr.2: 27444496-27444496 on Build GRCh38
rs953161312 Chr.2: 27444446-27444446 on Build GRCh38
rs547218491 Chr.2: 27444587-27444587 on Build GRCh38
rs781020503 Chr.2: 27444569-27444569 on Build GRCh38
rs780170408 Chr.2: 27444543-27444543 on Build GRCh38
rs532172685 Chr.2: 27444586-27444586 on Build GRCh38
rs368863616 Chr.2: 27444533-27444533 on Build GRCh38
rs757428770 Chr.2: 27444382-27444382 on Build GRCh38
rs1054911867 Chr.2: 27444380-27444380 on Build GRCh38

View Full Product Details