Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 159229626-159229897 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FNDC1
Entrez Gene ID
84624
Gene Name
fibronectin type III domain containing 1
Gene Aliases
AGS8, FNDC2, MEL4B3, bA243O10.1, dJ322A24.1
UniGene
Hs.520525
Gene Chromosome Location
Chr.6: 159169397-159272108 on Build GRCh38
Transcripts
NM_032532.2 XM_011536190.2 XM_011536191.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3921003 c.1240C>T p.P414S Chr.6: 159229874-159229874 on Build GRCh38
5524986 c.1230C>T p.S410S Chr.6: 159229864-159229864 on Build GRCh38
5700890 c.1222G>A p.A408T Chr.6: 159229856-159229856 on Build GRCh38
293875 c.1197C>T p.Y399Y Chr.6: 159229831-159229831 on Build GRCh38
4925082 c.1210A>C p.K404Q Chr.6: 159229844-159229844 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374491526 Chr.6: 159229858-159229858 on Build GRCh38
rs116342739 Chr.6: 159229816-159229816 on Build GRCh38
rs1032237076 Chr.6: 159229651-159229651 on Build GRCh38
rs944141093 Chr.6: 159229706-159229706 on Build GRCh38
rs146964795 Chr.6: 159229691-159229691 on Build GRCh38
rs762361935 Chr.6: 159229862-159229862 on Build GRCh38
rs756429306 Chr.6: 159229869-159229869 on Build GRCh38
rs763110426 Chr.6: 159229847-159229847 on Build GRCh38
rs534747334 Chr.6: 159229734-159229734 on Build GRCh38
rs764997490 Chr.6: 159229876-159229876 on Build GRCh38
rs775564641 Chr.6: 159229837-159229837 on Build GRCh38
rs148846766 Chr.6: 159229669-159229669 on Build GRCh38
rs1057019171 Chr.6: 159229813-159229813 on Build GRCh38
rs927236800 Chr.6: 159229747-159229747 on Build GRCh38
rs937582113 Chr.6: 159229776-159229776 on Build GRCh38
rs565844308 Chr.6: 159229724-159229724 on Build GRCh38
rs1017844974 Chr.6: 159229650-159229650 on Build GRCh38
rs1013371748 Chr.6: 159229867-159229867 on Build GRCh38
rs746878793 Chr.6: 159229828-159229828 on Build GRCh38
rs750839843 Chr.6: 159229868-159229868 on Build GRCh38
rs777538821 Chr.6: 159229848-159229848 on Build GRCh38
rs867891743 Chr.6: 159229656-159229656 on Build GRCh38
rs914599574 Chr.6: 159229725-159229725 on Build GRCh38
rs754709121 Chr.6: 159229791-159229791 on Build GRCh38
rs778397856 Chr.6: 159229807-159229807 on Build GRCh38
rs200720425 Chr.6: 159229836-159229836 on Build GRCh38
rs757702933 Chr.6: 159229782-159229782 on Build GRCh38
rs976243496 Chr.6: 159229688-159229688 on Build GRCh38
rs377223835 Chr.6: 159229831-159229831 on Build GRCh38
rs776356434 Chr.6: 159229832-159229832 on Build GRCh38
rs55642955 Chr.6: 159229735-159229735 on Build GRCh38
rs57447691 Chr.6: 159229690-159229690 on Build GRCh38
rs116058921 Chr.6: 159229772-159229772 on Build GRCh38
rs947420595 Chr.6: 159229728-159229728 on Build GRCh38
rs376869929 Chr.6: 159229863-159229863 on Build GRCh38
rs757832266 Chr.6: 159229820-159229820 on Build GRCh38
rs767955169 Chr.6: 159229783-159229783 on Build GRCh38
rs189890238 Chr.6: 159229785-159229785 on Build GRCh38
rs753456171 Chr.6: 159229784-159229784 on Build GRCh38
rs1048661310 Chr.6: 159229766-159229766 on Build GRCh38
rs777699032 Chr.6: 159229826-159229826 on Build GRCh38

View Full Product Details