Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 22209197-22209468 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EBLN1
Entrez Gene ID
340900
Gene Name
endogenous Bornavirus-like nucleoprotein 1
Gene Aliases
EBLN-1
UniGene
Hs.450519
Gene Chromosome Location
Chr.10: 22208814-22209983 on Build GRCh38
Transcripts
NM_001199938.1

SNP Detail

SNP ID Chromosome Location
rs1008306097 Chr.10: 22209272-22209272 on Build GRCh38
rs149078001 Chr.10: 22209419-22209419 on Build GRCh38
rs766493721 Chr.10: 22209358-22209358 on Build GRCh38
rs760723828 Chr.10: 22209354-22209354 on Build GRCh38
rs16922091 Chr.10: 22209330-22209330 on Build GRCh38
rs992517915 Chr.10: 22209351-22209351 on Build GRCh38
rs535338802 Chr.10: 22209237-22209237 on Build GRCh38
rs762309021 Chr.10: 22209414-22209414 on Build GRCh38
rs751918539 Chr.10: 22209219-22209219 on Build GRCh38
rs779897239 Chr.10: 22209263-22209263 on Build GRCh38
rs116468417 Chr.10: 22209366-22209366 on Build GRCh38
rs771897402 Chr.10: 22209220-22209220 on Build GRCh38
rs750554782 Chr.10: 22209250-22209250 on Build GRCh38
rs774978159 Chr.10: 22209410-22209410 on Build GRCh38
rs983321021 Chr.10: 22209338-22209338 on Build GRCh38
rs776306586 Chr.10: 22209384-22209384 on Build GRCh38
rs934604854 Chr.10: 22209382-22209382 on Build GRCh38
rs188558021 Chr.10: 22209370-22209370 on Build GRCh38
rs184227086 Chr.10: 22209283-22209283 on Build GRCh38
rs944562408 Chr.10: 22209439-22209439 on Build GRCh38
rs766070786 Chr.10: 22209280-22209281 on Build GRCh38
rs771732946 Chr.10: 22209288-22209288 on Build GRCh38
rs772561065 Chr.10: 22209418-22209418 on Build GRCh38
rs996880167 Chr.10: 22209307-22209307 on Build GRCh38
rs960523497 Chr.10: 22209350-22209350 on Build GRCh38
rs777378822 Chr.10: 22209294-22209294 on Build GRCh38
rs756197746 Chr.10: 22209260-22209260 on Build GRCh38
rs767809807 Chr.10: 22209239-22209239 on Build GRCh38
rs746537765 Chr.10: 22209321-22209321 on Build GRCh38
rs978670085 Chr.10: 22209336-22209336 on Build GRCh38
rs144989106 Chr.10: 22209375-22209375 on Build GRCh38
rs745332913 Chr.10: 22209387-22209387 on Build GRCh38
rs777465881 Chr.10: 22209377-22209377 on Build GRCh38
rs770466695 Chr.10: 22209326-22209326 on Build GRCh38
rs2243897 Chr.10: 22209280-22209280 on Build GRCh38
rs980325687 Chr.10: 22209398-22209398 on Build GRCh38
rs747896082 Chr.10: 22209284-22209284 on Build GRCh38
rs866871648 Chr.10: 22209317-22209317 on Build GRCh38
rs1015265877 Chr.10: 22209304-22209304 on Build GRCh38
rs775922559 Chr.10: 22209327-22209327 on Build GRCh38
rs572736524 Chr.10: 22209218-22209218 on Build GRCh38
rs769311906 Chr.10: 22209406-22209406 on Build GRCh38
rs562469933 Chr.10: 22209340-22209340 on Build GRCh38
rs754937725 Chr.10: 22209360-22209360 on Build GRCh38
rs746665379 Chr.10: 22209381-22209381 on Build GRCh38
rs753801020 Chr.10: 22209271-22209271 on Build GRCh38
rs547550731 Chr.10: 22209359-22209359 on Build GRCh38
rs757560137 Chr.10: 22209221-22209221 on Build GRCh38

View Full Product Details