Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153962274-153962533 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR6737
Entrez Gene ID
102465441
Gene Name
microRNA 6737
Gene Aliases
hsa-mir-6737
UniGene
Gene Chromosome Location
Chr.1: 153962351-153962420 on Build GRCh38
Transcripts
NR_106795.1 NM_001271957.1 NM_001271958.1 NM_001271959.1 NM_001271960.1 NM_001271961.1 NM_014437.4

Gene Symbol
SLC39A1
Entrez Gene ID
27173
Gene Name
solute carrier family 39 member 1
Gene Aliases
ZIP1, ZIRTL
UniGene
Hs.743291
Gene Chromosome Location
Chr.1: 153959099-153968184 on Build GRCh38
Transcripts
NR_106795.1 NM_001271957.1 NM_001271958.1 NM_001271959.1 NM_001271960.1 NM_001271961.1 NM_014437.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4612294 c.233_234insG p.V79fs*13 Chr.1: 153962304-153962305 on Build GRCh38
896473 c.244G>A p.A82T Chr.1: 153962294-153962294 on Build GRCh38
4022705 c.194G>A p.R65H Chr.1: 153962344-153962344 on Build GRCh38
4481951 c.255C>T p.L85L Chr.1: 153962283-153962283 on Build GRCh38
4727302 c.213A>G p.L71L Chr.1: 153962325-153962325 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs936516295 Chr.1: 153962438-153962438 on Build GRCh38
rs950976494 Chr.1: 153962293-153962293 on Build GRCh38
rs945023675 Chr.1: 153962354-153962354 on Build GRCh38
rs775850060 Chr.1: 153962387-153962387 on Build GRCh38
rs556623713 Chr.1: 153962421-153962421 on Build GRCh38
rs988937601 Chr.1: 153962395-153962395 on Build GRCh38
rs748825997 Chr.1: 153962458-153962458 on Build GRCh38
rs535826560 Chr.1: 153962306-153962306 on Build GRCh38
rs116493308 Chr.1: 153962337-153962337 on Build GRCh38
rs140644728 Chr.1: 153962464-153962464 on Build GRCh38
rs539511149 Chr.1: 153962405-153962405 on Build GRCh38
rs370004083 Chr.1: 153962303-153962303 on Build GRCh38
rs11554761 Chr.1: 153962343-153962343 on Build GRCh38
rs781141959 Chr.1: 153962312-153962312 on Build GRCh38
rs957568654 Chr.1: 153962375-153962375 on Build GRCh38
rs974029102 Chr.1: 153962483-153962483 on Build GRCh38
rs138894603 Chr.1: 153962310-153962310 on Build GRCh38
rs1054943687 Chr.1: 153962441-153962441 on Build GRCh38
rs765410913 Chr.1: 153962374-153962374 on Build GRCh38
rs374344642 Chr.1: 153962465-153962465 on Build GRCh38
rs753893443 Chr.1: 153962304-153962304 on Build GRCh38
rs777346622 Chr.1: 153962440-153962440 on Build GRCh38
rs371731583 Chr.1: 153962487-153962487 on Build GRCh38
rs570195454 Chr.1: 153962313-153962313 on Build GRCh38
rs766338599 Chr.1: 153962412-153962412 on Build GRCh38
rs754983562 Chr.1: 153962422-153962422 on Build GRCh38
rs763102282 Chr.1: 153962304-153962305 on Build GRCh38
rs1033651556 Chr.1: 153962358-153962358 on Build GRCh38
rs374574755 Chr.1: 153962365-153962365 on Build GRCh38
rs770280183 Chr.1: 153962461-153962461 on Build GRCh38
rs781084732 Chr.1: 153962432-153962432 on Build GRCh38
rs574361150 Chr.1: 153962345-153962345 on Build GRCh38
rs760040371 Chr.1: 153962355-153962355 on Build GRCh38
rs200374799 Chr.1: 153962324-153962324 on Build GRCh38
rs762736678 Chr.1: 153962467-153962467 on Build GRCh38
rs201334475 Chr.1: 153962295-153962295 on Build GRCh38
rs541888675 Chr.1: 153962391-153962391 on Build GRCh38
rs757341439 Chr.1: 153962407-153962407 on Build GRCh38
rs755761550 Chr.1: 153962434-153962434 on Build GRCh38
rs140573490 Chr.1: 153962311-153962311 on Build GRCh38
rs764259486 Chr.1: 153962404-153962404 on Build GRCh38
rs771389957 Chr.1: 153962345-153962345 on Build GRCh38
rs775798578 Chr.1: 153962485-153962485 on Build GRCh38
rs61755890 Chr.1: 153962344-153962344 on Build GRCh38
rs765728788 Chr.1: 153962410-153962411 on Build GRCh38

View Full Product Details