Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 21473478-21473721 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RECQL
Entrez Gene ID
5965
Gene Name
RecQ like helicase
Gene Aliases
RECQL1, RecQ1
UniGene
Hs.235069
Gene Chromosome Location
Chr.12: 21468910-21501669 on Build GRCh38
Transcripts
NM_002907.3 NM_032941.2 XM_005253461.3 XM_005253462.4 XM_005253463.3 XM_005253464.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4720943 c.1413T>A p.C471* Chr.12: 21473585-21473585 on Build GRCh38
72405 c.1416C>A p.N472K Chr.12: 21473582-21473582 on Build GRCh38
6764676 c.1435T>C p.C479R Chr.12: 21473563-21473563 on Build GRCh38
6462486 c.1447G>A p.A483T Chr.12: 21473551-21473551 on Build GRCh38
7282578 c.1361G>A p.R454H Chr.12: 21473637-21473637 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759170220 Chr.12: 21473601-21473601 on Build GRCh38
rs767408144 Chr.12: 21473552-21473552 on Build GRCh38
rs750279410 Chr.12: 21473633-21473635 on Build GRCh38
rs199740132 Chr.12: 21473635-21473635 on Build GRCh38
rs903746277 Chr.12: 21473590-21473590 on Build GRCh38
rs1007904090 Chr.12: 21473628-21473628 on Build GRCh38
rs201851180 Chr.12: 21473562-21473562 on Build GRCh38
rs373470216 Chr.12: 21473508-21473508 on Build GRCh38
rs778148225 Chr.12: 21473634-21473634 on Build GRCh38
rs761292621 Chr.12: 21473667-21473667 on Build GRCh38
rs754146708 Chr.12: 21473576-21473576 on Build GRCh38
rs772188113 Chr.12: 21473624-21473624 on Build GRCh38
rs762542665 Chr.12: 21473507-21473507 on Build GRCh38
rs761361397 Chr.12: 21473523-21473523 on Build GRCh38
rs776939203 Chr.12: 21473637-21473637 on Build GRCh38
rs142226004 Chr.12: 21473573-21473573 on Build GRCh38
rs772961527 Chr.12: 21473537-21473539 on Build GRCh38
rs779257050 Chr.12: 21473614-21473614 on Build GRCh38
rs762926806 Chr.12: 21473552-21473554 on Build GRCh38
rs751105885 Chr.12: 21473514-21473514 on Build GRCh38
rs753875849 Chr.12: 21473521-21473521 on Build GRCh38
rs1044095789 Chr.12: 21473666-21473666 on Build GRCh38
rs755020112 Chr.12: 21473649-21473650 on Build GRCh38
rs773050438 Chr.12: 21473683-21473683 on Build GRCh38
rs766294304 Chr.12: 21473616-21473618 on Build GRCh38
rs370420341 Chr.12: 21473649-21473649 on Build GRCh38
rs1055710310 Chr.12: 21473623-21473623 on Build GRCh38
rs376377526 Chr.12: 21473638-21473638 on Build GRCh38
rs765049715 Chr.12: 21473525-21473526 on Build GRCh38
rs766998762 Chr.12: 21473678-21473678 on Build GRCh38
rs201573409 Chr.12: 21473616-21473616 on Build GRCh38
rs892257635 Chr.12: 21473511-21473511 on Build GRCh38
rs200257717 Chr.12: 21473548-21473548 on Build GRCh38
rs200259739 Chr.12: 21473572-21473572 on Build GRCh38
rs761479946 Chr.12: 21473506-21473507 on Build GRCh38
rs768343455 Chr.12: 21473660-21473660 on Build GRCh38
rs754953232 Chr.12: 21473529-21473529 on Build GRCh38
rs755294309 Chr.12: 21473582-21473582 on Build GRCh38

View Full Product Details