Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 103414623-103414878 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF13
Entrez Gene ID
25879
Gene Name
DDB1 and CUL4 associated factor 13
Gene Aliases
GM83, HSPC064, WDSOF1
UniGene
Hs.532265
Gene Chromosome Location
Chr.8: 103414714-103443453 on Build GRCh38
Transcripts
NM_015420.6 NM_030780.4 NR_102337.1 NR_102338.1 XM_017013877.1

Gene Symbol
SLC25A32
Entrez Gene ID
81034
Gene Name
solute carrier family 25 member 32
Gene Aliases
MFT, MFTC, RREI
UniGene
Hs.532265
Gene Chromosome Location
Chr.8: 103398638-103415335 on Build GRCh38
Transcripts
NM_015420.6 NM_030780.4 NR_102337.1 NR_102338.1 XM_017013877.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
126222 c.130G>T p.D44Y Chr.8: 103414808-103414808 on Build GRCh38
4918685 c.97G>A p.G33S Chr.8: 103414841-103414841 on Build GRCh38
6193758 c.70G>C p.E24Q Chr.8: 103414868-103414868 on Build GRCh38
5714488 c.106T>C p.S36P Chr.8: 103414832-103414832 on Build GRCh38
1226124 c.124C>T p.P42S Chr.8: 103414814-103414814 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs549541582 Chr.8: 103414723-103414723 on Build GRCh38
rs764578714 Chr.8: 103414677-103414678 on Build GRCh38
rs535047258 Chr.8: 103414804-103414804 on Build GRCh38
rs945834133 Chr.8: 103414706-103414706 on Build GRCh38
rs760727387 Chr.8: 103414837-103414838 on Build GRCh38
rs565852534 Chr.8: 103414826-103414826 on Build GRCh38
rs114156661 Chr.8: 103414682-103414682 on Build GRCh38
rs376551963 Chr.8: 103414803-103414803 on Build GRCh38
rs369791895 Chr.8: 103414820-103414820 on Build GRCh38
rs5893653 Chr.8: 103414680-103414680 on Build GRCh38
rs774063632 Chr.8: 103414705-103414705 on Build GRCh38
rs777686799 Chr.8: 103414827-103414827 on Build GRCh38
rs561438030 Chr.8: 103414650-103414650 on Build GRCh38
rs769677268 Chr.8: 103414771-103414771 on Build GRCh38
rs190381842 Chr.8: 103414663-103414663 on Build GRCh38
rs768604116 Chr.8: 103414762-103414762 on Build GRCh38
rs1028800984 Chr.8: 103414652-103414652 on Build GRCh38
rs779454169 Chr.8: 103414737-103414737 on Build GRCh38
rs760874486 Chr.8: 103414680-103414682 on Build GRCh38
rs1037549926 Chr.8: 103414800-103414800 on Build GRCh38
rs766681889 Chr.8: 103414808-103414808 on Build GRCh38
rs781274066 Chr.8: 103414769-103414769 on Build GRCh38
rs749105713 Chr.8: 103414740-103414740 on Build GRCh38
rs898571659 Chr.8: 103414818-103414818 on Build GRCh38
rs369006861 Chr.8: 103414752-103414752 on Build GRCh38
rs533638821 Chr.8: 103414686-103414686 on Build GRCh38
rs386728404 Chr.8: 103414678-103414682 on Build GRCh38
rs748078750 Chr.8: 103414770-103414770 on Build GRCh38
rs775026329 Chr.8: 103414794-103414794 on Build GRCh38
rs878927639 Chr.8: 103414802-103414802 on Build GRCh38
rs916091653 Chr.8: 103414725-103414725 on Build GRCh38
rs568680947 Chr.8: 103414690-103414691 on Build GRCh38
rs777488699 Chr.8: 103414678-103414678 on Build GRCh38
rs776995991 Chr.8: 103414806-103414806 on Build GRCh38
rs531125360 Chr.8: 103414713-103414713 on Build GRCh38
rs528889896 Chr.8: 103414658-103414658 on Build GRCh38
rs768543989 Chr.8: 103414748-103414749 on Build GRCh38
rs367610017 Chr.8: 103414680-103414680 on Build GRCh38
rs567748395 Chr.8: 103414795-103414795 on Build GRCh38
rs373956646 Chr.8: 103414836-103414836 on Build GRCh38
rs201906277 Chr.8: 103414765-103414765 on Build GRCh38
rs539672574 Chr.8: 103414835-103414835 on Build GRCh38
rs10709873 Chr.8: 103414677-103414677 on Build GRCh38
rs770687702 Chr.8: 103414778-103414778 on Build GRCh38
rs964151286 Chr.8: 103414643-103414643 on Build GRCh38
rs5025988 Chr.8: 103414678-103414678 on Build GRCh38
rs149273860 Chr.8: 103414810-103414810 on Build GRCh38
rs765817450 Chr.8: 103414796-103414796 on Build GRCh38
rs938585948 Chr.8: 103414729-103414729 on Build GRCh38
rs551694914 Chr.8: 103414688-103414688 on Build GRCh38
rs202211338 Chr.8: 103414676-103414677 on Build GRCh38
rs766591219 Chr.8: 103414680-103414681 on Build GRCh38
rs759172485 Chr.8: 103414785-103414785 on Build GRCh38
rs139201908 Chr.8: 103414678-103414679 on Build GRCh38
rs762711361 Chr.8: 103414777-103414777 on Build GRCh38
rs774282121 Chr.8: 103414781-103414782 on Build GRCh38
rs5893654 Chr.8: 103414682-103414682 on Build GRCh38
rs941806000 Chr.8: 103414788-103414788 on Build GRCh38
rs769458251 Chr.8: 103414679-103414680 on Build GRCh38
rs138554712 Chr.8: 103414783-103414783 on Build GRCh38
rs781158650 Chr.8: 103414821-103414821 on Build GRCh38
rs759116286 Chr.8: 103414760-103414760 on Build GRCh38
rs144470699 Chr.8: 103414823-103414823 on Build GRCh38
rs750767608 Chr.8: 103414751-103414751 on Build GRCh38
rs774888842 Chr.8: 103414717-103414717 on Build GRCh38
rs367940278 Chr.8: 103414682-103414683 on Build GRCh38
rs891154692 Chr.8: 103414832-103414832 on Build GRCh38
rs773008806 Chr.8: 103414775-103414775 on Build GRCh38
rs570188831 Chr.8: 103414704-103414704 on Build GRCh38
rs766944140 Chr.8: 103414790-103414790 on Build GRCh38

View Full Product Details