Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 95087414-95087639 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRPS5
Entrez Gene ID
64969
Gene Name
mitochondrial ribosomal protein S5
Gene Aliases
MRP-S5, S5mt
UniGene
Hs.655259
Gene Chromosome Location
Chr.2: 95085391-95122027 on Build GRCh38
Transcripts
NM_001321995.1 NM_001321996.1 NM_001321997.1 NM_031902.4 NR_136152.1 NR_136153.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4401323 c.1165G>T p.G389W Chr.2: 95087485-95087485 on Build GRCh38
312944 c.1163G>T p.R388L Chr.2: 95087487-95087487 on Build GRCh38
143570 c.1121_1122GG>AA p.R374Q Chr.2: 95087528-95087529 on Build GRCh38
5140006 c.1163G>C p.R388P Chr.2: 95087487-95087487 on Build GRCh38
81430 c.1221C>T p.D407D Chr.2: 95087429-95087429 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763102361 Chr.2: 95087448-95087448 on Build GRCh38
rs201344145 Chr.2: 95087489-95087489 on Build GRCh38
rs369803780 Chr.2: 95087487-95087487 on Build GRCh38
rs562769679 Chr.2: 95087542-95087542 on Build GRCh38
rs937387408 Chr.2: 95087566-95087566 on Build GRCh38
rs143922359 Chr.2: 95087563-95087563 on Build GRCh38
rs547323684 Chr.2: 95087529-95087529 on Build GRCh38
rs148386913 Chr.2: 95087495-95087495 on Build GRCh38
rs546161255 Chr.2: 95087498-95087498 on Build GRCh38
rs551219520 Chr.2: 95087455-95087455 on Build GRCh38
rs761105808 Chr.2: 95087521-95087521 on Build GRCh38
rs201626536 Chr.2: 95087446-95087446 on Build GRCh38
rs767977505 Chr.2: 95087469-95087469 on Build GRCh38
rs373301376 Chr.2: 95087506-95087506 on Build GRCh38
rs554740592 Chr.2: 95087470-95087470 on Build GRCh38
rs1055811630 Chr.2: 95087604-95087604 on Build GRCh38
rs770115321 Chr.2: 95087440-95087440 on Build GRCh38
rs149040333 Chr.2: 95087494-95087494 on Build GRCh38
rs189335343 Chr.2: 95087449-95087449 on Build GRCh38
rs376665122 Chr.2: 95087459-95087459 on Build GRCh38
rs150457221 Chr.2: 95087497-95087497 on Build GRCh38
rs200747313 Chr.2: 95087493-95087493 on Build GRCh38
rs908844762 Chr.2: 95087481-95087481 on Build GRCh38
rs749418808 Chr.2: 95087496-95087496 on Build GRCh38
rs565531126 Chr.2: 95087571-95087571 on Build GRCh38
rs754559313 Chr.2: 95087488-95087488 on Build GRCh38
rs199604207 Chr.2: 95087486-95087486 on Build GRCh38
rs867288022 Chr.2: 95087545-95087545 on Build GRCh38
rs764963431 Chr.2: 95087530-95087530 on Build GRCh38
rs373403826 Chr.2: 95087441-95087441 on Build GRCh38
rs762265036 Chr.2: 95087461-95087461 on Build GRCh38
rs377726287 Chr.2: 95087525-95087525 on Build GRCh38
rs773586492 Chr.2: 95087512-95087512 on Build GRCh38
rs745686458 Chr.2: 95087592-95087592 on Build GRCh38
rs746758795 Chr.2: 95087439-95087439 on Build GRCh38
rs760932309 Chr.2: 95087471-95087471 on Build GRCh38

View Full Product Details